Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153981971-153982481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB13
Entrez Gene ID
5872
Gene Name
RAB13, member RAS oncogene family
Gene Aliases
GIG4
UniGene
Hs.151536
Gene Chromosome Location
Chr.1: 153981617-153990644 on Build GRCh38
Transcripts
NM_001272038.1 NM_002870.3 NR_073553.1 XM_017001959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6761259 c.527G>A p.R176Q Chr.1: 153982398-153982398 on Build GRCh38
4553159 c.582G>A p.K194K Chr.1: 153982129-153982129 on Build GRCh38
4994028 c.552T>A p.P184P Chr.1: 153982159-153982159 on Build GRCh38
4576298 c.585_587delGAA p.K195delK Chr.1: 153982124-153982126 on Build GRCh38
1723267 c.488delG p.S163fs*27 Chr.1: 153982437-153982437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372691843 Chr.1: 153982333-153982336 on Build GRCh38
rs370358416 Chr.1: 153982288-153982288 on Build GRCh38
rs376694573 Chr.1: 153982255-153982255 on Build GRCh38
rs188759070 Chr.1: 153982286-153982286 on Build GRCh38
rs777438879 Chr.1: 153982440-153982440 on Build GRCh38
rs770721850 Chr.1: 153982080-153982080 on Build GRCh38
rs765598996 Chr.1: 153982134-153982134 on Build GRCh38
rs907967118 Chr.1: 153982246-153982246 on Build GRCh38
rs1040359354 Chr.1: 153982273-153982273 on Build GRCh38
rs747431635 Chr.1: 153982305-153982306 on Build GRCh38
rs763178325 Chr.1: 153982048-153982048 on Build GRCh38
rs376518466 Chr.1: 153982187-153982187 on Build GRCh38
rs373352018 Chr.1: 153982213-153982213 on Build GRCh38
rs948526029 Chr.1: 153982331-153982336 on Build GRCh38
rs991023742 Chr.1: 153982264-153982264 on Build GRCh38
rs371012129 Chr.1: 153982425-153982425 on Build GRCh38
rs763522490 Chr.1: 153982199-153982199 on Build GRCh38
rs746688772 Chr.1: 153982180-153982180 on Build GRCh38
rs2230324 Chr.1: 153982170-153982170 on Build GRCh38
rs56791628 Chr.1: 153982445-153982445 on Build GRCh38
rs768905424 Chr.1: 153982069-153982069 on Build GRCh38
rs777882041 Chr.1: 153982224-153982224 on Build GRCh38
rs144566464 Chr.1: 153982139-153982139 on Build GRCh38
rs944725884 Chr.1: 153982074-153982074 on Build GRCh38
rs920728984 Chr.1: 153982002-153982002 on Build GRCh38
rs199992836 Chr.1: 153982352-153982352 on Build GRCh38
rs755835667 Chr.1: 153982340-153982341 on Build GRCh38
rs550367961 Chr.1: 153982303-153982310 on Build GRCh38
rs767311920 Chr.1: 153982211-153982214 on Build GRCh38
rs778778077 Chr.1: 153982175-153982175 on Build GRCh38
rs767907083 Chr.1: 153982400-153982400 on Build GRCh38
rs771845750 Chr.1: 153982094-153982094 on Build GRCh38
rs748737518 Chr.1: 153982451-153982451 on Build GRCh38
rs202131118 Chr.1: 153982426-153982426 on Build GRCh38
rs751871439 Chr.1: 153982155-153982155 on Build GRCh38
rs371625659 Chr.1: 153982303-153982304 on Build GRCh38
rs920847085 Chr.1: 153982233-153982233 on Build GRCh38
rs756322497 Chr.1: 153982412-153982412 on Build GRCh38
rs761296585 Chr.1: 153982189-153982189 on Build GRCh38
rs767565680 Chr.1: 153982212-153982212 on Build GRCh38
rs12071941 Chr.1: 153982340-153982340 on Build GRCh38
rs553808166 Chr.1: 153982428-153982428 on Build GRCh38
rs1045425556 Chr.1: 153982061-153982061 on Build GRCh38
rs374582048 Chr.1: 153982073-153982073 on Build GRCh38
rs766934698 Chr.1: 153982384-153982384 on Build GRCh38
rs750937101 Chr.1: 153982136-153982136 on Build GRCh38
rs370009244 Chr.1: 153982049-153982049 on Build GRCh38
rs747786017 Chr.1: 153982433-153982433 on Build GRCh38
rs769225876 Chr.1: 153982362-153982362 on Build GRCh38
rs1036918492 Chr.1: 153982392-153982392 on Build GRCh38
rs553304345 Chr.1: 153982283-153982283 on Build GRCh38
rs56853500 Chr.1: 153982332-153982335 on Build GRCh38
rs762397229 Chr.1: 153982114-153982114 on Build GRCh38
rs533820369 Chr.1: 153982341-153982341 on Build GRCh38
rs756202383 Chr.1: 153982222-153982222 on Build GRCh38
rs386635618 Chr.1: 153982198-153982199 on Build GRCh38
rs752731240 Chr.1: 153982341-153982342 on Build GRCh38
rs759880514 Chr.1: 153982399-153982399 on Build GRCh38
rs113430755 Chr.1: 153982336-153982336 on Build GRCh38
rs763627838 Chr.1: 153982098-153982098 on Build GRCh38
rs759182899 Chr.1: 153982082-153982082 on Build GRCh38
rs867817720 Chr.1: 153982302-153982302 on Build GRCh38
rs930748656 Chr.1: 153982033-153982033 on Build GRCh38
rs768136455 Chr.1: 153982182-153982182 on Build GRCh38
rs560548741 Chr.1: 153982160-153982160 on Build GRCh38
rs769439687 Chr.1: 153982436-153982436 on Build GRCh38
rs563763984 Chr.1: 153982304-153982305 on Build GRCh38
rs200514734 Chr.1: 153982205-153982205 on Build GRCh38
rs547812764 Chr.1: 153982334-153982334 on Build GRCh38
rs71584158 Chr.1: 153982303-153982306 on Build GRCh38
rs556345789 Chr.1: 153982424-153982424 on Build GRCh38
rs573012181 Chr.1: 153982449-153982449 on Build GRCh38
rs143645497 Chr.1: 153982157-153982157 on Build GRCh38
rs530111525 Chr.1: 153982303-153982308 on Build GRCh38
rs753241673 Chr.1: 153982102-153982102 on Build GRCh38
rs201338105 Chr.1: 153982344-153982344 on Build GRCh38
rs771898668 Chr.1: 153982177-153982177 on Build GRCh38
rs1012398410 Chr.1: 153982064-153982064 on Build GRCh38
rs1049574987 Chr.1: 153982410-153982410 on Build GRCh38
rs966741898 Chr.1: 153982429-153982429 on Build GRCh38
rs1054210810 Chr.1: 153981997-153981997 on Build GRCh38
rs958400512 Chr.1: 153982393-153982393 on Build GRCh38
rs201485878 Chr.1: 153982190-153982190 on Build GRCh38
rs774857336 Chr.1: 153982398-153982398 on Build GRCh38
rs772806404 Chr.1: 153982365-153982365 on Build GRCh38
rs948471635 Chr.1: 153982047-153982047 on Build GRCh38
rs930843045 Chr.1: 153982241-153982241 on Build GRCh38
rs541731981 Chr.1: 153982081-153982081 on Build GRCh38
rs764018486 Chr.1: 153982388-153982389 on Build GRCh38
rs983618665 Chr.1: 153982244-153982244 on Build GRCh38
rs531680525 Chr.1: 153982207-153982207 on Build GRCh38
rs763575836 Chr.1: 153982383-153982383 on Build GRCh38
rs557220660 Chr.1: 153982062-153982062 on Build GRCh38
rs780542973 Chr.1: 153982151-153982151 on Build GRCh38
rs775229993 Chr.1: 153982179-153982179 on Build GRCh38
rs200374379 Chr.1: 153982171-153982171 on Build GRCh38
rs754046939 Chr.1: 153982439-153982440 on Build GRCh38
rs112589571 Chr.1: 153982279-153982279 on Build GRCh38
rs572146621 Chr.1: 153982124-153982126 on Build GRCh38
rs371708064 Chr.1: 153982313-153982340 on Build GRCh38
rs760293906 Chr.1: 153982095-153982095 on Build GRCh38
rs747220822 Chr.1: 153982056-153982056 on Build GRCh38
rs61805527 Chr.1: 153982332-153982332 on Build GRCh38
rs746741597 Chr.1: 153982350-153982350 on Build GRCh38
rs537894434 Chr.1: 153982353-153982353 on Build GRCh38
rs751993794 Chr.1: 153982247-153982247 on Build GRCh38
rs754779874 Chr.1: 153982427-153982427 on Build GRCh38
rs574680321 Chr.1: 153982071-153982071 on Build GRCh38
rs894261075 Chr.1: 153982001-153982001 on Build GRCh38
rs10908578 Chr.1: 153982198-153982198 on Build GRCh38
rs766879641 Chr.1: 153982203-153982203 on Build GRCh38
rs779840771 Chr.1: 153982346-153982346 on Build GRCh38
rs752908416 Chr.1: 153982404-153982404 on Build GRCh38

View Full Product Details