Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153982254-153982770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB13
Entrez Gene ID
5872
Gene Name
RAB13, member RAS oncogene family
Gene Aliases
GIG4
UniGene
Hs.151536
Gene Chromosome Location
Chr.1: 153981617-153990644 on Build GRCh38
Transcripts
NM_001272038.1 NM_002870.3 NR_073553.1 XM_017001959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6761259 c.527G>A p.R176Q Chr.1: 153982398-153982398 on Build GRCh38
4022710 c.466A>G p.M156V Chr.1: 153982549-153982549 on Build GRCh38
6761260 c.378G>T p.E126D Chr.1: 153982755-153982755 on Build GRCh38
4022711 c.444C>T p.F148F Chr.1: 153982571-153982571 on Build GRCh38
6761261 c.481-2A>T p.? Chr.1: 153982446-153982446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049605926 Chr.1: 153982705-153982705 on Build GRCh38
rs188759070 Chr.1: 153982286-153982286 on Build GRCh38
rs370358416 Chr.1: 153982288-153982288 on Build GRCh38
rs372691843 Chr.1: 153982333-153982336 on Build GRCh38
rs777438879 Chr.1: 153982440-153982440 on Build GRCh38
rs200896887 Chr.1: 153982723-153982723 on Build GRCh38
rs747431635 Chr.1: 153982305-153982306 on Build GRCh38
rs756924739 Chr.1: 153982714-153982714 on Build GRCh38
rs754055347 Chr.1: 153982511-153982511 on Build GRCh38
rs948526029 Chr.1: 153982331-153982336 on Build GRCh38
rs145912655 Chr.1: 153982578-153982578 on Build GRCh38
rs371012129 Chr.1: 153982425-153982425 on Build GRCh38
rs751539122 Chr.1: 153982518-153982518 on Build GRCh38
rs760063487 Chr.1: 153982468-153982468 on Build GRCh38
rs56791628 Chr.1: 153982445-153982445 on Build GRCh38
rs545276571 Chr.1: 153982640-153982640 on Build GRCh38
rs775053648 Chr.1: 153982464-153982464 on Build GRCh38
rs780868590 Chr.1: 153982550-153982550 on Build GRCh38
rs931260449 Chr.1: 153982688-153982688 on Build GRCh38
rs199992836 Chr.1: 153982352-153982352 on Build GRCh38
rs776890093 Chr.1: 153982624-153982624 on Build GRCh38
rs755835667 Chr.1: 153982340-153982341 on Build GRCh38
rs894218542 Chr.1: 153982627-153982627 on Build GRCh38
rs550367961 Chr.1: 153982303-153982310 on Build GRCh38
rs748737518 Chr.1: 153982451-153982451 on Build GRCh38
rs767907083 Chr.1: 153982400-153982400 on Build GRCh38
rs377035618 Chr.1: 153982710-153982710 on Build GRCh38
rs754903283 Chr.1: 153982519-153982519 on Build GRCh38
rs371625659 Chr.1: 153982303-153982304 on Build GRCh38
rs202131118 Chr.1: 153982426-153982426 on Build GRCh38
rs1054366864 Chr.1: 153982523-153982523 on Build GRCh38
rs756322497 Chr.1: 153982412-153982412 on Build GRCh38
rs12071941 Chr.1: 153982340-153982340 on Build GRCh38
rs188353397 Chr.1: 153982684-153982684 on Build GRCh38
rs553808166 Chr.1: 153982428-153982428 on Build GRCh38
rs772575766 Chr.1: 153982605-153982605 on Build GRCh38
rs367690717 Chr.1: 153982594-153982594 on Build GRCh38
rs916766598 Chr.1: 153982515-153982515 on Build GRCh38
rs766934698 Chr.1: 153982384-153982384 on Build GRCh38
rs547968945 Chr.1: 153982711-153982711 on Build GRCh38
rs747786017 Chr.1: 153982433-153982433 on Build GRCh38
rs1036918492 Chr.1: 153982392-153982392 on Build GRCh38
rs769225876 Chr.1: 153982362-153982362 on Build GRCh38
rs553304345 Chr.1: 153982283-153982283 on Build GRCh38
rs56853500 Chr.1: 153982332-153982335 on Build GRCh38
rs533820369 Chr.1: 153982341-153982341 on Build GRCh38
rs752731240 Chr.1: 153982341-153982342 on Build GRCh38
rs759880514 Chr.1: 153982399-153982399 on Build GRCh38
rs529492772 Chr.1: 153982704-153982704 on Build GRCh38
rs113430755 Chr.1: 153982336-153982336 on Build GRCh38
rs867817720 Chr.1: 153982302-153982302 on Build GRCh38
rs118189503 Chr.1: 153982517-153982517 on Build GRCh38
rs767421720 Chr.1: 153982690-153982690 on Build GRCh38
rs573159730 Chr.1: 153982552-153982552 on Build GRCh38
rs769439687 Chr.1: 153982436-153982436 on Build GRCh38
rs563763984 Chr.1: 153982304-153982305 on Build GRCh38
rs562321650 Chr.1: 153982655-153982655 on Build GRCh38
rs757844951 Chr.1: 153982724-153982724 on Build GRCh38
rs547812764 Chr.1: 153982334-153982334 on Build GRCh38
rs71584158 Chr.1: 153982303-153982306 on Build GRCh38
rs556345789 Chr.1: 153982424-153982424 on Build GRCh38
rs573012181 Chr.1: 153982449-153982449 on Build GRCh38
rs530111525 Chr.1: 153982303-153982308 on Build GRCh38
rs201338105 Chr.1: 153982344-153982344 on Build GRCh38
rs1049574987 Chr.1: 153982410-153982410 on Build GRCh38
rs966741898 Chr.1: 153982429-153982429 on Build GRCh38
rs370214600 Chr.1: 153982617-153982617 on Build GRCh38
rs958400512 Chr.1: 153982393-153982393 on Build GRCh38
rs745309724 Chr.1: 153982737-153982737 on Build GRCh38
rs780668783 Chr.1: 153982725-153982725 on Build GRCh38
rs774857336 Chr.1: 153982398-153982398 on Build GRCh38
rs752569127 Chr.1: 153982698-153982698 on Build GRCh38
rs772806404 Chr.1: 153982365-153982365 on Build GRCh38
rs764018486 Chr.1: 153982388-153982389 on Build GRCh38
rs775878719 Chr.1: 153982499-153982499 on Build GRCh38
rs750465602 Chr.1: 153982650-153982650 on Build GRCh38
rs760825713 Chr.1: 153982503-153982503 on Build GRCh38
rs763575836 Chr.1: 153982383-153982383 on Build GRCh38
rs906997565 Chr.1: 153982590-153982590 on Build GRCh38
rs370977932 Chr.1: 153982507-153982507 on Build GRCh38
rs531692477 Chr.1: 153982592-153982592 on Build GRCh38
rs755725192 Chr.1: 153982566-153982566 on Build GRCh38
rs754046939 Chr.1: 153982439-153982440 on Build GRCh38
rs945957540 Chr.1: 153982532-153982532 on Build GRCh38
rs778582488 Chr.1: 153982716-153982716 on Build GRCh38
rs112589571 Chr.1: 153982279-153982279 on Build GRCh38
rs914493714 Chr.1: 153982530-153982530 on Build GRCh38
rs777631292 Chr.1: 153982574-153982574 on Build GRCh38
rs371708064 Chr.1: 153982313-153982340 on Build GRCh38
rs376277494 Chr.1: 153982495-153982495 on Build GRCh38
rs61805527 Chr.1: 153982332-153982332 on Build GRCh38
rs746741597 Chr.1: 153982350-153982350 on Build GRCh38
rs537894434 Chr.1: 153982353-153982353 on Build GRCh38
rs754779874 Chr.1: 153982427-153982427 on Build GRCh38
rs545834123 Chr.1: 153982553-153982553 on Build GRCh38
rs779840771 Chr.1: 153982346-153982346 on Build GRCh38
rs372299956 Chr.1: 153982621-153982621 on Build GRCh38
rs149250462 Chr.1: 153982593-153982593 on Build GRCh38
rs752908416 Chr.1: 153982404-153982404 on Build GRCh38
rs374620206 Chr.1: 153982512-153982512 on Build GRCh38

View Full Product Details