Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 56459065-56459605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
UniGene
Hs.604915
Gene Chromosome Location
Chr.6: 56457987-56954671 on Build GRCh38
Transcripts
NM_001144769.2 NM_001144770.1 NM_015548.4 NM_183380.3 XM_005249310.3 XM_005249315.3 XM_005249316.2 XM_005249318.2 XM_005249319.2 XM_005249320.4 XM_005249322.4 XM_005249323.3 XM_005249324.2 XM_011514824.2 XM_011514825.2 XM_011514826.2 XM_017011205.1 XM_017011206.1 XM_017011207.1 XM_017011208.1 XM_017011209.1 XM_017011210.1 XM_017011211.1 XM_017011212.1 XM_017011213.1 XM_017011214.1 XM_017011215.1 XM_017011216.1 XM_017011217.1 XM_017011218.1 XM_017011219.1 XM_017011220.1 XM_017011221.1 XM_017011222.1 XM_017011223.1 XM_017011224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4387992 c.15432G>T p.R5144R Chr.6: 56459089-56459089 on Build GRCh38
4619292 c.22542A>G p.R7514R Chr.6: 56459101-56459101 on Build GRCh38
374903 c.15288A>G p.R5096R Chr.6: 56459233-56459233 on Build GRCh38
1329616 c.22541G>A p.R7514Q Chr.6: 56459102-56459102 on Build GRCh38
3628944 c.15341C>T p.S5114L Chr.6: 56459180-56459180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924961968 Chr.6: 56459301-56459301 on Build GRCh38
rs559604901 Chr.6: 56459273-56459273 on Build GRCh38
rs1036786598 Chr.6: 56459152-56459152 on Build GRCh38
rs186873881 Chr.6: 56459299-56459299 on Build GRCh38
rs761116868 Chr.6: 56459090-56459090 on Build GRCh38
rs769636857 Chr.6: 56459109-56459109 on Build GRCh38
rs986482511 Chr.6: 56459482-56459482 on Build GRCh38
rs375978950 Chr.6: 56459102-56459102 on Build GRCh38
rs919739578 Chr.6: 56459552-56459552 on Build GRCh38
rs775570122 Chr.6: 56459317-56459317 on Build GRCh38
rs987858406 Chr.6: 56459530-56459530 on Build GRCh38
rs565020912 Chr.6: 56459466-56459466 on Build GRCh38
rs373455908 Chr.6: 56459246-56459246 on Build GRCh38
rs995801681 Chr.6: 56459223-56459223 on Build GRCh38
rs9370531 Chr.6: 56459381-56459381 on Build GRCh38
rs375160184 Chr.6: 56459283-56459283 on Build GRCh38
rs771266784 Chr.6: 56459141-56459141 on Build GRCh38
rs1044651182 Chr.6: 56459564-56459564 on Build GRCh38
rs770683363 Chr.6: 56459459-56459459 on Build GRCh38
rs1014870620 Chr.6: 56459146-56459146 on Build GRCh38
rs763576122 Chr.6: 56459304-56459304 on Build GRCh38
rs1024673310 Chr.6: 56459243-56459243 on Build GRCh38
rs774765327 Chr.6: 56459151-56459151 on Build GRCh38
rs183716103 Chr.6: 56459504-56459504 on Build GRCh38
rs766998736 Chr.6: 56459270-56459270 on Build GRCh38
rs577012766 Chr.6: 56459107-56459107 on Build GRCh38
rs373384542 Chr.6: 56459305-56459305 on Build GRCh38
rs775454333 Chr.6: 56459195-56459195 on Build GRCh38
rs1024766434 Chr.6: 56459494-56459494 on Build GRCh38
rs1010331369 Chr.6: 56459314-56459314 on Build GRCh38
rs777565037 Chr.6: 56459496-56459496 on Build GRCh38
rs868335418 Chr.6: 56459199-56459199 on Build GRCh38
rs764508787 Chr.6: 56459208-56459208 on Build GRCh38
rs965619323 Chr.6: 56459541-56459541 on Build GRCh38
rs765987178 Chr.6: 56459242-56459242 on Build GRCh38
rs541271813 Chr.6: 56459225-56459225 on Build GRCh38
rs888431191 Chr.6: 56459413-56459413 on Build GRCh38
rs749070889 Chr.6: 56459293-56459293 on Build GRCh38
rs754652342 Chr.6: 56459256-56459256 on Build GRCh38
rs772149151 Chr.6: 56459192-56459192 on Build GRCh38
rs142108560 Chr.6: 56459469-56459469 on Build GRCh38
rs147719000 Chr.6: 56459306-56459306 on Build GRCh38
rs962169183 Chr.6: 56459215-56459215 on Build GRCh38
rs759656642 Chr.6: 56459180-56459180 on Build GRCh38
rs777558951 Chr.6: 56459114-56459114 on Build GRCh38
rs932741600 Chr.6: 56459356-56459356 on Build GRCh38
rs777631996 Chr.6: 56459287-56459287 on Build GRCh38
rs931145632 Chr.6: 56459554-56459554 on Build GRCh38
rs957937826 Chr.6: 56459513-56459513 on Build GRCh38
rs766870965 Chr.6: 56459335-56459335 on Build GRCh38
rs914103348 Chr.6: 56459531-56459531 on Build GRCh38
rs939814173 Chr.6: 56459137-56459137 on Build GRCh38
rs757157314 Chr.6: 56459296-56459296 on Build GRCh38
rs774450561 Chr.6: 56459298-56459300 on Build GRCh38
rs756074647 Chr.6: 56459280-56459280 on Build GRCh38
rs969089370 Chr.6: 56459495-56459495 on Build GRCh38
rs754164327 Chr.6: 56459091-56459091 on Build GRCh38
rs749021445 Chr.6: 56459575-56459575 on Build GRCh38
rs899876251 Chr.6: 56459337-56459337 on Build GRCh38
rs780600756 Chr.6: 56459108-56459108 on Build GRCh38
rs944178802 Chr.6: 56459331-56459331 on Build GRCh38
rs762220017 Chr.6: 56459234-56459234 on Build GRCh38
rs369312019 Chr.6: 56459198-56459198 on Build GRCh38
rs779746657 Chr.6: 56459103-56459103 on Build GRCh38
rs191534310 Chr.6: 56459431-56459431 on Build GRCh38
rs777047077 Chr.6: 56459222-56459222 on Build GRCh38
rs968892575 Chr.6: 56459375-56459375 on Build GRCh38
rs751052704 Chr.6: 56459104-56459104 on Build GRCh38
rs749163618 Chr.6: 56459128-56459128 on Build GRCh38
rs558022526 Chr.6: 56459097-56459097 on Build GRCh38

View Full Product Details