Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63581511-63582053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTP4A1
Entrez Gene ID
7803
Gene Name
protein tyrosine phosphatase type IVA, member 1
Gene Aliases
HH72, PRL-1, PRL1, PTP(CAAX1), PTPCAAX1
UniGene
Hs.227777
Gene Chromosome Location
Chr.6: 63528021-63583588 on Build GRCh38
Transcripts
NM_003463.4 XM_011536111.1 XM_011536112.1 XM_017011270.1 XM_017011271.1

SNP Detail

SNP ID Chromosome Location
rs1048106424 Chr.6: 63581720-63581720 on Build GRCh38
rs1062182 Chr.6: 63581790-63581790 on Build GRCh38
rs896855225 Chr.6: 63581585-63581585 on Build GRCh38
rs931615716 Chr.6: 63581747-63581747 on Build GRCh38
rs891985104 Chr.6: 63581560-63581560 on Build GRCh38
rs544280994 Chr.6: 63581985-63581985 on Build GRCh38
rs146443685 Chr.6: 63581627-63581627 on Build GRCh38
rs1001309217 Chr.6: 63581734-63581734 on Build GRCh38
rs1062185 Chr.6: 63581841-63581841 on Build GRCh38
rs573197502 Chr.6: 63581826-63581826 on Build GRCh38
rs754953096 Chr.6: 63581787-63581787 on Build GRCh38
rs561990964 Chr.6: 63581770-63581770 on Build GRCh38
rs569458079 Chr.6: 63581651-63581651 on Build GRCh38
rs773256959 Chr.6: 63581547-63581547 on Build GRCh38
rs144571773 Chr.6: 63581936-63581936 on Build GRCh38
rs1062179 Chr.6: 63581588-63581588 on Build GRCh38
rs77882330 Chr.6: 63581775-63581775 on Build GRCh38
rs1027691066 Chr.6: 63581628-63581628 on Build GRCh38
rs575033403 Chr.6: 63581989-63581989 on Build GRCh38
rs953471896 Chr.6: 63581954-63581954 on Build GRCh38
rs907686028 Chr.6: 63581647-63581647 on Build GRCh38
rs1010187909 Chr.6: 63581881-63581881 on Build GRCh38
rs763266300 Chr.6: 63581644-63581644 on Build GRCh38
rs1062184 Chr.6: 63581831-63581831 on Build GRCh38
rs1052644664 Chr.6: 63581819-63581819 on Build GRCh38
rs924990858 Chr.6: 63581800-63581800 on Build GRCh38
rs921855027 Chr.6: 63581713-63581713 on Build GRCh38
rs1014479356 Chr.6: 63581568-63581568 on Build GRCh38
rs1053288837 Chr.6: 63581553-63581553 on Build GRCh38
rs536080435 Chr.6: 63581541-63581541 on Build GRCh38
rs975896808 Chr.6: 63581947-63581947 on Build GRCh38
rs866058268 Chr.6: 63581685-63581685 on Build GRCh38
rs1062187 Chr.6: 63581866-63581866 on Build GRCh38
rs371491460 Chr.6: 63581917-63581917 on Build GRCh38
rs1062183 Chr.6: 63581822-63581822 on Build GRCh38
rs935079125 Chr.6: 63581808-63581808 on Build GRCh38
rs975949239 Chr.6: 63581686-63581686 on Build GRCh38
rs771764111 Chr.6: 63581611-63581611 on Build GRCh38
rs952170773 Chr.6: 63581900-63581900 on Build GRCh38
rs1024579137 Chr.6: 63581599-63581599 on Build GRCh38
rs540155068 Chr.6: 63581727-63581727 on Build GRCh38
rs766304644 Chr.6: 63581655-63581655 on Build GRCh38
rs1020627821 Chr.6: 63581885-63581885 on Build GRCh38
rs140927727 Chr.6: 63581899-63581899 on Build GRCh38
rs891938529 Chr.6: 63581823-63581823 on Build GRCh38
rs921983051 Chr.6: 63581925-63581925 on Build GRCh38
rs965867728 Chr.6: 63581946-63581947 on Build GRCh38
rs953295725 Chr.6: 63581965-63581970 on Build GRCh38
rs979343828 Chr.6: 63582010-63582010 on Build GRCh38
rs186956654 Chr.6: 63581960-63581960 on Build GRCh38
rs983964817 Chr.6: 63581647-63581647 on Build GRCh38
rs970005052 Chr.6: 63581607-63581607 on Build GRCh38
rs1062180 Chr.6: 63581748-63581748 on Build GRCh38
rs761409438 Chr.6: 63581586-63581587 on Build GRCh38
rs1037104271 Chr.6: 63581571-63581571 on Build GRCh38
rs767630066 Chr.6: 63581868-63581868 on Build GRCh38
rs1045979116 Chr.6: 63581829-63581829 on Build GRCh38
rs1062186 Chr.6: 63581855-63581855 on Build GRCh38
rs751704599 Chr.6: 63581677-63581677 on Build GRCh38
rs752513401 Chr.6: 63581890-63581890 on Build GRCh38
rs1035510563 Chr.6: 63581752-63581752 on Build GRCh38
rs978776401 Chr.6: 63581916-63581916 on Build GRCh38

View Full Product Details