Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 112113931-112114432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMA4
Entrez Gene ID
3910
Gene Name
laminin subunit alpha 4
Gene Aliases
CMD1JJ, LAMA3, LAMA4*-1
UniGene
Hs.654572
Gene Chromosome Location
Chr.6: 112107931-112254722 on Build GRCh38
Transcripts
NM_001105206.2 NM_001105207.2 NM_002290.4 XM_005266983.3 XM_005266984.3 XM_017010854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4499023 c.5302C>T p.P1768S Chr.6: 112114079-112114079 on Build GRCh38
3828845 c.5272C>A p.H1758N Chr.6: 112114109-112114109 on Build GRCh38
4498586 c.5209C>A p.Q1737K Chr.6: 112114172-112114172 on Build GRCh38
4941112 c.5277G>C p.R1759S Chr.6: 112114104-112114104 on Build GRCh38
1072280 c.5304A>G p.P1768P Chr.6: 112114077-112114077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782227058 Chr.6: 112114118-112114118 on Build GRCh38
rs916580810 Chr.6: 112114169-112114169 on Build GRCh38
rs782244308 Chr.6: 112114135-112114135 on Build GRCh38
rs80354156 Chr.6: 112113983-112113983 on Build GRCh38
rs781928055 Chr.6: 112114233-112114233 on Build GRCh38
rs782677810 Chr.6: 112114221-112114221 on Build GRCh38
rs782239919 Chr.6: 112114237-112114237 on Build GRCh38
rs143630621 Chr.6: 112113973-112113973 on Build GRCh38
rs371370857 Chr.6: 112114148-112114148 on Build GRCh38
rs201734281 Chr.6: 112114018-112114018 on Build GRCh38
rs112726836 Chr.6: 112114329-112114329 on Build GRCh38
rs765685480 Chr.6: 112114388-112114388 on Build GRCh38
rs782765309 Chr.6: 112114076-112114076 on Build GRCh38
rs587649673 Chr.6: 112114333-112114333 on Build GRCh38
rs587773850 Chr.6: 112114252-112114252 on Build GRCh38
rs782487010 Chr.6: 112114207-112114207 on Build GRCh38
rs781964329 Chr.6: 112114029-112114029 on Build GRCh38
rs782561500 Chr.6: 112114111-112114111 on Build GRCh38
rs200177134 Chr.6: 112114132-112114132 on Build GRCh38
rs1052938515 Chr.6: 112114277-112114277 on Build GRCh38
rs890255814 Chr.6: 112114320-112114320 on Build GRCh38
rs587635507 Chr.6: 112114297-112114297 on Build GRCh38
rs587687200 Chr.6: 112114325-112114325 on Build GRCh38
rs375447272 Chr.6: 112114161-112114161 on Build GRCh38
rs3734290 Chr.6: 112114061-112114061 on Build GRCh38
rs782001026 Chr.6: 112114133-112114133 on Build GRCh38
rs934515853 Chr.6: 112114257-112114257 on Build GRCh38
rs782272670 Chr.6: 112114100-112114100 on Build GRCh38
rs371842580 Chr.6: 112114211-112114211 on Build GRCh38
rs781789940 Chr.6: 112114204-112114204 on Build GRCh38
rs782557878 Chr.6: 112114114-112114114 on Build GRCh38
rs34263758 Chr.6: 112114243-112114244 on Build GRCh38
rs782515304 Chr.6: 112114219-112114219 on Build GRCh38
rs782349629 Chr.6: 112114244-112114244 on Build GRCh38
rs782498118 Chr.6: 112114093-112114093 on Build GRCh38
rs145912332 Chr.6: 112113976-112113976 on Build GRCh38
rs782331662 Chr.6: 112114227-112114227 on Build GRCh38
rs1017493584 Chr.6: 112114339-112114339 on Build GRCh38
rs587647752 Chr.6: 112114192-112114192 on Build GRCh38
rs781826525 Chr.6: 112114084-112114084 on Build GRCh38
rs782710303 Chr.6: 112114200-112114200 on Build GRCh38
rs782679484 Chr.6: 112114098-112114098 on Build GRCh38
rs782532472 Chr.6: 112114195-112114195 on Build GRCh38
rs782776313 Chr.6: 112114190-112114190 on Build GRCh38
rs587714666 Chr.6: 112114356-112114356 on Build GRCh38
rs782078478 Chr.6: 112114230-112114230 on Build GRCh38
rs781896225 Chr.6: 112114124-112114124 on Build GRCh38
rs587726125 Chr.6: 112114144-112114144 on Build GRCh38
rs782078996 Chr.6: 112114047-112114047 on Build GRCh38
rs781975239 Chr.6: 112114171-112114171 on Build GRCh38
rs782086889 Chr.6: 112114189-112114189 on Build GRCh38
rs184641552 Chr.6: 112114007-112114007 on Build GRCh38
rs781991737 Chr.6: 112114066-112114066 on Build GRCh38
rs776325910 Chr.6: 112114240-112114240 on Build GRCh38
rs782446664 Chr.6: 112114104-112114104 on Build GRCh38
rs782070205 Chr.6: 112114158-112114158 on Build GRCh38
rs111835795 Chr.6: 112114241-112114242 on Build GRCh38
rs3734289 Chr.6: 112114070-112114070 on Build GRCh38
rs112265545 Chr.6: 112114209-112114209 on Build GRCh38
rs782426589 Chr.6: 112114143-112114143 on Build GRCh38
rs200663437 Chr.6: 112114244-112114245 on Build GRCh38
rs782281404 Chr.6: 112114225-112114225 on Build GRCh38
rs781913226 Chr.6: 112114240-112114241 on Build GRCh38

View Full Product Details