Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 112456772-112457276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFRD1
Entrez Gene ID
3475
Gene Name
interferon related developmental regulator 1
Gene Aliases
PC4, TIS7
UniGene
Hs.744747
Gene Chromosome Location
Chr.7: 112423144-112477203 on Build GRCh38
Transcripts
NM_001007245.2 NM_001197079.1 NM_001197080.1 NM_001550.3 NR_120333.1 XM_011516142.2 XM_011516143.2 XM_017012151.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1084118 c.313G>T p.G105C Chr.7: 112456942-112456942 on Build GRCh38
1084117 c.288G>A p.A96A Chr.7: 112456917-112456917 on Build GRCh38
1673462 c.395G>A p.R132H Chr.7: 112457024-112457024 on Build GRCh38
4169815 c.409+1G>C p.? Chr.7: 112457039-112457039 on Build GRCh38
5612147 c.333T>A p.A111A Chr.7: 112456962-112456962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73717557 Chr.7: 112457244-112457244 on Build GRCh38
rs767576072 Chr.7: 112456976-112456976 on Build GRCh38
rs192770035 Chr.7: 112457221-112457221 on Build GRCh38
rs756796129 Chr.7: 112456914-112456914 on Build GRCh38
rs6968084 Chr.7: 112457066-112457066 on Build GRCh38
rs768418717 Chr.7: 112457067-112457067 on Build GRCh38
rs6967894 Chr.7: 112456825-112456825 on Build GRCh38
rs200230784 Chr.7: 112457009-112457009 on Build GRCh38
rs548522773 Chr.7: 112457020-112457020 on Build GRCh38
rs754099331 Chr.7: 112457003-112457003 on Build GRCh38
rs907783955 Chr.7: 112456973-112456973 on Build GRCh38
rs760833178 Chr.7: 112457115-112457115 on Build GRCh38
rs570970894 Chr.7: 112457090-112457090 on Build GRCh38
rs537840872 Chr.7: 112457092-112457092 on Build GRCh38
rs766640868 Chr.7: 112456992-112456992 on Build GRCh38
rs1033213392 Chr.7: 112456806-112456806 on Build GRCh38
rs745386575 Chr.7: 112457028-112457028 on Build GRCh38
rs370064018 Chr.7: 112456864-112456864 on Build GRCh38
rs899954614 Chr.7: 112457034-112457034 on Build GRCh38
rs760744367 Chr.7: 112456990-112456990 on Build GRCh38
rs749131911 Chr.7: 112456937-112456937 on Build GRCh38
rs947803641 Chr.7: 112456869-112456869 on Build GRCh38
rs141889729 Chr.7: 112457033-112457033 on Build GRCh38
rs562479630 Chr.7: 112456971-112456971 on Build GRCh38
rs79917788 Chr.7: 112456827-112456827 on Build GRCh38
rs983180906 Chr.7: 112457181-112457181 on Build GRCh38
rs148147163 Chr.7: 112456980-112456980 on Build GRCh38
rs761788207 Chr.7: 112456974-112456974 on Build GRCh38
rs554079702 Chr.7: 112457215-112457215 on Build GRCh38
rs759032177 Chr.7: 112457024-112457024 on Build GRCh38
rs746476935 Chr.7: 112456872-112456872 on Build GRCh38
rs775079574 Chr.7: 112456917-112456917 on Build GRCh38
rs768440582 Chr.7: 112456943-112456943 on Build GRCh38
rs758887456 Chr.7: 112457078-112457080 on Build GRCh38
rs756305677 Chr.7: 112457162-112457162 on Build GRCh38
rs964082603 Chr.7: 112456824-112456824 on Build GRCh38
rs550593403 Chr.7: 112456916-112456916 on Build GRCh38
rs371480128 Chr.7: 112457045-112457045 on Build GRCh38
rs535715681 Chr.7: 112457161-112457161 on Build GRCh38
rs376871952 Chr.7: 112457023-112457023 on Build GRCh38
rs60461435 Chr.7: 112457241-112457241 on Build GRCh38
rs570093546 Chr.7: 112457106-112457106 on Build GRCh38
rs546422463 Chr.7: 112457222-112457222 on Build GRCh38
rs752038407 Chr.7: 112456796-112456796 on Build GRCh38
rs776877608 Chr.7: 112457124-112457124 on Build GRCh38
rs778104593 Chr.7: 112457171-112457171 on Build GRCh38
rs912523978 Chr.7: 112457247-112457247 on Build GRCh38
rs552367277 Chr.7: 112457072-112457072 on Build GRCh38
rs780843547 Chr.7: 112457027-112457027 on Build GRCh38
rs568116708 Chr.7: 112457160-112457160 on Build GRCh38
rs1000480162 Chr.7: 112457101-112457101 on Build GRCh38
rs937049450 Chr.7: 112457018-112457018 on Build GRCh38
rs919760025 Chr.7: 112456882-112456882 on Build GRCh38
rs189364948 Chr.7: 112457152-112457152 on Build GRCh38
rs771016283 Chr.7: 112457119-112457119 on Build GRCh38
rs778046574 Chr.7: 112457105-112457106 on Build GRCh38
rs199645932 Chr.7: 112457228-112457229 on Build GRCh38

View Full Product Details