Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33384872-33385370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP7
Entrez Gene ID
364
Gene Name
aquaporin 7
Gene Aliases
AQP7L, AQPap, GLYCQTL
UniGene
Hs.455323
Gene Chromosome Location
Chr.9: 33383135-33402682 on Build GRCh38
Transcripts
NM_001170.2 NM_001318156.1 NM_001318157.1 NM_001318158.1 NR_134513.1 NR_134514.1 NR_134515.1 XM_005251453.3 XM_006716765.3 XM_011517866.2 XM_011517867.2 XM_017014700.1 XM_017014701.1 XM_017014702.1 XM_017014703.1 XM_017014704.1 XM_017014705.1 XM_017014706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3656929 c.1013C>T p.A338V Chr.9: 33385021-33385021 on Build GRCh38
6183314 c.930G>T p.T310T Chr.9: 33385104-33385104 on Build GRCh38
7171764 c.919G>A p.E307K Chr.9: 33385112-33385112 on Build GRCh38
3656931 c.750G>A p.G250G Chr.9: 33385284-33385284 on Build GRCh38
3763887 c.791G>T p.G264V Chr.9: 33385243-33385243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774564666 Chr.9: 33385189-33385189 on Build GRCh38
rs541770524 Chr.9: 33385105-33385105 on Build GRCh38
rs76796426 Chr.9: 33384962-33384962 on Build GRCh38
rs61073250 Chr.9: 33385136-33385136 on Build GRCh38
rs755536859 Chr.9: 33385342-33385342 on Build GRCh38
rs766813164 Chr.9: 33385007-33385007 on Build GRCh38
rs755989154 Chr.9: 33385069-33385069 on Build GRCh38
rs749056562 Chr.9: 33385073-33385073 on Build GRCh38
rs774041307 Chr.9: 33384989-33384989 on Build GRCh38
rs769073570 Chr.9: 33385188-33385188 on Build GRCh38
rs777485460 Chr.9: 33385312-33385312 on Build GRCh38
rs144909659 Chr.9: 33385104-33385104 on Build GRCh38
rs13434 Chr.9: 33384979-33384979 on Build GRCh38
rs74557595 Chr.9: 33385289-33385289 on Build GRCh38
rs199871527 Chr.9: 33385042-33385042 on Build GRCh38
rs542757882 Chr.9: 33385153-33385153 on Build GRCh38
rs770805820 Chr.9: 33384959-33384959 on Build GRCh38
rs758073443 Chr.9: 33385311-33385311 on Build GRCh38
rs968498938 Chr.9: 33385260-33385260 on Build GRCh38
rs1014957126 Chr.9: 33384941-33384941 on Build GRCh38
rs759587407 Chr.9: 33385295-33385295 on Build GRCh38
rs561584740 Chr.9: 33385231-33385231 on Build GRCh38
rs773244652 Chr.9: 33385285-33385285 on Build GRCh38
rs769128601 Chr.9: 33385276-33385276 on Build GRCh38
rs745882902 Chr.9: 33385057-33385057 on Build GRCh38
rs752203742 Chr.9: 33385328-33385329 on Build GRCh38
rs751277502 Chr.9: 33385314-33385314 on Build GRCh38
rs201424877 Chr.9: 33385072-33385072 on Build GRCh38
rs757414925 Chr.9: 33385127-33385127 on Build GRCh38
rs752561448 Chr.9: 33385302-33385302 on Build GRCh38
rs768393072 Chr.9: 33385190-33385190 on Build GRCh38
rs770359710 Chr.9: 33385250-33385250 on Build GRCh38
rs770308564 Chr.9: 33385177-33385177 on Build GRCh38
rs751738116 Chr.9: 33385124-33385124 on Build GRCh38
rs760785390 Chr.9: 33385204-33385204 on Build GRCh38
rs1006473217 Chr.9: 33384912-33384912 on Build GRCh38
rs761959042 Chr.9: 33384982-33384982 on Build GRCh38
rs753896238 Chr.9: 33385229-33385229 on Build GRCh38
rs965074454 Chr.9: 33385322-33385322 on Build GRCh38
rs370340757 Chr.9: 33385202-33385202 on Build GRCh38
rs76602126 Chr.9: 33385313-33385313 on Build GRCh38
rs754341549 Chr.9: 33385269-33385271 on Build GRCh38
rs535195177 Chr.9: 33385011-33385011 on Build GRCh38
rs754560189 Chr.9: 33385283-33385283 on Build GRCh38
rs376205588 Chr.9: 33385147-33385147 on Build GRCh38
rs202043723 Chr.9: 33385237-33385237 on Build GRCh38
rs749852149 Chr.9: 33385273-33385273 on Build GRCh38
rs745346942 Chr.9: 33385339-33385339 on Build GRCh38
rs778264784 Chr.9: 33385317-33385326 on Build GRCh38
rs756889332 Chr.9: 33385326-33385326 on Build GRCh38
rs768396947 Chr.9: 33384986-33384986 on Build GRCh38
rs140886274 Chr.9: 33385137-33385137 on Build GRCh38
rs375356467 Chr.9: 33385278-33385278 on Build GRCh38
rs745663446 Chr.9: 33384966-33384966 on Build GRCh38
rs772587113 Chr.9: 33385090-33385090 on Build GRCh38
rs868186854 Chr.9: 33385081-33385081 on Build GRCh38
rs113571472 Chr.9: 33385055-33385055 on Build GRCh38
rs199594532 Chr.9: 33385085-33385085 on Build GRCh38
rs368879439 Chr.9: 33385155-33385155 on Build GRCh38
rs750788444 Chr.9: 33385086-33385086 on Build GRCh38
rs767736435 Chr.9: 33385022-33385023 on Build GRCh38
rs78431873 Chr.9: 33385049-33385049 on Build GRCh38
rs767600442 Chr.9: 33385191-33385191 on Build GRCh38
rs769564313 Chr.9: 33384971-33384971 on Build GRCh38
rs976861009 Chr.9: 33385343-33385343 on Build GRCh38
rs764313048 Chr.9: 33385123-33385123 on Build GRCh38
rs202003063 Chr.9: 33385180-33385180 on Build GRCh38
rs759960026 Chr.9: 33385014-33385014 on Build GRCh38
rs760977156 Chr.9: 33385286-33385286 on Build GRCh38
rs780728509 Chr.9: 33385338-33385338 on Build GRCh38
rs796349423 Chr.9: 33385079-33385079 on Build GRCh38
rs774092608 Chr.9: 33385086-33385086 on Build GRCh38
rs184118433 Chr.9: 33385244-33385244 on Build GRCh38
rs766932364 Chr.9: 33385165-33385165 on Build GRCh38
rs780095371 Chr.9: 33385142-33385142 on Build GRCh38
rs760974548 Chr.9: 33385093-33385093 on Build GRCh38
rs767693529 Chr.9: 33385129-33385129 on Build GRCh38
rs766426189 Chr.9: 33385205-33385205 on Build GRCh38
rs977722895 Chr.9: 33385113-33385113 on Build GRCh38
rs765046155 Chr.9: 33385297-33385297 on Build GRCh38
rs62542743 Chr.9: 33385243-33385243 on Build GRCh38
rs749112658 Chr.9: 33385145-33385145 on Build GRCh38
rs368432936 Chr.9: 33385021-33385021 on Build GRCh38
rs958540175 Chr.9: 33385022-33385022 on Build GRCh38
rs771682923 Chr.9: 33385087-33385087 on Build GRCh38
rs201527236 Chr.9: 33385159-33385159 on Build GRCh38
rs778848616 Chr.9: 33385149-33385149 on Build GRCh38
rs888067703 Chr.9: 33384906-33384906 on Build GRCh38
rs768450241 Chr.9: 33385080-33385080 on Build GRCh38
rs761341115 Chr.9: 33384999-33384999 on Build GRCh38
rs780673396 Chr.9: 33385255-33385255 on Build GRCh38
rs3569 Chr.9: 33385167-33385167 on Build GRCh38
rs775864068 Chr.9: 33385119-33385119 on Build GRCh38
rs1031970138 Chr.9: 33385301-33385301 on Build GRCh38
rs780994832 Chr.9: 33385056-33385056 on Build GRCh38
rs75724774 Chr.9: 33384953-33384953 on Build GRCh38
rs763311844 Chr.9: 33385184-33385184 on Build GRCh38
rs116974820 Chr.9: 33385284-33385284 on Build GRCh38
rs371523158 Chr.9: 33385122-33385122 on Build GRCh38
rs749003610 Chr.9: 33384980-33384980 on Build GRCh38
rs190625868 Chr.9: 33385198-33385198 on Build GRCh38
rs144994089 Chr.9: 33385158-33385158 on Build GRCh38
rs149110103 Chr.9: 33385017-33385017 on Build GRCh38
rs1002979398 Chr.9: 33384982-33384982 on Build GRCh38
rs766629103 Chr.9: 33385287-33385287 on Build GRCh38
rs758660571 Chr.9: 33385039-33385039 on Build GRCh38

View Full Product Details