Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37428403-37428900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHPR
Entrez Gene ID
9380
Gene Name
glyoxylate reductase/hydroxypyruvate reductase
Gene Aliases
GLXR, GLYD, PH2
UniGene
Hs.155742
Gene Chromosome Location
Chr.9: 37422666-37437782 on Build GRCh38
Transcripts
NM_012203.1 XM_005251631.1 XM_011518073.2 XM_017015320.1 XM_017015321.1 XM_017015322.1 XM_017015323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6674968 c.435G>A p.W145* Chr.9: 37428514-37428514 on Build GRCh38
4956668 c.469G>A p.G157S Chr.9: 37428548-37428548 on Build GRCh38
4956669 c.469G>A p.G157S Chr.9: 37428548-37428548 on Build GRCh38
5825295 c.432C>A p.L144L Chr.9: 37428511-37428511 on Build GRCh38
6674969 c.435G>A p.W145* Chr.9: 37428514-37428514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775855961 Chr.9: 37428636-37428636 on Build GRCh38
rs979531614 Chr.9: 37428452-37428452 on Build GRCh38
rs540651297 Chr.9: 37428441-37428441 on Build GRCh38
rs760782027 Chr.9: 37428566-37428566 on Build GRCh38
rs766613829 Chr.9: 37428717-37428717 on Build GRCh38
rs963807728 Chr.9: 37428790-37428790 on Build GRCh38
rs1057517333 Chr.9: 37428514-37428515 on Build GRCh38
rs978452673 Chr.9: 37428757-37428757 on Build GRCh38
rs754089807 Chr.9: 37428568-37428568 on Build GRCh38
rs992901517 Chr.9: 37428430-37428430 on Build GRCh38
rs948242663 Chr.9: 37428448-37428448 on Build GRCh38
rs1047080155 Chr.9: 37428851-37428851 on Build GRCh38
rs774233908 Chr.9: 37428548-37428548 on Build GRCh38
rs369699672 Chr.9: 37428601-37428601 on Build GRCh38
rs1013950815 Chr.9: 37428639-37428639 on Build GRCh38
rs768726104 Chr.9: 37428626-37428626 on Build GRCh38
rs759447197 Chr.9: 37428554-37428554 on Build GRCh38
rs933885886 Chr.9: 37428824-37428824 on Build GRCh38
rs979605409 Chr.9: 37428715-37428715 on Build GRCh38
rs763398450 Chr.9: 37428696-37428696 on Build GRCh38
rs752806700 Chr.9: 37428464-37428464 on Build GRCh38
rs941435896 Chr.9: 37428874-37428874 on Build GRCh38
rs746364242 Chr.9: 37428491-37428491 on Build GRCh38
rs760489628 Chr.9: 37428621-37428621 on Build GRCh38
rs944388378 Chr.9: 37428842-37428842 on Build GRCh38
rs769852092 Chr.9: 37428632-37428633 on Build GRCh38
rs746648380 Chr.9: 37428591-37428591 on Build GRCh38
rs147934112 Chr.9: 37428605-37428605 on Build GRCh38
rs146896070 Chr.9: 37428599-37428599 on Build GRCh38
rs755488609 Chr.9: 37428776-37428776 on Build GRCh38
rs376674882 Chr.9: 37428610-37428610 on Build GRCh38
rs748913563 Chr.9: 37428537-37428537 on Build GRCh38
rs1038425975 Chr.9: 37428875-37428875 on Build GRCh38
rs750803720 Chr.9: 37428684-37428684 on Build GRCh38
rs376052610 Chr.9: 37428427-37428427 on Build GRCh38
rs747664983 Chr.9: 37428534-37428534 on Build GRCh38
rs771019056 Chr.9: 37428532-37428533 on Build GRCh38
rs1057516941 Chr.9: 37428533-37428534 on Build GRCh38
rs180177312 Chr.9: 37428557-37428557 on Build GRCh38
rs759810267 Chr.9: 37428628-37428629 on Build GRCh38
rs371264136 Chr.9: 37428616-37428616 on Build GRCh38
rs925796156 Chr.9: 37428763-37428763 on Build GRCh38
rs935590752 Chr.9: 37428508-37428508 on Build GRCh38
rs776632391 Chr.9: 37428632-37428632 on Build GRCh38
rs763263606 Chr.9: 37428730-37428731 on Build GRCh38
rs757641237 Chr.9: 37428478-37428478 on Build GRCh38
rs770696432 Chr.9: 37428606-37428606 on Build GRCh38
rs764111479 Chr.9: 37428567-37428567 on Build GRCh38
rs142509393 Chr.9: 37428547-37428547 on Build GRCh38
rs776773819 Chr.9: 37428626-37428626 on Build GRCh38
rs309462 Chr.9: 37428799-37428799 on Build GRCh38
rs1019890612 Chr.9: 37428706-37428706 on Build GRCh38
rs778954463 Chr.9: 37428447-37428447 on Build GRCh38
rs780811992 Chr.9: 37428867-37428867 on Build GRCh38
rs891447278 Chr.9: 37428527-37428527 on Build GRCh38
rs754245462 Chr.9: 37428475-37428475 on Build GRCh38
rs140612021 Chr.9: 37428535-37428535 on Build GRCh38
rs377116116 Chr.9: 37428613-37428613 on Build GRCh38
rs747097881 Chr.9: 37428468-37428469 on Build GRCh38
rs779208888 Chr.9: 37428483-37428483 on Build GRCh38
rs959696480 Chr.9: 37428428-37428428 on Build GRCh38
rs1057517026 Chr.9: 37428520-37428521 on Build GRCh38
rs560441686 Chr.9: 37428526-37428526 on Build GRCh38
rs376781208 Chr.9: 37428807-37428807 on Build GRCh38
rs762078399 Chr.9: 37428651-37428651 on Build GRCh38
rs780588937 Chr.9: 37428592-37428592 on Build GRCh38
rs967118114 Chr.9: 37428730-37428730 on Build GRCh38
rs180177313 Chr.9: 37428574-37428574 on Build GRCh38
rs566785032 Chr.9: 37428674-37428674 on Build GRCh38
rs758880537 Chr.9: 37428499-37428499 on Build GRCh38
rs556203401 Chr.9: 37428712-37428712 on Build GRCh38
rs780635481 Chr.9: 37428509-37428509 on Build GRCh38
rs41303225 Chr.9: 37428581-37428581 on Build GRCh38
rs767483014 Chr.9: 37428556-37428556 on Build GRCh38
rs767510376 Chr.9: 37428453-37428453 on Build GRCh38
rs756355572 Chr.9: 37428468-37428468 on Build GRCh38
rs752115702 Chr.9: 37428718-37428718 on Build GRCh38
rs935609701 Chr.9: 37428734-37428734 on Build GRCh38
rs373472631 Chr.9: 37428604-37428604 on Build GRCh38
rs912782306 Chr.9: 37428810-37428810 on Build GRCh38
rs765596317 Chr.9: 37428585-37428585 on Build GRCh38
rs373213853 Chr.9: 37428617-37428617 on Build GRCh38
rs201661750 Chr.9: 37428519-37428519 on Build GRCh38
rs908515557 Chr.9: 37428868-37428868 on Build GRCh38
rs975647392 Chr.9: 37428798-37428798 on Build GRCh38
rs566239341 Chr.9: 37428498-37428498 on Build GRCh38
rs765415889 Chr.9: 37428670-37428670 on Build GRCh38
rs1053220809 Chr.9: 37428514-37428514 on Build GRCh38
rs141090601 Chr.9: 37428586-37428586 on Build GRCh38
rs374838995 Chr.9: 37428600-37428600 on Build GRCh38

View Full Product Details