Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121339659-121340157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STOM
Entrez Gene ID
2040
Gene Name
stomatin
Gene Aliases
BND7, EPB7, EPB72
UniGene
Hs.253903
Gene Chromosome Location
Chr.9: 121338988-121370304 on Build GRCh38
Transcripts
NM_001270526.1 NM_001270527.1 NM_004099.5 NM_198194.2 NR_073037.1

SNP Detail

SNP ID Chromosome Location
rs1027222932 Chr.9: 121340055-121340055 on Build GRCh38
rs964057417 Chr.9: 121339996-121339996 on Build GRCh38
rs1042977039 Chr.9: 121339755-121339755 on Build GRCh38
rs144554153 Chr.9: 121339691-121339691 on Build GRCh38
rs934302278 Chr.9: 121339844-121339844 on Build GRCh38
rs890127746 Chr.9: 121339888-121339888 on Build GRCh38
rs564728882 Chr.9: 121339933-121339933 on Build GRCh38
rs192441823 Chr.9: 121340043-121340043 on Build GRCh38
rs187270518 Chr.9: 121339893-121339893 on Build GRCh38
rs762236836 Chr.9: 121339928-121339928 on Build GRCh38
rs760123260 Chr.9: 121340110-121340113 on Build GRCh38
rs1051405918 Chr.9: 121339882-121339882 on Build GRCh38
rs79009054 Chr.9: 121340080-121340080 on Build GRCh38
rs563727450 Chr.9: 121339890-121339890 on Build GRCh38
rs1001893037 Chr.9: 121339852-121339852 on Build GRCh38
rs957818898 Chr.9: 121340079-121340079 on Build GRCh38
rs1037433169 Chr.9: 121339878-121339878 on Build GRCh38
rs991988448 Chr.9: 121339702-121339702 on Build GRCh38
rs1019592253 Chr.9: 121339968-121339968 on Build GRCh38
rs16910556 Chr.9: 121340072-121340072 on Build GRCh38
rs947243594 Chr.9: 121339725-121339725 on Build GRCh38
rs913759453 Chr.9: 121340083-121340083 on Build GRCh38
rs764740314 Chr.9: 121339960-121339960 on Build GRCh38
rs1008924777 Chr.9: 121339964-121339964 on Build GRCh38
rs562063791 Chr.9: 121340081-121340081 on Build GRCh38
rs924546772 Chr.9: 121339841-121339841 on Build GRCh38
rs762649751 Chr.9: 121340074-121340074 on Build GRCh38
rs41273434 Chr.9: 121339904-121339904 on Build GRCh38
rs975465671 Chr.9: 121340064-121340064 on Build GRCh38
rs991582267 Chr.9: 121340109-121340109 on Build GRCh38
rs561757880 Chr.9: 121339804-121339804 on Build GRCh38
rs960040452 Chr.9: 121340106-121340106 on Build GRCh38
rs915713524 Chr.9: 121339708-121339708 on Build GRCh38

View Full Product Details