Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206557469-206557969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASSF5
Entrez Gene ID
83593
Gene Name
Ras association domain family member 5
Gene Aliases
Maxp1, NORE1, NORE1A, NORE1B, RAPL, RASSF3
UniGene
Hs.497579
Gene Chromosome Location
Chr.1: 206507530-206589284 on Build GRCh38
Transcripts
NM_182663.3 NM_182664.3 NM_182665.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1927465 c.7G>A p.V3M Chr.1: 206557580-206557580 on Build GRCh38
36818 c.48G>A p.E16E Chr.1: 206557621-206557621 on Build GRCh38
1172100 c.31T>G p.Y11D Chr.1: 206557604-206557604 on Build GRCh38
6449206 c.32A>G p.Y11C Chr.1: 206557605-206557605 on Build GRCh38
1263862 c.10G>A p.D4N Chr.1: 206557583-206557583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031851824 Chr.1: 206557803-206557803 on Build GRCh38
rs782048681 Chr.1: 206557624-206557624 on Build GRCh38
rs782276943 Chr.1: 206557532-206557532 on Build GRCh38
rs112324473 Chr.1: 206557721-206557721 on Build GRCh38
rs782345773 Chr.1: 206557555-206557555 on Build GRCh38
rs535225587 Chr.1: 206557763-206557763 on Build GRCh38
rs782113842 Chr.1: 206557586-206557586 on Build GRCh38
rs782523796 Chr.1: 206557693-206557693 on Build GRCh38
rs149369928 Chr.1: 206557791-206557791 on Build GRCh38
rs941241170 Chr.1: 206557607-206557607 on Build GRCh38
rs909914004 Chr.1: 206557572-206557572 on Build GRCh38
rs782496235 Chr.1: 206557702-206557702 on Build GRCh38
rs782668776 Chr.1: 206557531-206557531 on Build GRCh38
rs781825252 Chr.1: 206557656-206557656 on Build GRCh38
rs897081405 Chr.1: 206557637-206557637 on Build GRCh38
rs267598332 Chr.1: 206557621-206557621 on Build GRCh38
rs373175972 Chr.1: 206557525-206557525 on Build GRCh38
rs781931729 Chr.1: 206557618-206557618 on Build GRCh38
rs557046579 Chr.1: 206557884-206557884 on Build GRCh38
rs782277990 Chr.1: 206557726-206557726 on Build GRCh38
rs968927244 Chr.1: 206557505-206557505 on Build GRCh38
rs782745953 Chr.1: 206557676-206557676 on Build GRCh38
rs557045537 Chr.1: 206557862-206557862 on Build GRCh38
rs575145537 Chr.1: 206557828-206557828 on Build GRCh38
rs781930220 Chr.1: 206557736-206557736 on Build GRCh38
rs186212530 Chr.1: 206557927-206557927 on Build GRCh38
rs782389622 Chr.1: 206557729-206557729 on Build GRCh38
rs782239625 Chr.1: 206557553-206557553 on Build GRCh38
rs1036927921 Chr.1: 206557625-206557625 on Build GRCh38
rs782609162 Chr.1: 206557523-206557523 on Build GRCh38
rs917461067 Chr.1: 206557829-206557829 on Build GRCh38
rs782747121 Chr.1: 206557628-206557628 on Build GRCh38
rs782280839 Chr.1: 206557530-206557530 on Build GRCh38
rs1044784340 Chr.1: 206557567-206557567 on Build GRCh38
rs782494932 Chr.1: 206557727-206557728 on Build GRCh38
rs548126357 Chr.1: 206557515-206557515 on Build GRCh38
rs566301613 Chr.1: 206557755-206557755 on Build GRCh38
rs782734291 Chr.1: 206557695-206557695 on Build GRCh38
rs998386774 Chr.1: 206557710-206557710 on Build GRCh38
rs782002567 Chr.1: 206557562-206557562 on Build GRCh38
rs782264072 Chr.1: 206557714-206557714 on Build GRCh38
rs181919601 Chr.1: 206557886-206557886 on Build GRCh38
rs781974231 Chr.1: 206557730-206557730 on Build GRCh38
rs941314791 Chr.1: 206557943-206557943 on Build GRCh38
rs782780335 Chr.1: 206557722-206557723 on Build GRCh38
rs199814122 Chr.1: 206557549-206557549 on Build GRCh38
rs956320478 Chr.1: 206557809-206557809 on Build GRCh38
rs1021948327 Chr.1: 206557488-206557488 on Build GRCh38
rs201031974 Chr.1: 206557595-206557595 on Build GRCh38
rs781799398 Chr.1: 206557700-206557700 on Build GRCh38
rs190811411 Chr.1: 206557832-206557832 on Build GRCh38
rs782608734 Chr.1: 206557703-206557703 on Build GRCh38
rs781870698 Chr.1: 206557723-206557724 on Build GRCh38
rs781899492 Chr.1: 206557691-206557691 on Build GRCh38
rs112833004 Chr.1: 206557732-206557732 on Build GRCh38
rs373288171 Chr.1: 206557733-206557733 on Build GRCh38
rs147521013 Chr.1: 206557585-206557585 on Build GRCh38
rs782126968 Chr.1: 206557671-206557671 on Build GRCh38
rs782436172 Chr.1: 206557608-206557608 on Build GRCh38
rs782515055 Chr.1: 206557720-206557720 on Build GRCh38

View Full Product Details