Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236548714-236549226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR1
Entrez Gene ID
55127
Gene Name
HEAT repeat containing 1
Gene Aliases
BAP28, UTP10
UniGene
Hs.708114
Gene Chromosome Location
Chr.1: 236549005-236604541 on Build GRCh38
Transcripts
NM_018072.5 NM_006499.4 NM_201543.2 NM_201544.2 NM_201545.2

Gene Symbol
LGALS8
Entrez Gene ID
3964
Gene Name
galectin 8
Gene Aliases
Gal-8, PCTA-1, PCTA1, Po66-CBP
UniGene
Hs.4082
Gene Chromosome Location
Chr.1: 236518214-236552981 on Build GRCh38
Transcripts
NM_018072.5 NM_006499.4 NM_201543.2 NM_201544.2 NM_201545.2

SNP Detail

SNP ID Chromosome Location
rs150029148 Chr.1: 236548775-236548778 on Build GRCh38
rs535572535 Chr.1: 236549148-236549148 on Build GRCh38
rs917242970 Chr.1: 236548974-236548974 on Build GRCh38
rs1050573161 Chr.1: 236549037-236549037 on Build GRCh38
rs144176241 Chr.1: 236548795-236548795 on Build GRCh38
rs912983717 Chr.1: 236548828-236548828 on Build GRCh38
rs568135333 Chr.1: 236549081-236549081 on Build GRCh38
rs77856957 Chr.1: 236549021-236549021 on Build GRCh38
rs747812016 Chr.1: 236548774-236548775 on Build GRCh38
rs183565332 Chr.1: 236548966-236548966 on Build GRCh38
rs749512352 Chr.1: 236548995-236548995 on Build GRCh38
rs1045148139 Chr.1: 236549062-236549062 on Build GRCh38
rs561998674 Chr.1: 236549202-236549202 on Build GRCh38
rs781055506 Chr.1: 236548927-236548927 on Build GRCh38
rs779198509 Chr.1: 236549170-236549170 on Build GRCh38
rs1000831805 Chr.1: 236549098-236549098 on Build GRCh38
rs998157494 Chr.1: 236549173-236549173 on Build GRCh38
rs1038008921 Chr.1: 236548979-236548979 on Build GRCh38
rs866128273 Chr.1: 236548801-236548801 on Build GRCh38
rs1009898337 Chr.1: 236549162-236549162 on Build GRCh38
rs976395878 Chr.1: 236548956-236548957 on Build GRCh38
rs561804394 Chr.1: 236548807-236548807 on Build GRCh38
rs746162309 Chr.1: 236549182-236549182 on Build GRCh38
rs940992330 Chr.1: 236548978-236548978 on Build GRCh38
rs187119597 Chr.1: 236548972-236548972 on Build GRCh38
rs937272536 Chr.1: 236548971-236548971 on Build GRCh38
rs754809537 Chr.1: 236548755-236548755 on Build GRCh38
rs549818738 Chr.1: 236549033-236549033 on Build GRCh38
rs889262534 Chr.1: 236549052-236549052 on Build GRCh38
rs531298834 Chr.1: 236549028-236549028 on Build GRCh38
rs1023753151 Chr.1: 236548845-236548845 on Build GRCh38
rs1031216752 Chr.1: 236548934-236548934 on Build GRCh38
rs1020072969 Chr.1: 236549167-236549167 on Build GRCh38
rs965040307 Chr.1: 236548784-236548784 on Build GRCh38
rs2758200 Chr.1: 236548841-236548841 on Build GRCh38
rs1019153833 Chr.1: 236549086-236549086 on Build GRCh38
rs896360114 Chr.1: 236548829-236548829 on Build GRCh38
rs77196793 Chr.1: 236548778-236548778 on Build GRCh38
rs144850744 Chr.1: 236548973-236548973 on Build GRCh38
rs957185170 Chr.1: 236548938-236548938 on Build GRCh38
rs1030999620 Chr.1: 236549117-236549117 on Build GRCh38
rs541594301 Chr.1: 236548970-236548970 on Build GRCh38
rs148576474 Chr.1: 236548999-236548999 on Build GRCh38
rs1019711641 Chr.1: 236548748-236548748 on Build GRCh38
rs1012728928 Chr.1: 236548836-236548836 on Build GRCh38
rs576428042 Chr.1: 236548785-236548785 on Build GRCh38
rs968573175 Chr.1: 236548958-236548958 on Build GRCh38
rs10534773 Chr.1: 236548778-236548781 on Build GRCh38
rs1056409386 Chr.1: 236549011-236549011 on Build GRCh38
rs896298493 Chr.1: 236549016-236549017 on Build GRCh38
rs1009570791 Chr.1: 236549077-236549077 on Build GRCh38
rs968010180 Chr.1: 236548914-236548914 on Build GRCh38
rs78333332 Chr.1: 236548993-236548993 on Build GRCh38
rs903938808 Chr.1: 236549093-236549093 on Build GRCh38

View Full Product Details