Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247450951-247451455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2B11
Entrez Gene ID
127623
Gene Name
olfactory receptor family 2 subfamily B member 11
Gene Aliases
-
UniGene
Hs.446999
Gene Chromosome Location
Chr.1: 247451029-247451982 on Build GRCh38
Transcripts
NM_001004492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2232038 c.589G>A p.A197T Chr.1: 247451394-247451394 on Build GRCh38
4235486 c.616C>T p.L206L Chr.1: 247451367-247451367 on Build GRCh38
4382732 c.776C>T p.A259V Chr.1: 247451207-247451207 on Build GRCh38
5536078 c.760C>T p.L254F Chr.1: 247451223-247451223 on Build GRCh38
367667 c.552G>T p.E184D Chr.1: 247451431-247451431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199665492 Chr.1: 247451353-247451353 on Build GRCh38
rs745322436 Chr.1: 247451147-247451147 on Build GRCh38
rs141064927 Chr.1: 247451252-247451252 on Build GRCh38
rs769112106 Chr.1: 247451234-247451234 on Build GRCh38
rs754877720 Chr.1: 247451104-247451104 on Build GRCh38
rs767594904 Chr.1: 247451431-247451431 on Build GRCh38
rs922034015 Chr.1: 247451181-247451181 on Build GRCh38
rs966393569 Chr.1: 247451295-247451295 on Build GRCh38
rs755752355 Chr.1: 247451112-247451112 on Build GRCh38
rs758187486 Chr.1: 247451328-247451328 on Build GRCh38
rs756533262 Chr.1: 247451394-247451394 on Build GRCh38
rs771240286 Chr.1: 247451341-247451341 on Build GRCh38
rs149603815 Chr.1: 247451163-247451163 on Build GRCh38
rs780372908 Chr.1: 247451230-247451230 on Build GRCh38
rs202210424 Chr.1: 247451074-247451074 on Build GRCh38
rs141488287 Chr.1: 247451389-247451389 on Build GRCh38
rs748966978 Chr.1: 247451013-247451013 on Build GRCh38
rs769864730 Chr.1: 247451121-247451122 on Build GRCh38
rs563361078 Chr.1: 247451209-247451209 on Build GRCh38
rs552689091 Chr.1: 247450992-247450992 on Build GRCh38
rs148864378 Chr.1: 247451101-247451101 on Build GRCh38
rs1029939794 Chr.1: 247451256-247451256 on Build GRCh38
rs12070953 Chr.1: 247451084-247451084 on Build GRCh38
rs12065526 Chr.1: 247451105-247451105 on Build GRCh38
rs776237447 Chr.1: 247451422-247451422 on Build GRCh38
rs1036956700 Chr.1: 247451239-247451239 on Build GRCh38
rs74583618 Chr.1: 247451304-247451304 on Build GRCh38
rs761449331 Chr.1: 247451282-247451282 on Build GRCh38
rs12086048 Chr.1: 247451251-247451251 on Build GRCh38
rs752668750 Chr.1: 247451107-247451107 on Build GRCh38
rs746183122 Chr.1: 247451129-247451129 on Build GRCh38
rs376330488 Chr.1: 247451132-247451132 on Build GRCh38
rs1037042590 Chr.1: 247451245-247451245 on Build GRCh38
rs776723686 Chr.1: 247451166-247451166 on Build GRCh38
rs764029001 Chr.1: 247451278-247451278 on Build GRCh38
rs866290262 Chr.1: 247451180-247451180 on Build GRCh38
rs775557634 Chr.1: 247451063-247451063 on Build GRCh38
rs779179137 Chr.1: 247451121-247451121 on Build GRCh38
rs778682038 Chr.1: 247451221-247451221 on Build GRCh38
rs766639771 Chr.1: 247451395-247451395 on Build GRCh38
rs772045250 Chr.1: 247451259-247451259 on Build GRCh38
rs138609579 Chr.1: 247451383-247451383 on Build GRCh38
rs143103868 Chr.1: 247451427-247451427 on Build GRCh38
rs998415094 Chr.1: 247451110-247451110 on Build GRCh38
rs970577243 Chr.1: 247450977-247450977 on Build GRCh38
rs777626988 Chr.1: 247451007-247451007 on Build GRCh38
rs4925663 Chr.1: 247451315-247451315 on Build GRCh38
rs140643495 Chr.1: 247451428-247451428 on Build GRCh38
rs772666502 Chr.1: 247451365-247451365 on Build GRCh38
rs750035161 Chr.1: 247451223-247451223 on Build GRCh38
rs762507891 Chr.1: 247451430-247451430 on Build GRCh38
rs201253456 Chr.1: 247451260-247451260 on Build GRCh38
rs746386655 Chr.1: 247451048-247451048 on Build GRCh38
rs368268590 Chr.1: 247451229-247451229 on Build GRCh38
rs939236650 Chr.1: 247451412-247451412 on Build GRCh38
rs150630761 Chr.1: 247451254-247451254 on Build GRCh38
rs772594367 Chr.1: 247451051-247451051 on Build GRCh38
rs747596995 Chr.1: 247450980-247450980 on Build GRCh38
rs374651468 Chr.1: 247451326-247451326 on Build GRCh38
rs746052771 Chr.1: 247451419-247451419 on Build GRCh38
rs200390008 Chr.1: 247451198-247451198 on Build GRCh38
rs113868131 Chr.1: 247451145-247451145 on Build GRCh38
rs766108420 Chr.1: 247451096-247451096 on Build GRCh38
rs753702882 Chr.1: 247451280-247451280 on Build GRCh38
rs569352082 Chr.1: 247451352-247451352 on Build GRCh38
rs959084651 Chr.1: 247451025-247451025 on Build GRCh38
rs372166908 Chr.1: 247451413-247451413 on Build GRCh38
rs756887640 Chr.1: 247451120-247451120 on Build GRCh38
rs781136061 Chr.1: 247451237-247451237 on Build GRCh38
rs6695302 Chr.1: 247451391-247451391 on Build GRCh38
rs765958164 Chr.1: 247451372-247451372 on Build GRCh38
rs755403448 Chr.1: 247451382-247451382 on Build GRCh38
rs777942359 Chr.1: 247451397-247451397 on Build GRCh38
rs77553834 Chr.1: 247451115-247451115 on Build GRCh38
rs755846678 Chr.1: 247451003-247451003 on Build GRCh38
rs912097071 Chr.1: 247451161-247451161 on Build GRCh38
rs368167912 Chr.1: 247451206-247451206 on Build GRCh38
rs941388191 Chr.1: 247451241-247451244 on Build GRCh38
rs764823291 Chr.1: 247451362-247451362 on Build GRCh38
rs553020154 Chr.1: 247451426-247451426 on Build GRCh38
rs1049076706 Chr.1: 247451210-247451210 on Build GRCh38
rs780601371 Chr.1: 247451158-247451158 on Build GRCh38
rs145019591 Chr.1: 247451271-247451271 on Build GRCh38
rs763722180 Chr.1: 247451212-247451212 on Build GRCh38
rs370314539 Chr.1: 247451175-247451175 on Build GRCh38
rs138984792 Chr.1: 247451343-247451343 on Build GRCh38
rs756868155 Chr.1: 247451217-247451217 on Build GRCh38
rs781752543 Chr.1: 247451307-247451307 on Build GRCh38
rs751199940 Chr.1: 247451373-247451373 on Build GRCh38
rs755487668 Chr.1: 247451306-247451306 on Build GRCh38
rs879391227 Chr.1: 247450983-247450983 on Build GRCh38
rs771243297 Chr.1: 247451028-247451028 on Build GRCh38
rs757995115 Chr.1: 247451300-247451300 on Build GRCh38
rs150244239 Chr.1: 247451291-247451291 on Build GRCh38
rs148369409 Chr.1: 247451214-247451214 on Build GRCh38
rs780425804 Chr.1: 247451303-247451303 on Build GRCh38
rs149870519 Chr.1: 247451425-247451425 on Build GRCh38
rs748202669 Chr.1: 247451240-247451240 on Build GRCh38
rs534641794 Chr.1: 247451408-247451408 on Build GRCh38
rs200299205 Chr.1: 247451162-247451162 on Build GRCh38
rs977774699 Chr.1: 247451324-247451326 on Build GRCh38
rs865850308 Chr.1: 247451225-247451225 on Build GRCh38
rs200000820 Chr.1: 247451410-247451410 on Build GRCh38
rs769990807 Chr.1: 247451183-247451183 on Build GRCh38
rs73140586 Chr.1: 247451355-247451355 on Build GRCh38
rs201556264 Chr.1: 247451369-247451369 on Build GRCh38
rs147839044 Chr.1: 247451421-247451421 on Build GRCh38
rs764151335 Chr.1: 247451083-247451083 on Build GRCh38
rs367979713 Chr.1: 247451407-247451407 on Build GRCh38
rs748598350 Chr.1: 247451311-247451311 on Build GRCh38
rs764619184 Chr.1: 247451286-247451286 on Build GRCh38
rs748047379 Chr.1: 247451061-247451062 on Build GRCh38
rs749022491 Chr.1: 247450975-247450975 on Build GRCh38
rs1003751173 Chr.1: 247451002-247451002 on Build GRCh38
rs769171531 Chr.1: 247451207-247451207 on Build GRCh38
rs774760268 Chr.1: 247451035-247451035 on Build GRCh38
rs141217332 Chr.1: 247451118-247451118 on Build GRCh38
rs777506166 Chr.1: 247451122-247451122 on Build GRCh38
rs939277811 Chr.1: 247451187-247451187 on Build GRCh38
rs966667457 Chr.1: 247451342-247451342 on Build GRCh38
rs200507462 Chr.1: 247451077-247451077 on Build GRCh38
rs868431495 Chr.1: 247451080-247451080 on Build GRCh38
rs149924080 Chr.1: 247451270-247451270 on Build GRCh38
rs1033482031 Chr.1: 247451263-247451263 on Build GRCh38
rs774030305 Chr.1: 247451186-247451186 on Build GRCh38

View Full Product Details