Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70139599-70140094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYSND1
Entrez Gene ID
219743
Gene Name
trypsin domain containing 1
Gene Aliases
NET41
UniGene
Hs.533655
Gene Chromosome Location
Chr.10: 70137981-70146740 on Build GRCh38
Transcripts
NM_001040273.2 NM_173555.3 NR_073580.1 NR_073581.1 NR_073582.1 NR_073591.1 NR_073592.1 NR_073593.1 NR_073594.1 NR_073595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7152572 c.1666C>T p.R556W Chr.10: 70139959-70139959 on Build GRCh38
5678065 c.1603C>G p.L535V Chr.10: 70140022-70140022 on Build GRCh38
4738550 c.1577T>C p.L526P Chr.10: 70140048-70140048 on Build GRCh38
1231484 c.1630G>A p.A544T Chr.10: 70139995-70139995 on Build GRCh38
386286 c.1633G>T p.A545S Chr.10: 70139992-70139992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748321500 Chr.10: 70140014-70140014 on Build GRCh38
rs1022119664 Chr.10: 70139890-70139890 on Build GRCh38
rs779591485 Chr.10: 70139920-70139920 on Build GRCh38
rs755270667 Chr.10: 70140010-70140010 on Build GRCh38
rs1019275526 Chr.10: 70140035-70140035 on Build GRCh38
rs760546072 Chr.10: 70140050-70140050 on Build GRCh38
rs755360645 Chr.10: 70140073-70140073 on Build GRCh38
rs912304713 Chr.10: 70139781-70139781 on Build GRCh38
rs977835118 Chr.10: 70140055-70140055 on Build GRCh38
rs775236896 Chr.10: 70140044-70140044 on Build GRCh38
rs566446925 Chr.10: 70139796-70139796 on Build GRCh38
rs370786831 Chr.10: 70139983-70139983 on Build GRCh38
rs377549834 Chr.10: 70139938-70139938 on Build GRCh38
rs745863815 Chr.10: 70140030-70140030 on Build GRCh38
rs750906539 Chr.10: 70139935-70139935 on Build GRCh38
rs147866017 Chr.10: 70139880-70139880 on Build GRCh38
rs1032120159 Chr.10: 70140041-70140041 on Build GRCh38
rs762310351 Chr.10: 70140070-70140070 on Build GRCh38
rs761426307 Chr.10: 70139995-70139995 on Build GRCh38
rs181240600 Chr.10: 70139754-70139754 on Build GRCh38
rs752344565 Chr.10: 70139875-70139875 on Build GRCh38
rs76914789 Chr.10: 70139940-70139940 on Build GRCh38
rs751972246 Chr.10: 70140072-70140072 on Build GRCh38
rs953855849 Chr.10: 70139778-70139778 on Build GRCh38
rs775355576 Chr.10: 70139980-70139980 on Build GRCh38
rs145992545 Chr.10: 70139978-70139978 on Build GRCh38
rs764906341 Chr.10: 70140064-70140064 on Build GRCh38
rs1054464961 Chr.10: 70139944-70139944 on Build GRCh38
rs780433703 Chr.10: 70139939-70139939 on Build GRCh38
rs762635076 Chr.10: 70140032-70140032 on Build GRCh38
rs372019414 Chr.10: 70139782-70139782 on Build GRCh38
rs1012226722 Chr.10: 70139658-70139658 on Build GRCh38
rs894549058 Chr.10: 70139730-70139730 on Build GRCh38
rs766912909 Chr.10: 70140071-70140071 on Build GRCh38
rs768908211 Chr.10: 70139948-70139948 on Build GRCh38
rs61748802 Chr.10: 70140038-70140038 on Build GRCh38
rs753498194 Chr.10: 70139990-70139990 on Build GRCh38
rs148744638 Chr.10: 70140009-70140009 on Build GRCh38
rs149776228 Chr.10: 70139728-70139728 on Build GRCh38
rs769911833 Chr.10: 70140020-70140020 on Build GRCh38
rs756757452 Chr.10: 70139912-70139912 on Build GRCh38
rs764814307 Chr.10: 70139996-70139996 on Build GRCh38
rs922312453 Chr.10: 70139703-70139703 on Build GRCh38
rs1015470076 Chr.10: 70139724-70139724 on Build GRCh38
rs761523688 Chr.10: 70140059-70140059 on Build GRCh38
rs989261863 Chr.10: 70139857-70139857 on Build GRCh38
rs531964675 Chr.10: 70139695-70139695 on Build GRCh38
rs151252509 Chr.10: 70140066-70140066 on Build GRCh38
rs965141769 Chr.10: 70139807-70139807 on Build GRCh38
rs963762782 Chr.10: 70139934-70139934 on Build GRCh38
rs758725668 Chr.10: 70139766-70139766 on Build GRCh38
rs941514317 Chr.10: 70139721-70139721 on Build GRCh38
rs770453370 Chr.10: 70139906-70139906 on Build GRCh38
rs200072087 Chr.10: 70139937-70139937 on Build GRCh38
rs371863879 Chr.10: 70139943-70139943 on Build GRCh38
rs375494960 Chr.10: 70140013-70140013 on Build GRCh38
rs550364642 Chr.10: 70139715-70139715 on Build GRCh38
rs61748803 Chr.10: 70140000-70140000 on Build GRCh38
rs759495882 Chr.10: 70139968-70139968 on Build GRCh38
rs564621500 Chr.10: 70140054-70140054 on Build GRCh38
rs916810893 Chr.10: 70139946-70139946 on Build GRCh38
rs906575610 Chr.10: 70139708-70139708 on Build GRCh38
rs748231956 Chr.10: 70139959-70139959 on Build GRCh38
rs753388027 Chr.10: 70139910-70139910 on Build GRCh38
rs79175235 Chr.10: 70139684-70139684 on Build GRCh38
rs61748804 Chr.10: 70139987-70139987 on Build GRCh38
rs774294361 Chr.10: 70139967-70139967 on Build GRCh38
rs924143305 Chr.10: 70139859-70139859 on Build GRCh38
rs956861718 Chr.10: 70139826-70139826 on Build GRCh38
rs1027324410 Chr.10: 70140003-70140003 on Build GRCh38
rs1030896504 Chr.10: 70139791-70139791 on Build GRCh38
rs547916571 Chr.10: 70139769-70139769 on Build GRCh38
rs750932595 Chr.10: 70139997-70139997 on Build GRCh38
rs763767085 Chr.10: 70139908-70139908 on Build GRCh38
rs781332731 Chr.10: 70139958-70139958 on Build GRCh38
rs141863700 Chr.10: 70139998-70139998 on Build GRCh38
rs985107379 Chr.10: 70139779-70139779 on Build GRCh38
rs974418772 Chr.10: 70139742-70139742 on Build GRCh38
rs777803234 Chr.10: 70140022-70140022 on Build GRCh38
rs80270209 Chr.10: 70139720-70139720 on Build GRCh38
rs537662254 Chr.10: 70139961-70139961 on Build GRCh38
rs377182465 Chr.10: 70139856-70139856 on Build GRCh38
rs1047275994 Chr.10: 70139713-70139713 on Build GRCh38
rs533675811 Chr.10: 70139831-70139831 on Build GRCh38

View Full Product Details