Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89383624-89384132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFIT1B
Entrez Gene ID
439996
Gene Name
interferon induced protein with tetratricopeptide repeats 1B
Gene Aliases
IFIT1L, bA149I23.6
UniGene
Hs.500491
Gene Chromosome Location
Chr.10: 89378056-89385205 on Build GRCh38
Transcripts
NM_001010987.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5371260 c.458_459GT>AA p.G153E Chr.10: 89383771-89383772 on Build GRCh38
6683829 c.465G>T p.K155N Chr.10: 89383778-89383778 on Build GRCh38
5425484 c.459T>A p.G153G Chr.10: 89383772-89383772 on Build GRCh38
3978889 c.784C>G p.R262G Chr.10: 89384097-89384097 on Build GRCh38
6683831 c.502C>A p.L168M Chr.10: 89383815-89383815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762070382 Chr.10: 89383873-89383873 on Build GRCh38
rs758792760 Chr.10: 89383935-89383935 on Build GRCh38
rs1062963 Chr.10: 89384077-89384077 on Build GRCh38
rs7072728 Chr.10: 89383876-89383876 on Build GRCh38
rs761591779 Chr.10: 89383805-89383805 on Build GRCh38
rs1062962 Chr.10: 89384064-89384064 on Build GRCh38
rs1031513135 Chr.10: 89383861-89383861 on Build GRCh38
rs978068691 Chr.10: 89383767-89383767 on Build GRCh38
rs767563469 Chr.10: 89383709-89383709 on Build GRCh38
rs772157106 Chr.10: 89383789-89383789 on Build GRCh38
rs749702479 Chr.10: 89383660-89383660 on Build GRCh38
rs536366720 Chr.10: 89383874-89383874 on Build GRCh38
rs304494 Chr.10: 89383772-89383772 on Build GRCh38
rs369472118 Chr.10: 89383813-89383813 on Build GRCh38
rs758674845 Chr.10: 89384030-89384030 on Build GRCh38
rs762591705 Chr.10: 89383968-89383968 on Build GRCh38
rs767791005 Chr.10: 89383872-89383872 on Build GRCh38
rs773132110 Chr.10: 89383882-89383882 on Build GRCh38
rs764241446 Chr.10: 89383798-89383798 on Build GRCh38
rs774921894 Chr.10: 89383776-89383776 on Build GRCh38
rs886189506 Chr.10: 89383651-89383651 on Build GRCh38
rs780621453 Chr.10: 89383860-89383860 on Build GRCh38
rs781680465 Chr.10: 89384055-89384055 on Build GRCh38
rs760243271 Chr.10: 89384007-89384007 on Build GRCh38
rs373013476 Chr.10: 89383830-89383830 on Build GRCh38
rs543130541 Chr.10: 89384086-89384086 on Build GRCh38
rs760097231 Chr.10: 89384101-89384101 on Build GRCh38
rs750985695 Chr.10: 89383835-89383835 on Build GRCh38
rs202170099 Chr.10: 89383750-89383750 on Build GRCh38
rs776841075 Chr.10: 89383800-89383800 on Build GRCh38
rs758308539 Chr.10: 89383826-89383826 on Build GRCh38
rs755783048 Chr.10: 89383920-89383920 on Build GRCh38
rs753590815 Chr.10: 89384016-89384016 on Build GRCh38
rs148786050 Chr.10: 89384074-89384074 on Build GRCh38
rs138882406 Chr.10: 89383865-89383865 on Build GRCh38
rs191011735 Chr.10: 89383847-89383847 on Build GRCh38
rs147758253 Chr.10: 89383926-89383926 on Build GRCh38
rs201077082 Chr.10: 89383857-89383857 on Build GRCh38
rs960791799 Chr.10: 89383790-89383790 on Build GRCh38
rs146689135 Chr.10: 89384049-89384049 on Build GRCh38
rs1062961 Chr.10: 89384050-89384050 on Build GRCh38
rs770613588 Chr.10: 89384085-89384085 on Build GRCh38
rs142531389 Chr.10: 89383947-89383947 on Build GRCh38
rs748260211 Chr.10: 89383956-89383956 on Build GRCh38
rs369800890 Chr.10: 89383754-89383754 on Build GRCh38
rs536601061 Chr.10: 89383880-89383880 on Build GRCh38
rs753578723 Chr.10: 89383722-89383722 on Build GRCh38
rs777855225 Chr.10: 89384068-89384068 on Build GRCh38
rs775389107 Chr.10: 89383796-89383796 on Build GRCh38
rs775756065 Chr.10: 89383710-89383710 on Build GRCh38
rs142964444 Chr.10: 89383875-89383875 on Build GRCh38
rs757912496 Chr.10: 89383760-89383760 on Build GRCh38
rs748168634 Chr.10: 89384058-89384058 on Build GRCh38
rs764329251 Chr.10: 89383720-89383720 on Build GRCh38
rs374200876 Chr.10: 89384106-89384106 on Build GRCh38
rs376214034 Chr.10: 89383974-89383974 on Build GRCh38
rs746271112 Chr.10: 89383856-89383856 on Build GRCh38
rs199669012 Chr.10: 89384098-89384098 on Build GRCh38
rs750352996 Chr.10: 89383820-89383820 on Build GRCh38
rs751550518 Chr.10: 89383878-89383878 on Build GRCh38
rs909362712 Chr.10: 89383745-89383745 on Build GRCh38
rs370101022 Chr.10: 89384104-89384104 on Build GRCh38
rs138241421 Chr.10: 89383891-89383891 on Build GRCh38
rs1030979188 Chr.10: 89383946-89383946 on Build GRCh38
rs147571645 Chr.10: 89383761-89383761 on Build GRCh38
rs199716259 Chr.10: 89384083-89384083 on Build GRCh38
rs772407661 Chr.10: 89383858-89383858 on Build GRCh38
rs113472557 Chr.10: 89384025-89384025 on Build GRCh38
rs780418889 Chr.10: 89383936-89383936 on Build GRCh38
rs373382761 Chr.10: 89384097-89384097 on Build GRCh38
rs774420284 Chr.10: 89383975-89383975 on Build GRCh38
rs766778806 Chr.10: 89384048-89384048 on Build GRCh38
rs749609629 Chr.10: 89383769-89383769 on Build GRCh38
rs759598005 Chr.10: 89383699-89383699 on Build GRCh38
rs781289875 Chr.10: 89383953-89383953 on Build GRCh38
rs754551476 Chr.10: 89383836-89383836 on Build GRCh38
rs765090733 Chr.10: 89383740-89383740 on Build GRCh38
rs142404774 Chr.10: 89384081-89384081 on Build GRCh38
rs148517080 Chr.10: 89383711-89383711 on Build GRCh38
rs201423042 Chr.10: 89383815-89383815 on Build GRCh38
rs767590809 Chr.10: 89383995-89383995 on Build GRCh38
rs769862207 Chr.10: 89383963-89383963 on Build GRCh38
rs148271216 Chr.10: 89383839-89383839 on Build GRCh38
rs866753126 Chr.10: 89383901-89383901 on Build GRCh38
rs751872884 Chr.10: 89384108-89384108 on Build GRCh38
rs752202646 Chr.10: 89383998-89383998 on Build GRCh38
rs140309299 Chr.10: 89383850-89383850 on Build GRCh38
rs531532009 Chr.10: 89383713-89383713 on Build GRCh38
rs757056673 Chr.10: 89383730-89383730 on Build GRCh38
rs763583940 Chr.10: 89384012-89384012 on Build GRCh38
rs141939113 Chr.10: 89383762-89383762 on Build GRCh38
rs948404442 Chr.10: 89384069-89384069 on Build GRCh38
rs751014638 Chr.10: 89383992-89383992 on Build GRCh38
rs916759248 Chr.10: 89383799-89383799 on Build GRCh38
rs765880730 Chr.10: 89383941-89383941 on Build GRCh38
rs760763316 Chr.10: 89383884-89383884 on Build GRCh38
rs762004870 Chr.10: 89383779-89383779 on Build GRCh38
rs746423448 Chr.10: 89383782-89383782 on Build GRCh38
rs73369750 Chr.10: 89383851-89383851 on Build GRCh38
rs777499902 Chr.10: 89383925-89383925 on Build GRCh38
rs771089871 Chr.10: 89383647-89383647 on Build GRCh38
rs781015137 Chr.10: 89383763-89383763 on Build GRCh38
rs183153471 Chr.10: 89383866-89383866 on Build GRCh38
rs763717028 Chr.10: 89384102-89384102 on Build GRCh38
rs12262020 Chr.10: 89383691-89383691 on Build GRCh38

View Full Product Details