Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 98392750-98393185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PYROXD2
Entrez Gene ID
84795
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
Gene Aliases
C10orf33, FP3420
UniGene
Hs.238303
Gene Chromosome Location
Chr.10: 98383565-98415221 on Build GRCh38
Transcripts
NM_032709.2 XM_011540293.2 XM_011540301.2 XM_017016835.1 XM_017016836.1 XM_017016837.1 XM_017016838.1 XM_017016839.1 XM_017016840.1 XM_017016841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5999350 c.927+8G>A p.? Chr.10: 98392934-98392934 on Build GRCh38
3433391 c.866C>T p.A289V Chr.10: 98393003-98393003 on Build GRCh38
3433390 c.897C>T p.T299T Chr.10: 98392972-98392972 on Build GRCh38
2057085 c.854G>A p.G285E Chr.10: 98393015-98393015 on Build GRCh38
2057081 c.907G>T p.A303S Chr.10: 98392962-98392962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028728501 Chr.10: 98392887-98392887 on Build GRCh38
rs139454799 Chr.10: 98392991-98392991 on Build GRCh38
rs565862311 Chr.10: 98392987-98392987 on Build GRCh38
rs892179497 Chr.10: 98393115-98393115 on Build GRCh38
rs763710091 Chr.10: 98393029-98393029 on Build GRCh38
rs774596569 Chr.10: 98392949-98392949 on Build GRCh38
rs1039630042 Chr.10: 98393034-98393034 on Build GRCh38
rs367994394 Chr.10: 98392977-98392977 on Build GRCh38
rs766761349 Chr.10: 98393022-98393022 on Build GRCh38
rs572261550 Chr.10: 98393101-98393101 on Build GRCh38
rs770713850 Chr.10: 98392901-98392901 on Build GRCh38
rs746951492 Chr.10: 98392872-98392872 on Build GRCh38
rs761148635 Chr.10: 98393015-98393015 on Build GRCh38
rs891876156 Chr.10: 98393155-98393155 on Build GRCh38
rs767895596 Chr.10: 98392967-98392967 on Build GRCh38
rs1009323707 Chr.10: 98393134-98393134 on Build GRCh38
rs891158518 Chr.10: 98392894-98392894 on Build GRCh38
rs374387844 Chr.10: 98393053-98393053 on Build GRCh38
rs754439453 Chr.10: 98392891-98392891 on Build GRCh38
rs1033673568 Chr.10: 98392955-98392955 on Build GRCh38
rs754916124 Chr.10: 98393039-98393039 on Build GRCh38
rs779815908 Chr.10: 98393045-98393045 on Build GRCh38
rs760905526 Chr.10: 98393119-98393119 on Build GRCh38
rs543707725 Chr.10: 98393163-98393163 on Build GRCh38
rs779509044 Chr.10: 98392954-98392954 on Build GRCh38
rs777248191 Chr.10: 98392986-98392986 on Build GRCh38
rs752123590 Chr.10: 98392981-98392981 on Build GRCh38
rs201557747 Chr.10: 98392868-98392868 on Build GRCh38
rs60153449 Chr.10: 98392803-98392803 on Build GRCh38
rs775549619 Chr.10: 98392995-98392995 on Build GRCh38
rs757963606 Chr.10: 98392985-98392985 on Build GRCh38
rs749270229 Chr.10: 98393065-98393065 on Build GRCh38
rs145164432 Chr.10: 98393031-98393031 on Build GRCh38
rs549061482 Chr.10: 98392922-98392922 on Build GRCh38
rs938413716 Chr.10: 98392786-98392786 on Build GRCh38
rs139584064 Chr.10: 98393012-98393012 on Build GRCh38
rs768565433 Chr.10: 98393088-98393088 on Build GRCh38
rs149406474 Chr.10: 98392958-98392958 on Build GRCh38
rs1011690003 Chr.10: 98393159-98393159 on Build GRCh38
rs770876332 Chr.10: 98392934-98392934 on Build GRCh38
rs370350320 Chr.10: 98392913-98392913 on Build GRCh38
rs1008241460 Chr.10: 98393128-98393128 on Build GRCh38
rs898691844 Chr.10: 98393055-98393055 on Build GRCh38
rs932878337 Chr.10: 98393068-98393068 on Build GRCh38
rs1000562093 Chr.10: 98392779-98392779 on Build GRCh38
rs773460892 Chr.10: 98393114-98393114 on Build GRCh38
rs940199096 Chr.10: 98392851-98392851 on Build GRCh38
rs150546939 Chr.10: 98392878-98392878 on Build GRCh38
rs370632716 Chr.10: 98393035-98393035 on Build GRCh38
rs200737429 Chr.10: 98392906-98392906 on Build GRCh38
rs772270891 Chr.10: 98393103-98393103 on Build GRCh38
rs781047555 Chr.10: 98393036-98393036 on Build GRCh38
rs766978479 Chr.10: 98392980-98392980 on Build GRCh38
rs768812481 Chr.10: 98393010-98393010 on Build GRCh38
rs771345773 Chr.10: 98393124-98393124 on Build GRCh38
rs769845311 Chr.10: 98392941-98392941 on Build GRCh38
rs980165865 Chr.10: 98392774-98392774 on Build GRCh38
rs188215241 Chr.10: 98392775-98392775 on Build GRCh38
rs1053156638 Chr.10: 98393129-98393129 on Build GRCh38
rs982998005 Chr.10: 98392999-98392999 on Build GRCh38
rs887248173 Chr.10: 98392842-98392842 on Build GRCh38
rs756060536 Chr.10: 98393076-98393077 on Build GRCh38
rs757059548 Chr.10: 98392990-98392990 on Build GRCh38
rs771644465 Chr.10: 98392806-98392807 on Build GRCh38
rs920924935 Chr.10: 98393047-98393047 on Build GRCh38
rs554216343 Chr.10: 98392943-98392943 on Build GRCh38
rs777528333 Chr.10: 98392920-98392920 on Build GRCh38
rs763153458 Chr.10: 98392946-98392946 on Build GRCh38
rs749299055 Chr.10: 98393006-98393006 on Build GRCh38
rs867294542 Chr.10: 98393007-98393007 on Build GRCh38
rs745731022 Chr.10: 98393054-98393056 on Build GRCh38
rs778303539 Chr.10: 98392904-98392904 on Build GRCh38
rs776941645 Chr.10: 98393135-98393135 on Build GRCh38
rs773564940 Chr.10: 98393013-98393013 on Build GRCh38
rs749166911 Chr.10: 98393023-98393023 on Build GRCh38
rs112160924 Chr.10: 98392935-98392935 on Build GRCh38
rs764202035 Chr.10: 98392948-98392948 on Build GRCh38
rs368360864 Chr.10: 98393100-98393100 on Build GRCh38
rs750876004 Chr.10: 98392973-98392973 on Build GRCh38
rs192159317 Chr.10: 98392925-98392925 on Build GRCh38
rs755898387 Chr.10: 98392852-98392852 on Build GRCh38
rs535112764 Chr.10: 98392936-98392936 on Build GRCh38

View Full Product Details