Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 537378-537871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC56
Entrez Gene ID
115399
Gene Name
leucine rich repeat containing 56
Gene Aliases
-
UniGene
Hs.567655
Gene Chromosome Location
Chr.11: 518967-554916 on Build GRCh38
Transcripts
NM_198075.3 XM_005252775.2 XM_005252776.2 XM_006718132.2 XM_011519875.2 XM_011519876.1 XM_011519877.2 XM_017017167.1 XM_017017168.1

SNP Detail

SNP ID Chromosome Location
rs933080514 Chr.11: 537430-537430 on Build GRCh38
rs148660518 Chr.11: 537567-537567 on Build GRCh38
rs904214068 Chr.11: 537820-537820 on Build GRCh38
rs748790813 Chr.11: 537645-537645 on Build GRCh38
rs1053343786 Chr.11: 537473-537473 on Build GRCh38
rs772921332 Chr.11: 537553-537553 on Build GRCh38
rs184968795 Chr.11: 537832-537832 on Build GRCh38
rs927626903 Chr.11: 537621-537621 on Build GRCh38
rs914935471 Chr.11: 537805-537805 on Build GRCh38
rs550477734 Chr.11: 537580-537580 on Build GRCh38
rs1028778242 Chr.11: 537758-537758 on Build GRCh38
rs564436228 Chr.11: 537742-537742 on Build GRCh38
rs559601504 Chr.11: 537541-537541 on Build GRCh38
rs375340021 Chr.11: 537681-537681 on Build GRCh38
rs987092334 Chr.11: 537448-537448 on Build GRCh38
rs1002996949 Chr.11: 537523-537523 on Build GRCh38
rs1057182917 Chr.11: 537718-537718 on Build GRCh38
rs995380581 Chr.11: 537417-537417 on Build GRCh38
rs953558513 Chr.11: 537653-537653 on Build GRCh38
rs61877795 Chr.11: 537502-537502 on Build GRCh38
rs1000373235 Chr.11: 537844-537844 on Build GRCh38
rs1041387597 Chr.11: 537795-537795 on Build GRCh38
rs373450962 Chr.11: 537637-537637 on Build GRCh38
rs904811872 Chr.11: 537618-537618 on Build GRCh38
rs934976308 Chr.11: 537589-537589 on Build GRCh38
rs996070932 Chr.11: 537754-537754 on Build GRCh38
rs115863335 Chr.11: 537507-537507 on Build GRCh38
rs374271384 Chr.11: 537440-537441 on Build GRCh38
rs987274980 Chr.11: 537572-537572 on Build GRCh38
rs1010355746 Chr.11: 537571-537571 on Build GRCh38
rs947648107 Chr.11: 537624-537624 on Build GRCh38
rs945717852 Chr.11: 537794-537794 on Build GRCh38
rs771004804 Chr.11: 537738-537738 on Build GRCh38
rs769295353 Chr.11: 537530-537530 on Build GRCh38
rs922006175 Chr.11: 537500-537500 on Build GRCh38
rs1001590700 Chr.11: 537432-537432 on Build GRCh38
rs1035340249 Chr.11: 537433-537433 on Build GRCh38
rs962065614 Chr.11: 537678-537678 on Build GRCh38
rs896953887 Chr.11: 537401-537401 on Build GRCh38
rs963281906 Chr.11: 537563-537563 on Build GRCh38
rs899073469 Chr.11: 537743-537743 on Build GRCh38
rs551847015 Chr.11: 537565-537565 on Build GRCh38
rs1048281931 Chr.11: 537558-537558 on Build GRCh38
rs978933239 Chr.11: 537488-537488 on Build GRCh38
rs911865974 Chr.11: 537504-537504 on Build GRCh38
rs1036950183 Chr.11: 537521-537521 on Build GRCh38
rs1056880410 Chr.11: 537780-537780 on Build GRCh38
rs112687793 Chr.11: 537469-537469 on Build GRCh38
rs974567967 Chr.11: 537407-537407 on Build GRCh38
rs927078577 Chr.11: 537761-537761 on Build GRCh38
rs931122546 Chr.11: 537557-537557 on Build GRCh38
rs376257909 Chr.11: 537569-537569 on Build GRCh38
rs1000301383 Chr.11: 537635-537635 on Build GRCh38
rs959988903 Chr.11: 537434-537434 on Build GRCh38
rs1029092282 Chr.11: 537650-537650 on Build GRCh38
rs747389358 Chr.11: 537462-537462 on Build GRCh38
rs938741168 Chr.11: 537692-537692 on Build GRCh38
rs181920602 Chr.11: 537513-537513 on Build GRCh38
rs1019488346 Chr.11: 537673-537673 on Build GRCh38
rs895588593 Chr.11: 537783-537783 on Build GRCh38
rs918490529 Chr.11: 537728-537728 on Build GRCh38
rs1016690209 Chr.11: 537739-537739 on Build GRCh38
rs954514667 Chr.11: 537765-537765 on Build GRCh38
rs1057292437 Chr.11: 537566-537566 on Build GRCh38
rs1036081745 Chr.11: 537527-537527 on Build GRCh38
rs112360405 Chr.11: 537466-537466 on Build GRCh38
rs200787111 Chr.11: 537770-537770 on Build GRCh38
rs543595493 Chr.11: 537501-537501 on Build GRCh38
rs577961755 Chr.11: 537484-537484 on Build GRCh38
rs552987920 Chr.11: 537828-537828 on Build GRCh38
rs528446656 Chr.11: 537559-537559 on Build GRCh38
rs572761826 Chr.11: 537418-537418 on Build GRCh38
rs989571193 Chr.11: 537595-537595 on Build GRCh38
rs968855164 Chr.11: 537481-537481 on Build GRCh38
rs987536375 Chr.11: 537655-537655 on Build GRCh38
rs905640762 Chr.11: 537423-537423 on Build GRCh38
rs533647552 Chr.11: 537624-537625 on Build GRCh38

View Full Product Details