Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1227527-1228018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC5B
Entrez Gene ID
727897
Gene Name
mucin 5B, oligomeric mucus/gel-forming
Gene Aliases
MG1, MUC-5B, MUC5, MUC9
UniGene
Hs.523395
Gene Chromosome Location
Chr.11: 1223065-1262176 on Build GRCh38
Transcripts
NM_002458.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1975360 c.669C>T p.N223N Chr.11: 1227676-1227676 on Build GRCh38
1215910 c.722C>T p.T241M Chr.11: 1227729-1227729 on Build GRCh38
4939960 c.756C>A p.A252A Chr.11: 1227763-1227763 on Build GRCh38
4939961 c.756C>A p.A252A Chr.11: 1227763-1227763 on Build GRCh38
6834929 c.751C>T p.P251S Chr.11: 1227758-1227758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761351446 Chr.11: 1227559-1227559 on Build GRCh38
rs556368529 Chr.11: 1227797-1227797 on Build GRCh38
rs936734291 Chr.11: 1227803-1227803 on Build GRCh38
rs970846839 Chr.11: 1227610-1227610 on Build GRCh38
rs370384874 Chr.11: 1227690-1227690 on Build GRCh38
rs551572926 Chr.11: 1227668-1227668 on Build GRCh38
rs982582607 Chr.11: 1227599-1227599 on Build GRCh38
rs553916093 Chr.11: 1227858-1227858 on Build GRCh38
rs555281324 Chr.11: 1227933-1227933 on Build GRCh38
rs986992018 Chr.11: 1227548-1227548 on Build GRCh38
rs761143345 Chr.11: 1227706-1227706 on Build GRCh38
rs1047827228 Chr.11: 1227733-1227733 on Build GRCh38
rs190771168 Chr.11: 1227778-1227778 on Build GRCh38
rs1010618665 Chr.11: 1227947-1227947 on Build GRCh38
rs976515023 Chr.11: 1227709-1227709 on Build GRCh38
rs370808078 Chr.11: 1227818-1227818 on Build GRCh38
rs916593678 Chr.11: 1227869-1227869 on Build GRCh38
rs942008757 Chr.11: 1227861-1227861 on Build GRCh38
rs999291951 Chr.11: 1227669-1227669 on Build GRCh38
rs1037983438 Chr.11: 1227553-1227553 on Build GRCh38
rs781471890 Chr.11: 1227774-1227774 on Build GRCh38
rs532958707 Chr.11: 1227655-1227655 on Build GRCh38
rs375187456 Chr.11: 1227783-1227783 on Build GRCh38
rs976649255 Chr.11: 1227807-1227807 on Build GRCh38
rs112851053 Chr.11: 1227766-1227766 on Build GRCh38
rs895304178 Chr.11: 1227555-1227555 on Build GRCh38
rs571742054 Chr.11: 1227819-1227819 on Build GRCh38
rs970644045 Chr.11: 1227955-1227955 on Build GRCh38
rs747988361 Chr.11: 1227650-1227650 on Build GRCh38
rs999662627 Chr.11: 1227785-1227785 on Build GRCh38
rs572317578 Chr.11: 1227913-1227913 on Build GRCh38
rs929862636 Chr.11: 1227595-1227595 on Build GRCh38
rs1052609396 Chr.11: 1227945-1227945 on Build GRCh38
rs2075855 Chr.11: 1227730-1227730 on Build GRCh38
rs981022979 Chr.11: 1227959-1227959 on Build GRCh38
rs1007713158 Chr.11: 1227719-1227719 on Build GRCh38
rs1029886164 Chr.11: 1227839-1227839 on Build GRCh38
rs777377205 Chr.11: 1227747-1227747 on Build GRCh38
rs767706508 Chr.11: 1227700-1227700 on Build GRCh38
rs573537880 Chr.11: 1227946-1227946 on Build GRCh38
rs890662047 Chr.11: 1227705-1227705 on Build GRCh38
rs775231661 Chr.11: 1227691-1227691 on Build GRCh38
rs752398820 Chr.11: 1227980-1227980 on Build GRCh38
rs746810803 Chr.11: 1227748-1227748 on Build GRCh38
rs948257690 Chr.11: 1227901-1227901 on Build GRCh38
rs753960470 Chr.11: 1227727-1227727 on Build GRCh38
rs745336157 Chr.11: 1227677-1227677 on Build GRCh38
rs765468012 Chr.11: 1227731-1227731 on Build GRCh38
rs147623221 Chr.11: 1227978-1227978 on Build GRCh38
rs74046271 Chr.11: 1227932-1227932 on Build GRCh38
rs372765890 Chr.11: 1227670-1227670 on Build GRCh38
rs752878889 Chr.11: 1227741-1227741 on Build GRCh38
rs113967601 Chr.11: 1227912-1227912 on Build GRCh38
rs911170152 Chr.11: 1227682-1227682 on Build GRCh38
rs56179538 Chr.11: 1227764-1227764 on Build GRCh38
rs982676867 Chr.11: 1227981-1227981 on Build GRCh38
rs377758054 Chr.11: 1227742-1227742 on Build GRCh38
rs34261330 Chr.11: 1227572-1227572 on Build GRCh38
rs1029609216 Chr.11: 1227979-1227979 on Build GRCh38
rs752933898 Chr.11: 1227825-1227825 on Build GRCh38
rs755260740 Chr.11: 1227625-1227625 on Build GRCh38
rs973365460 Chr.11: 1227862-1227862 on Build GRCh38
rs949214607 Chr.11: 1227639-1227639 on Build GRCh38
rs540273001 Chr.11: 1227963-1227963 on Build GRCh38
rs527543345 Chr.11: 1227760-1227760 on Build GRCh38
rs919855015 Chr.11: 1227557-1227557 on Build GRCh38
rs922467356 Chr.11: 1227712-1227712 on Build GRCh38
rs907986653 Chr.11: 1227857-1227857 on Build GRCh38
rs762298481 Chr.11: 1227694-1227694 on Build GRCh38
rs1024730528 Chr.11: 1227572-1227572 on Build GRCh38
rs951270758 Chr.11: 1227846-1227846 on Build GRCh38
rs1037950809 Chr.11: 1227759-1227759 on Build GRCh38
rs56274761 Chr.11: 1227958-1227958 on Build GRCh38
rs538772569 Chr.11: 1227827-1227827 on Build GRCh38
rs773781510 Chr.11: 1227704-1227704 on Build GRCh38
rs538278703 Chr.11: 1227794-1227794 on Build GRCh38
rs777076129 Chr.11: 1227801-1227801 on Build GRCh38
rs1017898300 Chr.11: 1227800-1227800 on Build GRCh38
rs768340355 Chr.11: 1227792-1227792 on Build GRCh38
rs1017865669 Chr.11: 1227962-1227962 on Build GRCh38
rs1018016006 Chr.11: 1227549-1227549 on Build GRCh38
rs963842902 Chr.11: 1227692-1227692 on Build GRCh38
rs766904961 Chr.11: 1227714-1227714 on Build GRCh38
rs753139720 Chr.11: 1227775-1227775 on Build GRCh38
rs779077787 Chr.11: 1227648-1227648 on Build GRCh38
rs951049594 Chr.11: 1227761-1227761 on Build GRCh38
rs562635105 Chr.11: 1227567-1227567 on Build GRCh38
rs781163769 Chr.11: 1227763-1227763 on Build GRCh38
rs1046665728 Chr.11: 1227911-1227911 on Build GRCh38
rs899584195 Chr.11: 1227919-1227919 on Build GRCh38
rs377531490 Chr.11: 1227676-1227676 on Build GRCh38
rs892729114 Chr.11: 1227678-1227678 on Build GRCh38
rs369516705 Chr.11: 1227664-1227664 on Build GRCh38
rs756531475 Chr.11: 1227847-1227847 on Build GRCh38
rs368290527 Chr.11: 1227793-1227793 on Build GRCh38

View Full Product Details