Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2269592-2270086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCL2
Entrez Gene ID
430
Gene Name
achaete-scute family bHLH transcription factor 2
Gene Aliases
ASH2, HASH2, MASH2, bHLHa45
UniGene
Hs.152475
Gene Chromosome Location
Chr.11: 2268498-2270952 on Build GRCh38
Transcripts
NM_005170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6567763 c.366G>A p.P122P Chr.11: 2269967-2269967 on Build GRCh38
6401688 c.418G>A p.A140T Chr.11: 2269915-2269915 on Build GRCh38
6395830 c.575G>T p.G192V Chr.11: 2269758-2269758 on Build GRCh38
6391898 c.508A>T p.S170C Chr.11: 2269825-2269825 on Build GRCh38
7329070 c.315G>T p.L105L Chr.11: 2270018-2270018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006119146 Chr.11: 2269804-2269804 on Build GRCh38
rs908504730 Chr.11: 2270033-2270033 on Build GRCh38
rs958209243 Chr.11: 2269862-2269862 on Build GRCh38
rs1048384715 Chr.11: 2270012-2270012 on Build GRCh38
rs929129306 Chr.11: 2269786-2269786 on Build GRCh38
rs1047537037 Chr.11: 2269945-2269945 on Build GRCh38
rs559140499 Chr.11: 2269651-2269651 on Build GRCh38
rs930554763 Chr.11: 2270006-2270006 on Build GRCh38
rs989125753 Chr.11: 2269626-2269626 on Build GRCh38
rs530856175 Chr.11: 2269929-2269929 on Build GRCh38
rs943060446 Chr.11: 2269720-2269720 on Build GRCh38
rs899863151 Chr.11: 2269684-2269684 on Build GRCh38
rs1033576596 Chr.11: 2269806-2269806 on Build GRCh38
rs989177609 Chr.11: 2269863-2269863 on Build GRCh38
rs755151474 Chr.11: 2269940-2269965 on Build GRCh38
rs199617658 Chr.11: 2269897-2269897 on Build GRCh38
rs1021039748 Chr.11: 2269879-2269879 on Build GRCh38
rs777405531 Chr.11: 2269807-2269807 on Build GRCh38
rs752446935 Chr.11: 2269769-2269769 on Build GRCh38
rs913680178 Chr.11: 2269630-2269630 on Build GRCh38
rs964688390 Chr.11: 2269885-2269885 on Build GRCh38
rs940131231 Chr.11: 2270043-2270043 on Build GRCh38
rs770434687 Chr.11: 2269822-2269822 on Build GRCh38
rs548309623 Chr.11: 2269922-2269922 on Build GRCh38
rs757830419 Chr.11: 2269775-2269775 on Build GRCh38
rs909027939 Chr.11: 2269952-2269952 on Build GRCh38
rs552532684 Chr.11: 2269935-2269935 on Build GRCh38
rs766105614 Chr.11: 2269735-2269735 on Build GRCh38
rs920540813 Chr.11: 2269988-2269988 on Build GRCh38
rs760610806 Chr.11: 2269710-2269710 on Build GRCh38
rs774424594 Chr.11: 2269905-2269905 on Build GRCh38
rs563419786 Chr.11: 2269923-2269923 on Build GRCh38
rs577073968 Chr.11: 2269615-2269615 on Build GRCh38
rs780516427 Chr.11: 2269829-2269829 on Build GRCh38
rs1047587557 Chr.11: 2269788-2269788 on Build GRCh38
rs936942796 Chr.11: 2269665-2269665 on Build GRCh38
rs750039631 Chr.11: 2269828-2269828 on Build GRCh38
rs1004665370 Chr.11: 2269765-2269765 on Build GRCh38
rs923006333 Chr.11: 2269932-2269932 on Build GRCh38
rs927484315 Chr.11: 2269658-2269658 on Build GRCh38
rs749698576 Chr.11: 2269839-2269839 on Build GRCh38
rs895110009 Chr.11: 2269808-2269808 on Build GRCh38
rs985679747 Chr.11: 2269877-2269877 on Build GRCh38
rs911744714 Chr.11: 2269948-2269948 on Build GRCh38
rs895068209 Chr.11: 2269983-2269983 on Build GRCh38
rs958261494 Chr.11: 2269841-2269841 on Build GRCh38
rs931337665 Chr.11: 2269686-2269686 on Build GRCh38
rs868494863 Chr.11: 2269639-2269639 on Build GRCh38
rs941881978 Chr.11: 2269964-2269964 on Build GRCh38
rs989795494 Chr.11: 2269692-2269692 on Build GRCh38
rs376558492 Chr.11: 2269965-2269965 on Build GRCh38
rs116549635 Chr.11: 2269820-2269820 on Build GRCh38
rs754589237 Chr.11: 2269752-2269752 on Build GRCh38
rs753659602 Chr.11: 2269747-2269747 on Build GRCh38
rs911677251 Chr.11: 2269653-2269653 on Build GRCh38
rs540919609 Chr.11: 2269638-2269638 on Build GRCh38
rs910194100 Chr.11: 2269698-2269698 on Build GRCh38
rs1033953387 Chr.11: 2269838-2269838 on Build GRCh38
rs911482274 Chr.11: 2269880-2269880 on Build GRCh38
rs1014003630 Chr.11: 2270053-2270053 on Build GRCh38
rs570710935 Chr.11: 2269958-2269958 on Build GRCh38
rs778401748 Chr.11: 2269766-2269766 on Build GRCh38
rs887682137 Chr.11: 2269797-2269797 on Build GRCh38
rs925525208 Chr.11: 2269664-2269664 on Build GRCh38
rs1000096186 Chr.11: 2269833-2269833 on Build GRCh38
rs887237728 Chr.11: 2269695-2269695 on Build GRCh38

View Full Product Details