Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36490157-36490664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF6
Entrez Gene ID
7189
Gene Name
TNF receptor associated factor 6
Gene Aliases
MGC:3310, RNF85
UniGene
Hs.444172
Gene Chromosome Location
Chr.11: 36483767-36510313 on Build GRCh38
Transcripts
NM_004620.3 NM_145803.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5014989 c.810G>T p.M270I Chr.11: 36490597-36490597 on Build GRCh38
926724 c.1032C>T p.I344I Chr.11: 36490375-36490375 on Build GRCh38
5014988 c.1204C>T p.R402C Chr.11: 36490203-36490203 on Build GRCh38
926723 c.1214A>G p.N405S Chr.11: 36490193-36490193 on Build GRCh38
3447041 c.1126C>T p.H376Y Chr.11: 36490281-36490281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771879519 Chr.11: 36490629-36490629 on Build GRCh38
rs78639425 Chr.11: 36490529-36490529 on Build GRCh38
rs751855450 Chr.11: 36490554-36490554 on Build GRCh38
rs139667746 Chr.11: 36490408-36490408 on Build GRCh38
rs146694009 Chr.11: 36490459-36490459 on Build GRCh38
rs539301101 Chr.11: 36490417-36490417 on Build GRCh38
rs201926351 Chr.11: 36490399-36490399 on Build GRCh38
rs781690437 Chr.11: 36490429-36490429 on Build GRCh38
rs746111041 Chr.11: 36490302-36490302 on Build GRCh38
rs148751232 Chr.11: 36490322-36490322 on Build GRCh38
rs187625425 Chr.11: 36490258-36490258 on Build GRCh38
rs772536661 Chr.11: 36490303-36490303 on Build GRCh38
rs143313499 Chr.11: 36490363-36490363 on Build GRCh38
rs748672638 Chr.11: 36490558-36490558 on Build GRCh38
rs755357439 Chr.11: 36490555-36490555 on Build GRCh38
rs773951368 Chr.11: 36490597-36490597 on Build GRCh38
rs765347176 Chr.11: 36490540-36490540 on Build GRCh38
rs766148455 Chr.11: 36490358-36490358 on Build GRCh38
rs141250243 Chr.11: 36490345-36490345 on Build GRCh38
rs750059919 Chr.11: 36490183-36490183 on Build GRCh38
rs751422527 Chr.11: 36490359-36490359 on Build GRCh38
rs900697253 Chr.11: 36490544-36490544 on Build GRCh38
rs776049763 Chr.11: 36490316-36490316 on Build GRCh38
rs548543739 Chr.11: 36490203-36490203 on Build GRCh38
rs759228997 Chr.11: 36490622-36490622 on Build GRCh38
rs759276140 Chr.11: 36490507-36490507 on Build GRCh38
rs149674427 Chr.11: 36490573-36490573 on Build GRCh38
rs147417401 Chr.11: 36490377-36490377 on Build GRCh38
rs780561325 Chr.11: 36490412-36490412 on Build GRCh38
rs144270884 Chr.11: 36490261-36490261 on Build GRCh38
rs781456089 Chr.11: 36490557-36490557 on Build GRCh38
rs145506174 Chr.11: 36490621-36490621 on Build GRCh38
rs776312804 Chr.11: 36490530-36490530 on Build GRCh38
rs762588981 Chr.11: 36490446-36490446 on Build GRCh38
rs963487647 Chr.11: 36490588-36490588 on Build GRCh38
rs535909443 Chr.11: 36490409-36490409 on Build GRCh38
rs1017786202 Chr.11: 36490492-36490492 on Build GRCh38
rs147645716 Chr.11: 36490551-36490551 on Build GRCh38
rs34165477 Chr.11: 36490221-36490222 on Build GRCh38
rs761520729 Chr.11: 36490543-36490548 on Build GRCh38
rs758560289 Chr.11: 36490547-36490547 on Build GRCh38
rs376933557 Chr.11: 36490563-36490563 on Build GRCh38
rs55840758 Chr.11: 36490571-36490571 on Build GRCh38
rs772654643 Chr.11: 36490441-36490441 on Build GRCh38
rs751366186 Chr.11: 36490327-36490327 on Build GRCh38
rs770500483 Chr.11: 36490467-36490467 on Build GRCh38
rs752651373 Chr.11: 36490528-36490528 on Build GRCh38
rs376266610 Chr.11: 36490326-36490326 on Build GRCh38
rs777821743 Chr.11: 36490252-36490252 on Build GRCh38
rs536779653 Chr.11: 36490294-36490294 on Build GRCh38
rs199830912 Chr.11: 36490202-36490202 on Build GRCh38
rs145275891 Chr.11: 36490432-36490432 on Build GRCh38
rs747509379 Chr.11: 36490321-36490321 on Build GRCh38
rs750597806 Chr.11: 36490545-36490545 on Build GRCh38
rs774002087 Chr.11: 36490480-36490480 on Build GRCh38
rs767244798 Chr.11: 36490512-36490512 on Build GRCh38
rs139449357 Chr.11: 36490564-36490564 on Build GRCh38
rs754966271 Chr.11: 36490231-36490231 on Build GRCh38
rs762641979 Chr.11: 36490349-36490349 on Build GRCh38
rs751475632 Chr.11: 36490213-36490213 on Build GRCh38
rs778902552 Chr.11: 36490407-36490407 on Build GRCh38
rs752595856 Chr.11: 36490386-36490386 on Build GRCh38
rs761330797 Chr.11: 36490331-36490331 on Build GRCh38

View Full Product Details