Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44260589-44261092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX4
Entrez Gene ID
60529
Gene Name
ALX homeobox 4
Gene Aliases
CRS5, FND2
UniGene
Hs.436055
Gene Chromosome Location
Chr.11: 44260728-44310166 on Build GRCh38
Transcripts
NM_021926.3

SNP Detail

SNP ID Chromosome Location
rs373059937 Chr.11: 44260911-44260911 on Build GRCh38
rs954376190 Chr.11: 44260819-44260819 on Build GRCh38
rs905705859 Chr.11: 44260872-44260872 on Build GRCh38
rs932128850 Chr.11: 44260709-44260709 on Build GRCh38
rs984906590 Chr.11: 44260719-44260719 on Build GRCh38
rs755986390 Chr.11: 44260978-44260978 on Build GRCh38
rs538499078 Chr.11: 44261029-44261029 on Build GRCh38
rs554409058 Chr.11: 44261070-44261070 on Build GRCh38
rs906517439 Chr.11: 44260647-44260647 on Build GRCh38
rs571701553 Chr.11: 44261012-44261012 on Build GRCh38
rs373307279 Chr.11: 44261049-44261049 on Build GRCh38
rs147497663 Chr.11: 44261062-44261062 on Build GRCh38
rs1020572536 Chr.11: 44260643-44260643 on Build GRCh38
rs988842130 Chr.11: 44260641-44260641 on Build GRCh38
rs1036770827 Chr.11: 44260764-44260764 on Build GRCh38
rs560087261 Chr.11: 44260749-44260749 on Build GRCh38
rs540638130 Chr.11: 44260717-44260717 on Build GRCh38
rs916748478 Chr.11: 44261009-44261009 on Build GRCh38
rs752581342 Chr.11: 44260867-44260867 on Build GRCh38
rs989764655 Chr.11: 44261008-44261008 on Build GRCh38
rs1009937911 Chr.11: 44261020-44261020 on Build GRCh38
rs897110837 Chr.11: 44260779-44260779 on Build GRCh38
rs34557775 Chr.11: 44260735-44260736 on Build GRCh38
rs369328027 Chr.11: 44260855-44260855 on Build GRCh38
rs924179957 Chr.11: 44260917-44260917 on Build GRCh38
rs528449092 Chr.11: 44260941-44260941 on Build GRCh38
rs778437510 Chr.11: 44260659-44260659 on Build GRCh38
rs892873917 Chr.11: 44260979-44260979 on Build GRCh38
rs531226738 Chr.11: 44260720-44260720 on Build GRCh38
rs976249932 Chr.11: 44260683-44260683 on Build GRCh38
rs936917265 Chr.11: 44260929-44260929 on Build GRCh38
rs1001739501 Chr.11: 44260949-44260949 on Build GRCh38
rs143536201 Chr.11: 44261042-44261042 on Build GRCh38
rs909255631 Chr.11: 44260736-44260736 on Build GRCh38
rs564010958 Chr.11: 44260627-44260627 on Build GRCh38
rs886373131 Chr.11: 44260710-44260710 on Build GRCh38
rs767299156 Chr.11: 44260814-44260814 on Build GRCh38
rs1032763125 Chr.11: 44260952-44260952 on Build GRCh38
rs532671795 Chr.11: 44260912-44260912 on Build GRCh38
rs1030516530 Chr.11: 44260760-44260760 on Build GRCh38
rs1005970390 Chr.11: 44260716-44260717 on Build GRCh38
rs796831292 Chr.11: 44260644-44260644 on Build GRCh38
rs978181962 Chr.11: 44260890-44260890 on Build GRCh38
rs759267585 Chr.11: 44260756-44260756 on Build GRCh38
rs886048283 Chr.11: 44260866-44260866 on Build GRCh38
rs765522183 Chr.11: 44260703-44260703 on Build GRCh38
rs546810144 Chr.11: 44260972-44260972 on Build GRCh38

View Full Product Details