Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119172229-119172765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRX1
Entrez Gene ID
79671
Gene Name
NLR family member X1
Gene Aliases
CLR11.3, DLNB26, NOD26, NOD5, NOD9
UniGene
Hs.524082
Gene Chromosome Location
Chr.11: 119168334-119184016 on Build GRCh38
Transcripts
NM_001282143.1 NM_001282144.1 NM_001282358.1 NM_024618.3 XM_005271669.1 XM_005271670.1 XM_005271671.1 XM_005271672.1 XM_006718904.1 XM_011542980.1 XM_017018294.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7425405 c.114A>T p.Q38H Chr.11: 119172399-119172399 on Build GRCh38
3444124 c.83C>T p.P28L Chr.11: 119172368-119172368 on Build GRCh38
110483 c.124C>T p.P42S Chr.11: 119172409-119172409 on Build GRCh38
7425406 c.114A>T p.Q38H Chr.11: 119172399-119172399 on Build GRCh38
4462759 c.125C>T p.P42L Chr.11: 119172410-119172410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015171527 Chr.11: 119172631-119172631 on Build GRCh38
rs778429968 Chr.11: 119172472-119172472 on Build GRCh38
rs759515461 Chr.11: 119172406-119172406 on Build GRCh38
rs571987792 Chr.11: 119172641-119172641 on Build GRCh38
rs1021489741 Chr.11: 119172732-119172732 on Build GRCh38
rs1056816055 Chr.11: 119172539-119172539 on Build GRCh38
rs151161124 Chr.11: 119172594-119172594 on Build GRCh38
rs372680679 Chr.11: 119172690-119172690 on Build GRCh38
rs777542512 Chr.11: 119172331-119172331 on Build GRCh38
rs373080354 Chr.11: 119172436-119172436 on Build GRCh38
rs776914089 Chr.11: 119172366-119172366 on Build GRCh38
rs991136394 Chr.11: 119172633-119172633 on Build GRCh38
rs760586955 Chr.11: 119172424-119172424 on Build GRCh38
rs199476036 Chr.11: 119172685-119172685 on Build GRCh38
rs938352460 Chr.11: 119172500-119172500 on Build GRCh38
rs770012581 Chr.11: 119172401-119172401 on Build GRCh38
rs58947677 Chr.11: 119172268-119172268 on Build GRCh38
rs1051333163 Chr.11: 119172524-119172524 on Build GRCh38
rs543104083 Chr.11: 119172585-119172585 on Build GRCh38
rs768753625 Chr.11: 119172364-119172364 on Build GRCh38
rs111941670 Chr.11: 119172544-119172544 on Build GRCh38
rs547167082 Chr.11: 119172621-119172621 on Build GRCh38
rs942715727 Chr.11: 119172551-119172551 on Build GRCh38
rs577488289 Chr.11: 119172445-119172445 on Build GRCh38
rs561361836 Chr.11: 119172586-119172586 on Build GRCh38
rs774391248 Chr.11: 119172405-119172405 on Build GRCh38
rs868294590 Chr.11: 119172503-119172503 on Build GRCh38
rs761922653 Chr.11: 119172383-119172383 on Build GRCh38
rs1003056725 Chr.11: 119172257-119172257 on Build GRCh38
rs919036529 Chr.11: 119172694-119172694 on Build GRCh38
rs371837882 Chr.11: 119172307-119172307 on Build GRCh38
rs201371895 Chr.11: 119172346-119172346 on Build GRCh38
rs924311356 Chr.11: 119172476-119172476 on Build GRCh38
rs142957206 Chr.11: 119172409-119172409 on Build GRCh38
rs375317314 Chr.11: 119172304-119172304 on Build GRCh38
rs903038871 Chr.11: 119172591-119172591 on Build GRCh38
rs569668529 Chr.11: 119172688-119172688 on Build GRCh38
rs539441331 Chr.11: 119172278-119172278 on Build GRCh38
rs563325829 Chr.11: 119172550-119172550 on Build GRCh38
rs775689434 Chr.11: 119172361-119172361 on Build GRCh38
rs551426776 Chr.11: 119172687-119172687 on Build GRCh38
rs537131759 Chr.11: 119172317-119172317 on Build GRCh38
rs958906235 Chr.11: 119172629-119172629 on Build GRCh38
rs113160059 Chr.11: 119172555-119172556 on Build GRCh38
rs1012334422 Chr.11: 119172298-119172298 on Build GRCh38
rs772389031 Chr.11: 119172358-119172358 on Build GRCh38
rs200678457 Chr.11: 119172557-119172557 on Build GRCh38
rs552852958 Chr.11: 119172439-119172439 on Build GRCh38
rs952270039 Chr.11: 119172303-119172303 on Build GRCh38
rs532841620 Chr.11: 119172684-119172684 on Build GRCh38
rs60612369 Chr.11: 119172362-119172362 on Build GRCh38
rs557754815 Chr.11: 119172285-119172285 on Build GRCh38
rs953195851 Chr.11: 119172367-119172367 on Build GRCh38
rs755452598 Chr.11: 119172536-119172536 on Build GRCh38
rs188616690 Chr.11: 119172410-119172410 on Build GRCh38
rs1030440699 Chr.11: 119172668-119172668 on Build GRCh38
rs997481031 Chr.11: 119172663-119172663 on Build GRCh38
rs967229978 Chr.11: 119172258-119172258 on Build GRCh38
rs893188458 Chr.11: 119172570-119172570 on Build GRCh38
rs902833322 Chr.11: 119172254-119172254 on Build GRCh38
rs545016598 Chr.11: 119172545-119172545 on Build GRCh38
rs1034097700 Chr.11: 119172739-119172739 on Build GRCh38
rs771369503 Chr.11: 119172315-119172315 on Build GRCh38
rs58592897 Chr.11: 119172556-119172557 on Build GRCh38
rs755698473 Chr.11: 119172319-119172319 on Build GRCh38
rs985939928 Chr.11: 119172389-119172389 on Build GRCh38
rs1023843657 Chr.11: 119172300-119172300 on Build GRCh38
rs748745248 Chr.11: 119172337-119172337 on Build GRCh38
rs752575821 Chr.11: 119172318-119172318 on Build GRCh38
rs752419860 Chr.11: 119172416-119172416 on Build GRCh38
rs887934394 Chr.11: 119172605-119172605 on Build GRCh38
rs754770235 Chr.11: 119172312-119172312 on Build GRCh38
rs555322970 Chr.11: 119172348-119172348 on Build GRCh38
rs769088403 Chr.11: 119172437-119172438 on Build GRCh38
rs1040145577 Chr.11: 119172564-119172564 on Build GRCh38
rs530937047 Chr.11: 119172581-119172581 on Build GRCh38
rs375464688 Chr.11: 119172311-119172311 on Build GRCh38

View Full Product Details