Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16989613-16990121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3849324 c.2592C>T p.C864C Chr.1: 16989708-16989708 on Build GRCh38
3849325 c.2460C>T p.C820C Chr.1: 16989708-16989708 on Build GRCh38
7356922 c.2413G>A p.D805N Chr.1: 16990003-16990003 on Build GRCh38
424620 c.2422C>T p.Q808* Chr.1: 16989994-16989994 on Build GRCh38
7324241 c.2593G>A p.E865K Chr.1: 16989707-16989707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766432992 Chr.1: 16989946-16989946 on Build GRCh38
rs761491667 Chr.1: 16990039-16990039 on Build GRCh38
rs761620417 Chr.1: 16989935-16989935 on Build GRCh38
rs755936102 Chr.1: 16990098-16990100 on Build GRCh38
rs986226126 Chr.1: 16989819-16989819 on Build GRCh38
rs747242869 Chr.1: 16989746-16989746 on Build GRCh38
rs905316433 Chr.1: 16989637-16989637 on Build GRCh38
rs587776890 Chr.1: 16989747-16989748 on Build GRCh38
rs763659305 Chr.1: 16989884-16989884 on Build GRCh38
rs765064192 Chr.1: 16989937-16989937 on Build GRCh38
rs77880760 Chr.1: 16990082-16990082 on Build GRCh38
rs147611385 Chr.1: 16989938-16989938 on Build GRCh38
rs745954026 Chr.1: 16989742-16989742 on Build GRCh38
rs761130666 Chr.1: 16989648-16989648 on Build GRCh38
rs537112220 Chr.1: 16989871-16989871 on Build GRCh38
rs148534162 Chr.1: 16989707-16989707 on Build GRCh38
rs1030482567 Chr.1: 16990050-16990050 on Build GRCh38
rs954971480 Chr.1: 16989800-16989800 on Build GRCh38
rs777472886 Chr.1: 16989708-16989708 on Build GRCh38
rs949240567 Chr.1: 16989810-16989810 on Build GRCh38
rs760318841 Chr.1: 16989882-16989882 on Build GRCh38
rs757153866 Chr.1: 16989841-16989841 on Build GRCh38
rs372376094 Chr.1: 16989965-16989965 on Build GRCh38
rs780146972 Chr.1: 16989854-16989854 on Build GRCh38
rs917176878 Chr.1: 16989891-16989891 on Build GRCh38
rs778229092 Chr.1: 16990087-16990087 on Build GRCh38
rs200809857 Chr.1: 16989878-16989878 on Build GRCh38
rs376237408 Chr.1: 16989955-16989955 on Build GRCh38
rs755157804 Chr.1: 16989957-16989957 on Build GRCh38
rs748463334 Chr.1: 16989869-16989869 on Build GRCh38
rs1037763231 Chr.1: 16990014-16990014 on Build GRCh38
rs376174719 Chr.1: 16989638-16989638 on Build GRCh38
rs563317561 Chr.1: 16989654-16989654 on Build GRCh38
rs766227206 Chr.1: 16990077-16990077 on Build GRCh38
rs778353567 Chr.1: 16989979-16989979 on Build GRCh38
rs747248855 Chr.1: 16989856-16989856 on Build GRCh38
rs897837480 Chr.1: 16989848-16989848 on Build GRCh38
rs535209471 Chr.1: 16989821-16989821 on Build GRCh38
rs954861510 Chr.1: 16989656-16989656 on Build GRCh38
rs771708290 Chr.1: 16989879-16989879 on Build GRCh38
rs374203880 Chr.1: 16990097-16990097 on Build GRCh38
rs776124539 Chr.1: 16990033-16990033 on Build GRCh38
rs866035312 Chr.1: 16989961-16989961 on Build GRCh38
rs759204814 Chr.1: 16989681-16989681 on Build GRCh38
rs751615609 Chr.1: 16990081-16990081 on Build GRCh38
rs763937012 Chr.1: 16989820-16989820 on Build GRCh38
rs781557299 Chr.1: 16989861-16989861 on Build GRCh38
rs774420241 Chr.1: 16989736-16989736 on Build GRCh38
rs773837281 Chr.1: 16989877-16989877 on Build GRCh38
rs765011219 Chr.1: 16990040-16990040 on Build GRCh38
rs751168600 Chr.1: 16989680-16989680 on Build GRCh38
rs941921003 Chr.1: 16990066-16990066 on Build GRCh38
rs566172222 Chr.1: 16989752-16989752 on Build GRCh38
rs774997717 Chr.1: 16989994-16989994 on Build GRCh38
rs886045579 Chr.1: 16989737-16989737 on Build GRCh38
rs374008623 Chr.1: 16989779-16989779 on Build GRCh38
rs55929813 Chr.1: 16989826-16989826 on Build GRCh38
rs765905631 Chr.1: 16989675-16989675 on Build GRCh38
rs759142662 Chr.1: 16989784-16989784 on Build GRCh38
rs994103230 Chr.1: 16990035-16990035 on Build GRCh38
rs868725919 Chr.1: 16989764-16989764 on Build GRCh38
rs368244226 Chr.1: 16989977-16989977 on Build GRCh38
rs781298587 Chr.1: 16989960-16989960 on Build GRCh38
rs768307347 Chr.1: 16990004-16990004 on Build GRCh38
rs778906228 Chr.1: 16989740-16989740 on Build GRCh38
rs773047666 Chr.1: 16990044-16990044 on Build GRCh38
rs756467036 Chr.1: 16990085-16990085 on Build GRCh38
rs778854959 Chr.1: 16989846-16989846 on Build GRCh38
rs752965372 Chr.1: 16990084-16990084 on Build GRCh38
rs377073826 Chr.1: 16989851-16989851 on Build GRCh38
rs373475292 Chr.1: 16989646-16989646 on Build GRCh38
rs541344554 Chr.1: 16990046-16990046 on Build GRCh38
rs772297257 Chr.1: 16989643-16989643 on Build GRCh38
rs749802732 Chr.1: 16989987-16989987 on Build GRCh38
rs756052051 Chr.1: 16990053-16990053 on Build GRCh38
rs746533808 Chr.1: 16990003-16990003 on Build GRCh38
rs760315801 Chr.1: 16989816-16989816 on Build GRCh38
rs769033175 Chr.1: 16989756-16989756 on Build GRCh38
rs771579345 Chr.1: 16989992-16989992 on Build GRCh38
rs375894941 Chr.1: 16989773-16989773 on Build GRCh38
rs1017231900 Chr.1: 16990083-16990083 on Build GRCh38
rs758449038 Chr.1: 16989853-16989853 on Build GRCh38
rs371083479 Chr.1: 16989860-16989860 on Build GRCh38
rs201321612 Chr.1: 16989825-16989825 on Build GRCh38
rs376087842 Chr.1: 16990052-16990052 on Build GRCh38
rs898009726 Chr.1: 16990017-16990017 on Build GRCh38
rs752494943 Chr.1: 16989701-16989701 on Build GRCh38
rs753813388 Chr.1: 16989719-16989719 on Build GRCh38
rs748454159 Chr.1: 16989976-16989976 on Build GRCh38
rs376194683 Chr.1: 16989655-16989655 on Build GRCh38
rs770434494 Chr.1: 16989776-16989776 on Build GRCh38
rs752367474 Chr.1: 16989795-16989795 on Build GRCh38
rs767492622 Chr.1: 16990079-16990086 on Build GRCh38
rs770381718 Chr.1: 16989876-16989876 on Build GRCh38
rs1045067591 Chr.1: 16989636-16989636 on Build GRCh38
rs587777053 Chr.1: 16989739-16989739 on Build GRCh38
rs776925680 Chr.1: 16989765-16989765 on Build GRCh38
rs774924482 Chr.1: 16989881-16989881 on Build GRCh38
rs772243999 Chr.1: 16989743-16989743 on Build GRCh38
rs776448394 Chr.1: 16989886-16989886 on Build GRCh38
rs55889979 Chr.1: 16989744-16989744 on Build GRCh38
rs986327732 Chr.1: 16989868-16989868 on Build GRCh38
rs905157690 Chr.1: 16989980-16989980 on Build GRCh38
rs370828598 Chr.1: 16989660-16989660 on Build GRCh38
rs935118855 Chr.1: 16989782-16989782 on Build GRCh38
rs767280251 Chr.1: 16989700-16989700 on Build GRCh38
rs750369195 Chr.1: 16990090-16990094 on Build GRCh38

View Full Product Details