Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78012782-78013308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D2B
Entrez Gene ID
23102
Gene Name
TBC1 domain family member 2B
Gene Aliases
-
UniGene
Hs.567426
Gene Chromosome Location
Chr.15: 77994985-78077708 on Build GRCh38
Transcripts
NM_015079.5 NM_144572.1 XM_011521387.2 XM_017022022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4477785 c.2183C>T p.T728I Chr.15: 78012910-78012910 on Build GRCh38
458954 c.1867C>G p.L623V Chr.15: 78013226-78013226 on Build GRCh38
6195019 c.1998G>A p.E666E Chr.15: 78013095-78013095 on Build GRCh38
3370269 c.2086C>T p.L696L Chr.15: 78013007-78013007 on Build GRCh38
4477784 c.2183C>T p.T728I Chr.15: 78012910-78012910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559426852 Chr.15: 78012966-78012966 on Build GRCh38
rs970444065 Chr.15: 78013144-78013144 on Build GRCh38
rs755446116 Chr.15: 78013097-78013097 on Build GRCh38
rs762894619 Chr.15: 78013066-78013066 on Build GRCh38
rs957757954 Chr.15: 78013207-78013207 on Build GRCh38
rs143151049 Chr.15: 78013252-78013252 on Build GRCh38
rs781029381 Chr.15: 78013111-78013111 on Build GRCh38
rs763924402 Chr.15: 78012993-78012993 on Build GRCh38
rs756184324 Chr.15: 78013029-78013029 on Build GRCh38
rs760327037 Chr.15: 78012945-78012945 on Build GRCh38
rs760656010 Chr.15: 78012976-78012976 on Build GRCh38
rs913797491 Chr.15: 78013250-78013250 on Build GRCh38
rs756992755 Chr.15: 78013082-78013082 on Build GRCh38
rs772558929 Chr.15: 78013107-78013107 on Build GRCh38
rs757438046 Chr.15: 78013009-78013009 on Build GRCh38
rs753849181 Chr.15: 78012842-78012842 on Build GRCh38
rs767279913 Chr.15: 78013166-78013166 on Build GRCh38
rs750087272 Chr.15: 78012813-78012813 on Build GRCh38
rs770231051 Chr.15: 78012909-78012909 on Build GRCh38
rs771877432 Chr.15: 78012887-78012887 on Build GRCh38
rs747927065 Chr.15: 78012860-78012860 on Build GRCh38
rs768925166 Chr.15: 78013041-78013041 on Build GRCh38
rs779563838 Chr.15: 78013247-78013247 on Build GRCh38
rs746009252 Chr.15: 78013023-78013023 on Build GRCh38
rs750665936 Chr.15: 78013174-78013174 on Build GRCh38
rs984175938 Chr.15: 78013283-78013283 on Build GRCh38
rs780061299 Chr.15: 78013086-78013086 on Build GRCh38
rs142452030 Chr.15: 78013098-78013098 on Build GRCh38
rs781266509 Chr.15: 78013013-78013013 on Build GRCh38
rs985235984 Chr.15: 78012915-78012915 on Build GRCh38
rs866980960 Chr.15: 78013137-78013137 on Build GRCh38
rs138268229 Chr.15: 78012806-78012806 on Build GRCh38
rs775966091 Chr.15: 78012916-78012916 on Build GRCh38
rs762817054 Chr.15: 78013122-78013122 on Build GRCh38
rs765837023 Chr.15: 78012947-78012947 on Build GRCh38
rs778230884 Chr.15: 78013242-78013242 on Build GRCh38
rs768023792 Chr.15: 78013256-78013256 on Build GRCh38
rs749781151 Chr.15: 78013036-78013036 on Build GRCh38
rs772789609 Chr.15: 78012942-78012942 on Build GRCh38
rs1021729301 Chr.15: 78013145-78013145 on Build GRCh38
rs774154833 Chr.15: 78013153-78013153 on Build GRCh38
rs565167596 Chr.15: 78012904-78012904 on Build GRCh38
rs77039980 Chr.15: 78012819-78012819 on Build GRCh38
rs769447237 Chr.15: 78013119-78013119 on Build GRCh38
rs201895110 Chr.15: 78013245-78013245 on Build GRCh38
rs776889349 Chr.15: 78013123-78013125 on Build GRCh38
rs531931885 Chr.15: 78012872-78012872 on Build GRCh38
rs780580937 Chr.15: 78013033-78013033 on Build GRCh38
rs891804312 Chr.15: 78013115-78013115 on Build GRCh38
rs752480801 Chr.15: 78012859-78012859 on Build GRCh38
rs774737781 Chr.15: 78013047-78013047 on Build GRCh38
rs765891435 Chr.15: 78012835-78012835 on Build GRCh38
rs775096530 Chr.15: 78013120-78013120 on Build GRCh38
rs376590157 Chr.15: 78012996-78012996 on Build GRCh38
rs373974886 Chr.15: 78013197-78013197 on Build GRCh38
rs776408611 Chr.15: 78013055-78013055 on Build GRCh38
rs781295772 Chr.15: 78012900-78012900 on Build GRCh38
rs754926191 Chr.15: 78012845-78012845 on Build GRCh38
rs772924996 Chr.15: 78013280-78013280 on Build GRCh38
rs377622910 Chr.15: 78013275-78013275 on Build GRCh38
rs547997471 Chr.15: 78013114-78013114 on Build GRCh38
rs771886267 Chr.15: 78013277-78013277 on Build GRCh38
rs569486767 Chr.15: 78013162-78013162 on Build GRCh38
rs760847886 Chr.15: 78013190-78013190 on Build GRCh38
rs770558751 Chr.15: 78013053-78013053 on Build GRCh38
rs530794756 Chr.15: 78013224-78013224 on Build GRCh38
rs767136041 Chr.15: 78012810-78012810 on Build GRCh38
rs778491182 Chr.15: 78013202-78013202 on Build GRCh38
rs749790935 Chr.15: 78013093-78013093 on Build GRCh38
rs201500278 Chr.15: 78012873-78012873 on Build GRCh38
rs139151755 Chr.15: 78013042-78013042 on Build GRCh38
rs753110801 Chr.15: 78013220-78013220 on Build GRCh38
rs150768376 Chr.15: 78012997-78012997 on Build GRCh38
rs913712270 Chr.15: 78012846-78012846 on Build GRCh38
rs536975479 Chr.15: 78013200-78013200 on Build GRCh38
rs769516617 Chr.15: 78013244-78013244 on Build GRCh38
rs773080972 Chr.15: 78012950-78012950 on Build GRCh38
rs763804734 Chr.15: 78013071-78013071 on Build GRCh38
rs752674926 Chr.15: 78012973-78012973 on Build GRCh38
rs778854483 Chr.15: 78012852-78012852 on Build GRCh38
rs146036476 Chr.15: 78013083-78013083 on Build GRCh38
rs745543777 Chr.15: 78013243-78013243 on Build GRCh38
rs756237475 Chr.15: 78013085-78013085 on Build GRCh38
rs945202011 Chr.15: 78012930-78012930 on Build GRCh38
rs758165409 Chr.15: 78012981-78012981 on Build GRCh38
rs774397112 Chr.15: 78013268-78013268 on Build GRCh38
rs766350273 Chr.15: 78013056-78013056 on Build GRCh38
rs199722270 Chr.15: 78012934-78012934 on Build GRCh38
rs149524007 Chr.15: 78013077-78013077 on Build GRCh38
rs141915348 Chr.15: 78012936-78012936 on Build GRCh38
rs764806059 Chr.15: 78013060-78013060 on Build GRCh38
rs552433664 Chr.15: 78013203-78013203 on Build GRCh38
rs775614174 Chr.15: 78013061-78013061 on Build GRCh38
rs746644370 Chr.15: 78013048-78013048 on Build GRCh38
rs779538966 Chr.15: 78013215-78013215 on Build GRCh38
rs768700129 Chr.15: 78013123-78013123 on Build GRCh38

View Full Product Details