Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83564977-83565475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3GL3
Entrez Gene ID
6457
Gene Name
SH3 domain containing GRB2 like 3, endophilin A3
Gene Aliases
CNSA3, EEN-B2, HsT19371, SH3D2C, SH3P13
UniGene
Hs.270055
Gene Chromosome Location
Chr.15: 83447123-83618743 on Build GRCh38
Transcripts
NM_001301108.1 NM_001301109.1 NM_001324182.1 NM_001324183.1 NM_001324184.1 NM_001324185.1 NM_001324186.1 NM_001324187.1 NM_003027.4 NR_125370.1 NR_136711.1 NR_136712.1 NR_136713.1 NR_136714.1 XM_011521889.1 XM_011521891.2 XM_011521892.2 XM_017022486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7389649 c.127A>T p.T43S Chr.15: 83565146-83565146 on Build GRCh38
4725663 c.171T>G p.Y57* Chr.15: 83565190-83565190 on Build GRCh38
1608763 c.145G>A p.E49K Chr.15: 83565164-83565164 on Build GRCh38
4995613 c.200A>T p.Q67L Chr.15: 83565195-83565195 on Build GRCh38
1588214 c.151C>A p.L51I Chr.15: 83565170-83565170 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762145370 Chr.15: 83565287-83565287 on Build GRCh38
rs556339318 Chr.15: 83565369-83565369 on Build GRCh38
rs978319893 Chr.15: 83565390-83565390 on Build GRCh38
rs763528544 Chr.15: 83565249-83565249 on Build GRCh38
rs764305405 Chr.15: 83565261-83565261 on Build GRCh38
rs939810743 Chr.15: 83565032-83565032 on Build GRCh38
rs539129066 Chr.15: 83565212-83565212 on Build GRCh38
rs946952265 Chr.15: 83565344-83565344 on Build GRCh38
rs1025052756 Chr.15: 83565357-83565357 on Build GRCh38
rs746705173 Chr.15: 83565207-83565207 on Build GRCh38
rs758301475 Chr.15: 83565314-83565314 on Build GRCh38
rs754796115 Chr.15: 83565302-83565302 on Build GRCh38
rs746126766 Chr.15: 83565034-83565034 on Build GRCh38
rs774530411 Chr.15: 83565273-83565273 on Build GRCh38
rs536822525 Chr.15: 83565031-83565031 on Build GRCh38
rs777243734 Chr.15: 83565191-83565191 on Build GRCh38
rs536010887 Chr.15: 83565355-83565355 on Build GRCh38
rs763064369 Chr.15: 83565108-83565108 on Build GRCh38
rs748072490 Chr.15: 83565174-83565174 on Build GRCh38
rs765615934 Chr.15: 83565097-83565097 on Build GRCh38
rs753669508 Chr.15: 83565301-83565301 on Build GRCh38
rs373247529 Chr.15: 83565228-83565228 on Build GRCh38
rs752680769 Chr.15: 83565310-83565310 on Build GRCh38
rs754796484 Chr.15: 83565105-83565108 on Build GRCh38
rs752839571 Chr.15: 83565106-83565106 on Build GRCh38
rs533824738 Chr.15: 83565150-83565150 on Build GRCh38
rs753525950 Chr.15: 83565143-83565143 on Build GRCh38
rs978373819 Chr.15: 83565048-83565048 on Build GRCh38
rs764252226 Chr.15: 83565117-83565117 on Build GRCh38
rs763636963 Chr.15: 83565050-83565050 on Build GRCh38
rs757962311 Chr.15: 83565183-83565183 on Build GRCh38
rs775932448 Chr.15: 83565236-83565236 on Build GRCh38
rs779927565 Chr.15: 83565127-83565127 on Build GRCh38
rs1043958241 Chr.15: 83565365-83565365 on Build GRCh38
rs567224148 Chr.15: 83565331-83565331 on Build GRCh38
rs770259067 Chr.15: 83565347-83565347 on Build GRCh38
rs767575839 Chr.15: 83565292-83565292 on Build GRCh38
rs867796792 Chr.15: 83565045-83565045 on Build GRCh38
rs751875660 Chr.15: 83565119-83565119 on Build GRCh38
rs562418217 Chr.15: 83565003-83565003 on Build GRCh38
rs770531560 Chr.15: 83565221-83565221 on Build GRCh38
rs1053973978 Chr.15: 83565081-83565081 on Build GRCh38
rs770342421 Chr.15: 83565235-83565235 on Build GRCh38
rs547245438 Chr.15: 83565308-83565308 on Build GRCh38
rs111704318 Chr.15: 83565426-83565426 on Build GRCh38
rs571266621 Chr.15: 83565069-83565069 on Build GRCh38
rs757519795 Chr.15: 83565126-83565126 on Build GRCh38
rs911437391 Chr.15: 83565340-83565340 on Build GRCh38
rs117749710 Chr.15: 83565432-83565432 on Build GRCh38
rs115956298 Chr.15: 83565427-83565427 on Build GRCh38
rs922123704 Chr.15: 83565440-83565440 on Build GRCh38
rs777079660 Chr.15: 83565223-83565223 on Build GRCh38
rs890497772 Chr.15: 83565156-83565156 on Build GRCh38
rs140175336 Chr.15: 83565022-83565022 on Build GRCh38

View Full Product Details