Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35151444-35151849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD7
Entrez Gene ID
53822
Gene Name
FXYD domain containing ion transport regulator 7
Gene Aliases
-
UniGene
Hs.134729
Gene Chromosome Location
Chr.19: 35143250-35154302 on Build GRCh38
Transcripts
NM_022006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3197415 c.161C>T p.A54V Chr.19: 35151464-35151464 on Build GRCh38
994897 c.143A>C p.K48T Chr.19: 35151446-35151446 on Build GRCh38
5855485 c.220G>A p.A74T Chr.19: 35151673-35151673 on Build GRCh38
3532447 c.213C>T p.P71P Chr.19: 35151666-35151666 on Build GRCh38
225772 c.194C>T p.S65F Chr.19: 35151647-35151647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111908466 Chr.19: 35151514-35151514 on Build GRCh38
rs150824677 Chr.19: 35151778-35151778 on Build GRCh38
rs1005887235 Chr.19: 35151586-35151586 on Build GRCh38
rs777311315 Chr.19: 35151698-35151698 on Build GRCh38
rs573302966 Chr.19: 35151764-35151764 on Build GRCh38
rs916603747 Chr.19: 35151774-35151774 on Build GRCh38
rs112317274 Chr.19: 35151811-35151811 on Build GRCh38
rs369408080 Chr.19: 35151529-35151529 on Build GRCh38
rs759257262 Chr.19: 35151534-35151534 on Build GRCh38
rs752322086 Chr.19: 35151694-35151694 on Build GRCh38
rs777644039 Chr.19: 35151520-35151520 on Build GRCh38
rs772263936 Chr.19: 35151721-35151721 on Build GRCh38
rs778434452 Chr.19: 35151515-35151515 on Build GRCh38
rs948685185 Chr.19: 35151540-35151540 on Build GRCh38
rs907060863 Chr.19: 35151571-35151571 on Build GRCh38
rs117378441 Chr.19: 35151782-35151782 on Build GRCh38
rs780779173 Chr.19: 35151587-35151587 on Build GRCh38
rs370869800 Chr.19: 35151723-35151723 on Build GRCh38
rs36072726 Chr.19: 35151748-35151748 on Build GRCh38
rs747632330 Chr.19: 35151516-35151516 on Build GRCh38
rs766842900 Chr.19: 35151667-35151667 on Build GRCh38
rs73588876 Chr.19: 35151580-35151580 on Build GRCh38
rs967342364 Chr.19: 35151747-35151747 on Build GRCh38
rs996536186 Chr.19: 35151736-35151737 on Build GRCh38
rs745627309 Chr.19: 35151708-35151708 on Build GRCh38
rs748933900 Chr.19: 35151476-35151476 on Build GRCh38
rs973934684 Chr.19: 35151780-35151780 on Build GRCh38
rs945550389 Chr.19: 35151507-35151507 on Build GRCh38
rs770789018 Chr.19: 35151704-35151704 on Build GRCh38
rs774604982 Chr.19: 35151715-35151715 on Build GRCh38
rs757892421 Chr.19: 35151697-35151697 on Build GRCh38
rs746946231 Chr.19: 35151702-35151702 on Build GRCh38
rs116812496 Chr.19: 35151824-35151824 on Build GRCh38
rs761913417 Chr.19: 35151596-35151596 on Build GRCh38
rs769511911 Chr.19: 35151714-35151714 on Build GRCh38
rs950098450 Chr.19: 35151825-35151825 on Build GRCh38
rs200372075 Chr.19: 35151531-35151531 on Build GRCh38
rs199623974 Chr.19: 35151628-35151628 on Build GRCh38
rs976768121 Chr.19: 35151729-35151729 on Build GRCh38
rs117593335 Chr.19: 35151572-35151572 on Build GRCh38
rs550509616 Chr.19: 35151733-35151733 on Build GRCh38
rs141725524 Chr.19: 35151477-35151477 on Build GRCh38
rs551902878 Chr.19: 35151776-35151777 on Build GRCh38
rs986446640 Chr.19: 35151753-35151753 on Build GRCh38
rs756667641 Chr.19: 35151520-35151520 on Build GRCh38
rs372801986 Chr.19: 35151575-35151575 on Build GRCh38
rs750946668 Chr.19: 35151610-35151610 on Build GRCh38
rs910183506 Chr.19: 35151465-35151465 on Build GRCh38
rs754161730 Chr.19: 35151674-35151674 on Build GRCh38
rs182197816 Chr.19: 35151519-35151519 on Build GRCh38
rs903396388 Chr.19: 35151657-35151657 on Build GRCh38
rs745755708 Chr.19: 35151678-35151678 on Build GRCh38
rs368538591 Chr.19: 35151779-35151779 on Build GRCh38
rs768869520 Chr.19: 35151583-35151583 on Build GRCh38
rs1030565946 Chr.19: 35151742-35151742 on Build GRCh38
rs1047556683 Chr.19: 35151822-35151822 on Build GRCh38
rs780612366 Chr.19: 35151474-35151474 on Build GRCh38
rs200383866 Chr.19: 35151506-35151506 on Build GRCh38
rs370412632 Chr.19: 35151567-35151567 on Build GRCh38
rs73588877 Chr.19: 35151817-35151817 on Build GRCh38
rs750999432 Chr.19: 35151466-35151466 on Build GRCh38
rs111645945 Chr.19: 35151749-35151749 on Build GRCh38
rs774459332 Chr.19: 35151585-35151585 on Build GRCh38
rs936742098 Chr.19: 35151750-35151750 on Build GRCh38
rs186468123 Chr.19: 35151804-35151804 on Build GRCh38
rs572815640 Chr.19: 35151773-35151773 on Build GRCh38
rs112381083 Chr.19: 35151486-35151486 on Build GRCh38
rs990900932 Chr.19: 35151769-35151769 on Build GRCh38
rs755307907 Chr.19: 35151680-35151680 on Build GRCh38
rs919744918 Chr.19: 35151781-35151781 on Build GRCh38
rs756518153 Chr.19: 35151473-35151473 on Build GRCh38
rs761993469 Chr.19: 35151718-35151718 on Build GRCh38
rs778578245 Chr.19: 35151688-35151688 on Build GRCh38
rs767746532 Chr.19: 35151605-35151605 on Build GRCh38
rs964399170 Chr.19: 35151623-35151623 on Build GRCh38
rs965331653 Chr.19: 35151469-35151469 on Build GRCh38
rs376911183 Chr.19: 35151522-35151522 on Build GRCh38
rs1045644239 Chr.19: 35151551-35151551 on Build GRCh38
rs781120968 Chr.19: 35151706-35151706 on Build GRCh38

View Full Product Details