Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55172425-55172931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLHC1
Entrez Gene ID
130162
Gene Name
clathrin heavy chain linker domain containing 1
Gene Aliases
C2orf63
UniGene
Hs.347014
Gene Chromosome Location
Chr.2: 55172547-55232563 on Build GRCh38
Transcripts
NM_001135598.1 NM_152385.2 XM_005264136.2 XM_006711937.3 XM_006711938.3 XM_006711939.3 XM_011532519.2 XM_011532520.2 XM_011532521.2 XM_011532522.2 XM_011532523.2 XM_011532524.1 XM_011532525.2 XM_011532527.2 XM_011532528.1 XM_011532529.2 XM_011532530.1 XM_011532534.1 XM_011532535.2 XM_017003333.1 XM_017003334.1 XM_017003335.1 XM_017003336.1 XM_017003337.1 XM_017003338.1 XM_017003339.1 XM_017003340.1 XM_017003341.1 XM_017003342.1 XM_017003343.1 XM_017003344.1 XM_017003345.1 XM_017003346.1 XM_017003347.1 XM_017003349.1 XM_017003350.1

SNP Detail

SNP ID Chromosome Location
rs4233959 Chr.2: 55172496-55172496 on Build GRCh38
rs397748783 Chr.2: 55172518-55172519 on Build GRCh38
rs55848760 Chr.2: 55172508-55172509 on Build GRCh38
rs978263581 Chr.2: 55172541-55172541 on Build GRCh38
rs1034191884 Chr.2: 55172725-55172725 on Build GRCh38
rs967138715 Chr.2: 55172789-55172789 on Build GRCh38
rs993574194 Chr.2: 55172773-55172791 on Build GRCh38
rs1019959601 Chr.2: 55172533-55172533 on Build GRCh38
rs962065570 Chr.2: 55172902-55172902 on Build GRCh38
rs930075799 Chr.2: 55172685-55172685 on Build GRCh38
rs117391002 Chr.2: 55172901-55172901 on Build GRCh38
rs62138271 Chr.2: 55172716-55172716 on Build GRCh38
rs147538630 Chr.2: 55172763-55172763 on Build GRCh38
rs893064559 Chr.2: 55172733-55172733 on Build GRCh38
rs922789071 Chr.2: 55172564-55172564 on Build GRCh38
rs796210245 Chr.2: 55172716-55172716 on Build GRCh38
rs942346050 Chr.2: 55172625-55172625 on Build GRCh38
rs910911258 Chr.2: 55172623-55172623 on Build GRCh38
rs534897143 Chr.2: 55172833-55172833 on Build GRCh38
rs972539155 Chr.2: 55172450-55172450 on Build GRCh38
rs911479044 Chr.2: 55172621-55172621 on Build GRCh38
rs187383804 Chr.2: 55172686-55172686 on Build GRCh38
rs11347337 Chr.2: 55172702-55172702 on Build GRCh38
rs869181869 Chr.2: 55172714-55172715 on Build GRCh38
rs372707510 Chr.2: 55172507-55172508 on Build GRCh38
rs1038411984 Chr.2: 55172636-55172636 on Build GRCh38
rs985684615 Chr.2: 55172620-55172620 on Build GRCh38
rs1000429465 Chr.2: 55172801-55172801 on Build GRCh38
rs374220487 Chr.2: 55172519-55172519 on Build GRCh38
rs551255762 Chr.2: 55172703-55172703 on Build GRCh38
rs1011191257 Chr.2: 55172757-55172757 on Build GRCh38
rs1030098207 Chr.2: 55172884-55172884 on Build GRCh38
rs907002275 Chr.2: 55172718-55172718 on Build GRCh38
rs549894391 Chr.2: 55172554-55172554 on Build GRCh38
rs565771270 Chr.2: 55172797-55172797 on Build GRCh38
rs373774572 Chr.2: 55172693-55172693 on Build GRCh38
rs114568702 Chr.2: 55172634-55172634 on Build GRCh38
rs997643986 Chr.2: 55172869-55172869 on Build GRCh38
rs934186047 Chr.2: 55172614-55172614 on Build GRCh38
rs1056433364 Chr.2: 55172861-55172861 on Build GRCh38
rs897235191 Chr.2: 55172643-55172643 on Build GRCh38
rs530153001 Chr.2: 55172715-55172716 on Build GRCh38
rs967568081 Chr.2: 55172534-55172534 on Build GRCh38
rs554488598 Chr.2: 55172883-55172883 on Build GRCh38
rs957014191 Chr.2: 55172454-55172454 on Build GRCh38
rs894505740 Chr.2: 55172870-55172870 on Build GRCh38
rs922343265 Chr.2: 55172644-55172644 on Build GRCh38
rs775460148 Chr.2: 55172560-55172560 on Build GRCh38
rs528660157 Chr.2: 55172719-55172719 on Build GRCh38

View Full Product Details