Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 119608360-119608904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP221
Entrez Gene ID
200373
Gene Name
cilia and flagella associated protein 221
Gene Aliases
FAP221, PCDP1
UniGene
Hs.496753
Gene Chromosome Location
Chr.2: 119544392-119688610 on Build GRCh38
Transcripts
NM_001271049.1 NR_073132.1 NR_073133.1 XM_006712353.3 XM_017003550.1 XM_017003551.1 XM_017003552.1 XM_017003553.1 XM_017003554.1 XM_017003555.1 XM_017003556.1 XM_017003557.1 XM_017003558.1 XM_017003559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6446477 c.346G>A p.A116T Chr.2: 119608572-119608572 on Build GRCh38
1287022 c.353C>G p.A118G Chr.2: 119608579-119608579 on Build GRCh38
6741314 c.299C>A p.P100H Chr.2: 119608525-119608525 on Build GRCh38
1185986 c.281T>G p.V94G Chr.2: 119608507-119608507 on Build GRCh38
6325636 c.331G>A p.E111K Chr.2: 119608557-119608557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901169527 Chr.2: 119608874-119608874 on Build GRCh38
rs772116880 Chr.2: 119608499-119608499 on Build GRCh38
rs765699546 Chr.2: 119608524-119608524 on Build GRCh38
rs878957316 Chr.2: 119608494-119608494 on Build GRCh38
rs773999512 Chr.2: 119608471-119608471 on Build GRCh38
rs756944517 Chr.2: 119608552-119608552 on Build GRCh38
rs1006196346 Chr.2: 119608646-119608646 on Build GRCh38
rs981961290 Chr.2: 119608417-119608417 on Build GRCh38
rs940766782 Chr.2: 119608411-119608411 on Build GRCh38
rs1023305003 Chr.2: 119608675-119608675 on Build GRCh38
rs775522316 Chr.2: 119608502-119608502 on Build GRCh38
rs559428425 Chr.2: 119608848-119608848 on Build GRCh38
rs371898156 Chr.2: 119608598-119608598 on Build GRCh38
rs138738841 Chr.2: 119608708-119608708 on Build GRCh38
rs990632151 Chr.2: 119608718-119608718 on Build GRCh38
rs11398750 Chr.2: 119608481-119608482 on Build GRCh38
rs796969009 Chr.2: 119608847-119608847 on Build GRCh38
rs749022533 Chr.2: 119608572-119608572 on Build GRCh38
rs553476454 Chr.2: 119608447-119608447 on Build GRCh38
rs867373975 Chr.2: 119608454-119608454 on Build GRCh38
rs748821223 Chr.2: 119608456-119608456 on Build GRCh38
rs754104887 Chr.2: 119608507-119608507 on Build GRCh38
rs762744970 Chr.2: 119608624-119608624 on Build GRCh38
rs907934292 Chr.2: 119608394-119608394 on Build GRCh38
rs981546726 Chr.2: 119608691-119608691 on Build GRCh38
rs386391028 Chr.2: 119608487-119608488 on Build GRCh38
rs141322055 Chr.2: 119608580-119608580 on Build GRCh38
rs750592832 Chr.2: 119608617-119608617 on Build GRCh38
rs139037503 Chr.2: 119608527-119608527 on Build GRCh38
rs1039633258 Chr.2: 119608873-119608873 on Build GRCh38
rs902218062 Chr.2: 119608534-119608534 on Build GRCh38
rs368000329 Chr.2: 119608565-119608565 on Build GRCh38
rs534287943 Chr.2: 119608482-119608483 on Build GRCh38
rs548329414 Chr.2: 119608829-119608829 on Build GRCh38
rs764205097 Chr.2: 119608505-119608505 on Build GRCh38
rs759591265 Chr.2: 119608482-119608482 on Build GRCh38
rs752020326 Chr.2: 119608550-119608550 on Build GRCh38
rs989330303 Chr.2: 119608838-119608838 on Build GRCh38
rs778376512 Chr.2: 119608633-119608633 on Build GRCh38
rs927800917 Chr.2: 119608419-119608419 on Build GRCh38
rs796585806 Chr.2: 119608482-119608483 on Build GRCh38
rs753451446 Chr.2: 119608483-119608484 on Build GRCh38
rs578213297 Chr.2: 119608482-119608482 on Build GRCh38
rs5833792 Chr.2: 119608493-119608494 on Build GRCh38
rs901589599 Chr.2: 119608882-119608882 on Build GRCh38
rs776569291 Chr.2: 119608606-119608606 on Build GRCh38
rs34210676 Chr.2: 119608853-119608853 on Build GRCh38
rs141884466 Chr.2: 119608521-119608521 on Build GRCh38
rs752071631 Chr.2: 119608637-119608637 on Build GRCh38
rs79983908 Chr.2: 119608545-119608545 on Build GRCh38
rs760871327 Chr.2: 119608503-119608503 on Build GRCh38
rs755622064 Chr.2: 119608554-119608554 on Build GRCh38
rs946362373 Chr.2: 119608515-119608515 on Build GRCh38
rs1521915 Chr.2: 119608719-119608719 on Build GRCh38
rs368674563 Chr.2: 119608607-119608607 on Build GRCh38
rs991776663 Chr.2: 119608648-119608648 on Build GRCh38
rs775429707 Chr.2: 119608709-119608709 on Build GRCh38
rs578016183 Chr.2: 119608464-119608464 on Build GRCh38
rs375225988 Chr.2: 119608602-119608602 on Build GRCh38
rs946624012 Chr.2: 119608777-119608777 on Build GRCh38
rs765354932 Chr.2: 119608611-119608611 on Build GRCh38
rs939221028 Chr.2: 119608420-119608420 on Build GRCh38
rs868381769 Chr.2: 119608500-119608500 on Build GRCh38
rs200577746 Chr.2: 119608496-119608496 on Build GRCh38
rs371925073 Chr.2: 119608625-119608625 on Build GRCh38
rs146574324 Chr.2: 119608402-119608402 on Build GRCh38
rs758952901 Chr.2: 119608528-119608528 on Build GRCh38
rs970441367 Chr.2: 119608694-119608694 on Build GRCh38
rs575681640 Chr.2: 119608601-119608601 on Build GRCh38
rs1039684640 Chr.2: 119608399-119608399 on Build GRCh38
rs386391029 Chr.2: 119608488-119608489 on Build GRCh38
rs778821197 Chr.2: 119608585-119608585 on Build GRCh38
rs901214991 Chr.2: 119608533-119608533 on Build GRCh38
rs1014919009 Chr.2: 119608652-119608652 on Build GRCh38
rs935134195 Chr.2: 119608778-119608778 on Build GRCh38
rs150187235 Chr.2: 119608665-119608665 on Build GRCh38
rs984790959 Chr.2: 119608392-119608392 on Build GRCh38
rs754099944 Chr.2: 119608860-119608860 on Build GRCh38
rs1055017527 Chr.2: 119608436-119608436 on Build GRCh38

View Full Product Details