Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1720799-1721299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM129
Entrez Gene ID
92305
Gene Name
transmembrane protein 129
Gene Aliases
D4S2561E
UniGene
Hs.518562
Gene Chromosome Location
Chr.4: 1715952-1721357 on Build GRCh38
Transcripts
NM_001127266.1 NM_138385.3 XM_005248039.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6414258 c.23T>A p.F8Y Chr.4: 1720815-1720815 on Build GRCh38
6414257 c.23T>A p.F8Y Chr.4: 1720815-1720815 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1052519136 Chr.4: 1721086-1721086 on Build GRCh38
rs113260869 Chr.4: 1720871-1720871 on Build GRCh38
rs1016233695 Chr.4: 1720916-1720916 on Build GRCh38
rs1027298751 Chr.4: 1720991-1720991 on Build GRCh38
rs897977615 Chr.4: 1721182-1721182 on Build GRCh38
rs986605717 Chr.4: 1720939-1720939 on Build GRCh38
rs1012999712 Chr.4: 1721242-1721242 on Build GRCh38
rs865891140 Chr.4: 1720825-1720825 on Build GRCh38
rs1028684009 Chr.4: 1720927-1720927 on Build GRCh38
rs971772845 Chr.4: 1720967-1720967 on Build GRCh38
rs955181135 Chr.4: 1720930-1720930 on Build GRCh38
rs924426483 Chr.4: 1721061-1721061 on Build GRCh38
rs1011198011 Chr.4: 1720892-1720892 on Build GRCh38
rs918192766 Chr.4: 1721124-1721124 on Build GRCh38
rs915821682 Chr.4: 1721043-1721043 on Build GRCh38
rs900536532 Chr.4: 1720919-1720919 on Build GRCh38
rs991455186 Chr.4: 1721033-1721033 on Build GRCh38
rs917417200 Chr.4: 1721055-1721055 on Build GRCh38
rs996610066 Chr.4: 1721213-1721213 on Build GRCh38
rs981290884 Chr.4: 1721056-1721056 on Build GRCh38
rs935380021 Chr.4: 1720861-1720861 on Build GRCh38
rs993244805 Chr.4: 1721212-1721212 on Build GRCh38
rs900483640 Chr.4: 1721209-1721209 on Build GRCh38
rs889387176 Chr.4: 1721116-1721116 on Build GRCh38
rs1050411839 Chr.4: 1721107-1721107 on Build GRCh38
rs6837127 Chr.4: 1721064-1721064 on Build GRCh38
rs757836155 Chr.4: 1720849-1720849 on Build GRCh38
rs951794255 Chr.4: 1721240-1721240 on Build GRCh38
rs935427433 Chr.4: 1721070-1721070 on Build GRCh38
rs1027350793 Chr.4: 1721223-1721223 on Build GRCh38
rs979597507 Chr.4: 1721078-1721078 on Build GRCh38
rs571390626 Chr.4: 1720890-1720890 on Build GRCh38
rs375704917 Chr.4: 1720848-1720848 on Build GRCh38
rs886617254 Chr.4: 1721161-1721161 on Build GRCh38
rs750776470 Chr.4: 1720863-1720863 on Build GRCh38
rs368824267 Chr.4: 1721047-1721047 on Build GRCh38
rs1020815527 Chr.4: 1720940-1720940 on Build GRCh38
rs1047847387 Chr.4: 1721143-1721143 on Build GRCh38
rs1030348249 Chr.4: 1721236-1721236 on Build GRCh38
rs951798301 Chr.4: 1720998-1720998 on Build GRCh38
rs891027290 Chr.4: 1721256-1721256 on Build GRCh38
rs556726322 Chr.4: 1721005-1721005 on Build GRCh38
rs752779738 Chr.4: 1720866-1720890 on Build GRCh38
rs1038231720 Chr.4: 1721196-1721196 on Build GRCh38
rs966407940 Chr.4: 1720948-1720948 on Build GRCh38
rs971733107 Chr.4: 1721264-1721264 on Build GRCh38
rs1040546565 Chr.4: 1721150-1721150 on Build GRCh38
rs866493095 Chr.4: 1721113-1721113 on Build GRCh38
rs10030268 Chr.4: 1721132-1721132 on Build GRCh38
rs6846887 Chr.4: 1721081-1721081 on Build GRCh38
rs942112851 Chr.4: 1721193-1721193 on Build GRCh38
rs537171723 Chr.4: 1720910-1720910 on Build GRCh38
rs369001628 Chr.4: 1720851-1720851 on Build GRCh38
rs1019049031 Chr.4: 1720931-1720931 on Build GRCh38
rs765904780 Chr.4: 1720853-1720853 on Build GRCh38
rs1008149682 Chr.4: 1721271-1721271 on Build GRCh38
rs6837277 Chr.4: 1721072-1721072 on Build GRCh38
rs924963238 Chr.4: 1721090-1721090 on Build GRCh38
rs949957961 Chr.4: 1721051-1721051 on Build GRCh38
rs867341081 Chr.4: 1721101-1721101 on Build GRCh38
rs758727950 Chr.4: 1720867-1720867 on Build GRCh38
rs996775102 Chr.4: 1720925-1720925 on Build GRCh38
rs375429325 Chr.4: 1720847-1720847 on Build GRCh38
rs767078157 Chr.4: 1720839-1720839 on Build GRCh38
rs759258858 Chr.4: 1720830-1720830 on Build GRCh38

View Full Product Details