Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 52116140-52116593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST18
Entrez Gene ID
9705
Gene Name
ST18, C2H2C-type zinc finger
Gene Aliases
NZF3, ZC2H2C3, ZC2HC10, ZNF387
UniGene
Hs.655499
Gene Chromosome Location
Chr.8: 52110832-52409879 on Build GRCh38
Transcripts
NM_014682.2 XM_006716487.1 XM_011517629.1 XM_011517631.1 XM_011517632.1 XM_011517633.1 XM_011517634.1 XM_011517635.1 XM_011517636.2 XM_011517637.1 XM_011517638.2 XM_011517641.1 XM_011517642.1 XM_017014047.1 XM_017014048.1 XM_017014049.1 XM_017014050.1 XM_017014051.1 XM_017014052.1 XM_017014053.1 XM_017014054.1 XM_017014055.1 XM_017014056.1 XM_017014057.1 XM_017014058.1 XM_017014059.1 XM_017014060.1 XM_017014061.1 XM_017014062.1 XM_017014063.1 XM_017014064.1 XM_017014065.1 XM_017014066.1 XM_017014067.1 XM_017014068.1 XM_017014069.1 XM_017014070.1 XM_017014071.1 XM_017014072.1 XM_017014073.1 XM_017014074.1 XM_017014075.1 XM_017014076.1 XM_017014077.1 XM_017014078.1 XM_017014079.1 XM_017014080.1 XM_017014081.1 XM_017014082.1 XM_017014083.1 XM_017014084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4422001 c.2979T>C p.L993L Chr.8: 52116299-52116299 on Build GRCh38
6791026 c.2998C>T p.Q1000* Chr.8: 52116280-52116280 on Build GRCh38
4517193 c.2950_2951GG>AA p.G984N Chr.8: 52116327-52116328 on Build GRCh38
1236642 c.2947G>T p.A983S Chr.8: 52116331-52116331 on Build GRCh38
1330959 c.2932C>T p.L978L Chr.8: 52116346-52116346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368520409 Chr.8: 52116318-52116318 on Build GRCh38
rs536467531 Chr.8: 52116474-52116474 on Build GRCh38
rs944928647 Chr.8: 52116331-52116331 on Build GRCh38
rs375870926 Chr.8: 52116467-52116467 on Build GRCh38
rs746566336 Chr.8: 52116341-52116341 on Build GRCh38
rs983449393 Chr.8: 52116328-52116328 on Build GRCh38
rs771570944 Chr.8: 52116264-52116264 on Build GRCh38
rs879231977 Chr.8: 52116195-52116195 on Build GRCh38
rs377522624 Chr.8: 52116250-52116250 on Build GRCh38
rs761756701 Chr.8: 52116301-52116301 on Build GRCh38
rs903604628 Chr.8: 52116460-52116460 on Build GRCh38
rs765135070 Chr.8: 52116302-52116302 on Build GRCh38
rs960606838 Chr.8: 52116209-52116209 on Build GRCh38
rs79619736 Chr.8: 52116207-52116207 on Build GRCh38
rs956431622 Chr.8: 52116537-52116537 on Build GRCh38
rs372846323 Chr.8: 52116342-52116342 on Build GRCh38
rs2303459 Chr.8: 52116198-52116198 on Build GRCh38
rs762921002 Chr.8: 52116383-52116383 on Build GRCh38
rs896434013 Chr.8: 52116529-52116529 on Build GRCh38
rs760691387 Chr.8: 52116406-52116406 on Build GRCh38
rs997875573 Chr.8: 52116489-52116489 on Build GRCh38
rs899464503 Chr.8: 52116483-52116483 on Build GRCh38
rs750367441 Chr.8: 52116497-52116497 on Build GRCh38
rs561592003 Chr.8: 52116184-52116184 on Build GRCh38
rs956718854 Chr.8: 52116203-52116203 on Build GRCh38
rs551060800 Chr.8: 52116332-52116332 on Build GRCh38
rs763878159 Chr.8: 52116278-52116278 on Build GRCh38
rs757493526 Chr.8: 52116439-52116439 on Build GRCh38
rs569206286 Chr.8: 52116338-52116338 on Build GRCh38
rs761558739 Chr.8: 52116357-52116357 on Build GRCh38
rs776876915 Chr.8: 52116251-52116251 on Build GRCh38
rs1023514139 Chr.8: 52116532-52116532 on Build GRCh38
rs976333786 Chr.8: 52116362-52116362 on Build GRCh38
rs199592058 Chr.8: 52116254-52116254 on Build GRCh38
rs776562044 Chr.8: 52116287-52116287 on Build GRCh38
rs750793196 Chr.8: 52116450-52116450 on Build GRCh38
rs571649544 Chr.8: 52116235-52116235 on Build GRCh38
rs756357334 Chr.8: 52116316-52116316 on Build GRCh38
rs146863606 Chr.8: 52116390-52116390 on Build GRCh38
rs1013932083 Chr.8: 52116531-52116531 on Build GRCh38
rs572834609 Chr.8: 52116490-52116490 on Build GRCh38
rs746366348 Chr.8: 52116256-52116256 on Build GRCh38
rs952062312 Chr.8: 52116268-52116268 on Build GRCh38
rs544004934 Chr.8: 52116382-52116382 on Build GRCh38
rs764165610 Chr.8: 52116425-52116425 on Build GRCh38
rs533814858 Chr.8: 52116528-52116528 on Build GRCh38
rs980734250 Chr.8: 52116214-52116214 on Build GRCh38
rs757551599 Chr.8: 52116330-52116330 on Build GRCh38
rs1043873709 Chr.8: 52116414-52116414 on Build GRCh38
rs2303458 Chr.8: 52116475-52116475 on Build GRCh38
rs578202236 Chr.8: 52116221-52116221 on Build GRCh38
rs905035117 Chr.8: 52116535-52116535 on Build GRCh38
rs751544423 Chr.8: 52116344-52116344 on Build GRCh38
rs935035018 Chr.8: 52116466-52116466 on Build GRCh38
rs781132197 Chr.8: 52116230-52116230 on Build GRCh38
rs999113226 Chr.8: 52116200-52116200 on Build GRCh38
rs766296777 Chr.8: 52116389-52116389 on Build GRCh38
rs1023875020 Chr.8: 52116220-52116220 on Build GRCh38
rs758294624 Chr.8: 52116313-52116313 on Build GRCh38
rs754020397 Chr.8: 52116437-52116437 on Build GRCh38
rs752763322 Chr.8: 52116315-52116315 on Build GRCh38
rs908148976 Chr.8: 52116329-52116329 on Build GRCh38
rs758937463 Chr.8: 52116452-52116452 on Build GRCh38
rs760351916 Chr.8: 52116276-52116276 on Build GRCh38
rs12334321 Chr.8: 52116570-52116570 on Build GRCh38
rs778082620 Chr.8: 52116249-52116249 on Build GRCh38
rs779313094 Chr.8: 52116442-52116442 on Build GRCh38
rs1027783039 Chr.8: 52116547-52116547 on Build GRCh38
rs769668358 Chr.8: 52116358-52116358 on Build GRCh38
rs776362008 Chr.8: 52116346-52116346 on Build GRCh38
rs756343534 Chr.8: 52116246-52116246 on Build GRCh38
rs113744338 Chr.8: 52116237-52116237 on Build GRCh38
rs754075407 Chr.8: 52116321-52116321 on Build GRCh38
rs774885425 Chr.8: 52116266-52116266 on Build GRCh38
rs1013441521 Chr.8: 52116216-52116216 on Build GRCh38
rs772503453 Chr.8: 52116334-52116334 on Build GRCh38
rs995346455 Chr.8: 52116538-52116538 on Build GRCh38

View Full Product Details