Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 70071058-70071556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM14
Entrez Gene ID
63978
Gene Name
PR domain 14
Gene Aliases
PFM11
UniGene
Hs.736037
Gene Chromosome Location
Chr.8: 70051126-70071327 on Build GRCh38
Transcripts
NM_024504.3

SNP Detail

SNP ID Chromosome Location
rs942951533 Chr.8: 70071515-70071515 on Build GRCh38
rs976107294 Chr.8: 70071220-70071220 on Build GRCh38
rs947620495 Chr.8: 70071331-70071331 on Build GRCh38
rs534958935 Chr.8: 70071466-70071466 on Build GRCh38
rs746905044 Chr.8: 70071333-70071333 on Build GRCh38
rs545546773 Chr.8: 70071168-70071168 on Build GRCh38
rs941450323 Chr.8: 70071282-70071282 on Build GRCh38
rs1024217924 Chr.8: 70071119-70071120 on Build GRCh38
rs894295414 Chr.8: 70071097-70071097 on Build GRCh38
rs1016140595 Chr.8: 70071402-70071402 on Build GRCh38
rs1017303827 Chr.8: 70071386-70071386 on Build GRCh38
rs1044209487 Chr.8: 70071298-70071298 on Build GRCh38
rs984752714 Chr.8: 70071238-70071238 on Build GRCh38
rs113983474 Chr.8: 70071286-70071286 on Build GRCh38
rs964388180 Chr.8: 70071390-70071390 on Build GRCh38
rs114909879 Chr.8: 70071488-70071488 on Build GRCh38
rs748386332 Chr.8: 70071133-70071133 on Build GRCh38
rs3750230 Chr.8: 70071249-70071249 on Build GRCh38
rs954274248 Chr.8: 70071125-70071125 on Build GRCh38
rs1006979023 Chr.8: 70071129-70071129 on Build GRCh38
rs138124001 Chr.8: 70071382-70071382 on Build GRCh38
rs971512122 Chr.8: 70071122-70071122 on Build GRCh38
rs867271969 Chr.8: 70071264-70071264 on Build GRCh38
rs768457541 Chr.8: 70071362-70071362 on Build GRCh38
rs181064572 Chr.8: 70071306-70071306 on Build GRCh38
rs908519566 Chr.8: 70071275-70071275 on Build GRCh38
rs769312065 Chr.8: 70071282-70071282 on Build GRCh38
rs918614209 Chr.8: 70071211-70071211 on Build GRCh38
rs938422039 Chr.8: 70071458-70071458 on Build GRCh38
rs1012750691 Chr.8: 70071325-70071325 on Build GRCh38
rs761579411 Chr.8: 70071401-70071401 on Build GRCh38
rs901235420 Chr.8: 70071372-70071372 on Build GRCh38
rs536610176 Chr.8: 70071533-70071533 on Build GRCh38
rs984492491 Chr.8: 70071407-70071407 on Build GRCh38
rs115207527 Chr.8: 70071118-70071118 on Build GRCh38
rs560589565 Chr.8: 70071291-70071291 on Build GRCh38
rs528003874 Chr.8: 70071232-70071232 on Build GRCh38
rs965461692 Chr.8: 70071172-70071172 on Build GRCh38
rs890036951 Chr.8: 70071348-70071348 on Build GRCh38
rs542802290 Chr.8: 70071115-70071115 on Build GRCh38
rs985110150 Chr.8: 70071400-70071400 on Build GRCh38
rs565256126 Chr.8: 70071169-70071169 on Build GRCh38
rs952776911 Chr.8: 70071398-70071398 on Build GRCh38
rs1043124080 Chr.8: 70071490-70071490 on Build GRCh38
rs112145305 Chr.8: 70071403-70071403 on Build GRCh38
rs988613932 Chr.8: 70071469-70071469 on Build GRCh38
rs574075421 Chr.8: 70071100-70071100 on Build GRCh38
rs150170021 Chr.8: 70071478-70071478 on Build GRCh38
rs916002117 Chr.8: 70071513-70071513 on Build GRCh38
rs549612726 Chr.8: 70071316-70071316 on Build GRCh38
rs565866332 Chr.8: 70071428-70071428 on Build GRCh38
rs938446382 Chr.8: 70071315-70071315 on Build GRCh38
rs998536518 Chr.8: 70071146-70071146 on Build GRCh38
rs10216606 Chr.8: 70071153-70071153 on Build GRCh38
rs928487474 Chr.8: 70071414-70071414 on Build GRCh38
rs891491528 Chr.8: 70071329-70071329 on Build GRCh38
rs1011857008 Chr.8: 70071077-70071077 on Build GRCh38
rs890013238 Chr.8: 70071507-70071507 on Build GRCh38
rs994275834 Chr.8: 70071380-70071380 on Build GRCh38
rs752449029 Chr.8: 70071207-70071224 on Build GRCh38
rs777846834 Chr.8: 70071214-70071214 on Build GRCh38
rs972701105 Chr.8: 70071188-70071188 on Build GRCh38

View Full Product Details