Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 12775735-12776242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LURAP1L
Entrez Gene ID
286343
Gene Name
leucine rich adaptor protein 1 like
Gene Aliases
C9orf150, HYST0841, LRAP35b, bA3L8.2
UniGene
Hs.445356
Gene Chromosome Location
Chr.9: 12774990-12823060 on Build GRCh38
Transcripts
NM_203403.1 XM_005251443.2 NR_125775.1

Gene Symbol
LURAP1L-AS1
Entrez Gene ID
101929467
Gene Name
LURAP1L antisense RNA 1
Gene Aliases
-
UniGene
Hs.586443
Gene Chromosome Location
Chr.9: 12699999-12814389 on Build GRCh38
Transcripts
NM_203403.1 XM_005251443.2 NR_125775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6044238 c.60A>G p.V20V Chr.9: 12775775-12775775 on Build GRCh38
3324831 c.114G>A p.R38R Chr.9: 12775829-12775829 on Build GRCh38
5977067 c.148_150delGGC p.G55delG Chr.9: 12775863-12775865 on Build GRCh38
265673 c.148_156delGGCGGCGGC p.G50_G52delGGG Chr.9: 12775863-12775871 on Build GRCh38
6208102 c.239G>A p.G80D Chr.9: 12775954-12775954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753416448 Chr.9: 12776054-12776054 on Build GRCh38
rs374144313 Chr.9: 12775800-12775800 on Build GRCh38
rs750540640 Chr.9: 12775789-12775789 on Build GRCh38
rs567788672 Chr.9: 12775824-12775824 on Build GRCh38
rs114639122 Chr.9: 12776188-12776188 on Build GRCh38
rs762776237 Chr.9: 12775838-12775838 on Build GRCh38
rs3833707 Chr.9: 12775880-12775881 on Build GRCh38
rs779311221 Chr.9: 12775849-12775849 on Build GRCh38
rs148872560 Chr.9: 12775970-12775970 on Build GRCh38
rs757788448 Chr.9: 12775872-12775873 on Build GRCh38
rs769453501 Chr.9: 12775854-12775856 on Build GRCh38
rs757545946 Chr.9: 12775781-12775781 on Build GRCh38
rs759281702 Chr.9: 12775767-12775767 on Build GRCh38
rs533656677 Chr.9: 12775847-12775847 on Build GRCh38
rs1043557818 Chr.9: 12776130-12776130 on Build GRCh38
rs891052461 Chr.9: 12776182-12776182 on Build GRCh38
rs749750519 Chr.9: 12775799-12775799 on Build GRCh38
rs574948841 Chr.9: 12776116-12776116 on Build GRCh38
rs769385779 Chr.9: 12775925-12775925 on Build GRCh38
rs75227666 Chr.9: 12775895-12775895 on Build GRCh38
rs775587418 Chr.9: 12775927-12775927 on Build GRCh38
rs770436223 Chr.9: 12775813-12775813 on Build GRCh38
rs577351025 Chr.9: 12776109-12776109 on Build GRCh38
rs768615218 Chr.9: 12775947-12775947 on Build GRCh38
rs780649620 Chr.9: 12775910-12775910 on Build GRCh38
rs572021640 Chr.9: 12775856-12775856 on Build GRCh38
rs778078503 Chr.9: 12775898-12775898 on Build GRCh38
rs756918427 Chr.9: 12775900-12775900 on Build GRCh38
rs776200067 Chr.9: 12775862-12775862 on Build GRCh38
rs771725447 Chr.9: 12776008-12776008 on Build GRCh38
rs769928103 Chr.9: 12776026-12776026 on Build GRCh38
rs377029141 Chr.9: 12776053-12776053 on Build GRCh38
rs755731741 Chr.9: 12775984-12775984 on Build GRCh38
rs560683720 Chr.9: 12776144-12776144 on Build GRCh38
rs767749277 Chr.9: 12776048-12776048 on Build GRCh38
rs372303319 Chr.9: 12776203-12776203 on Build GRCh38
rs577160285 Chr.9: 12776156-12776156 on Build GRCh38
rs764588878 Chr.9: 12776039-12776039 on Build GRCh38
rs768377441 Chr.9: 12775926-12775928 on Build GRCh38
rs758901842 Chr.9: 12775969-12775969 on Build GRCh38
rs750127973 Chr.9: 12776049-12776049 on Build GRCh38
rs751967116 Chr.9: 12775973-12775973 on Build GRCh38
rs920831163 Chr.9: 12775794-12775794 on Build GRCh38
rs776154291 Chr.9: 12775844-12775845 on Build GRCh38
rs927935062 Chr.9: 12776086-12776086 on Build GRCh38
rs747987088 Chr.9: 12776077-12776077 on Build GRCh38
rs576435207 Chr.9: 12776047-12776047 on Build GRCh38
rs765252228 Chr.9: 12775963-12775963 on Build GRCh38
rs202014896 Chr.9: 12775995-12775995 on Build GRCh38
rs902393345 Chr.9: 12776159-12776159 on Build GRCh38
rs777434609 Chr.9: 12775854-12775854 on Build GRCh38
rs748950577 Chr.9: 12775886-12775887 on Build GRCh38
rs745384628 Chr.9: 12775924-12775924 on Build GRCh38
rs3750501 Chr.9: 12775890-12775890 on Build GRCh38
rs951335773 Chr.9: 12776179-12776195 on Build GRCh38
rs199677889 Chr.9: 12775853-12775854 on Build GRCh38
rs772358620 Chr.9: 12775853-12775853 on Build GRCh38
rs781478824 Chr.9: 12775782-12775782 on Build GRCh38
rs760720545 Chr.9: 12775802-12775802 on Build GRCh38
rs752956210 Chr.9: 12775863-12775864 on Build GRCh38
rs765007237 Chr.9: 12775830-12775830 on Build GRCh38
rs985183267 Chr.9: 12775906-12775906 on Build GRCh38
rs780163680 Chr.9: 12775846-12775846 on Build GRCh38
rs760923198 Chr.9: 12775886-12775886 on Build GRCh38
rs745389792 Chr.9: 12775874-12775875 on Build GRCh38
rs202004319 Chr.9: 12776018-12776018 on Build GRCh38
rs759015410 Chr.9: 12776046-12776046 on Build GRCh38
rs747983719 Chr.9: 12776007-12776007 on Build GRCh38
rs745336606 Chr.9: 12775851-12775852 on Build GRCh38
rs772795261 Chr.9: 12775809-12775809 on Build GRCh38
rs751869777 Chr.9: 12775780-12775780 on Build GRCh38
rs762963213 Chr.9: 12776036-12776036 on Build GRCh38
rs997924253 Chr.9: 12775801-12775801 on Build GRCh38
rs756167380 Chr.9: 12775845-12775845 on Build GRCh38
rs753252927 Chr.9: 12775966-12775966 on Build GRCh38
rs768703196 Chr.9: 12775999-12775999 on Build GRCh38
rs995700561 Chr.9: 12776190-12776190 on Build GRCh38
rs373322755 Chr.9: 12775990-12775990 on Build GRCh38
rs778661483 Chr.9: 12776003-12776003 on Build GRCh38
rs200424165 Chr.9: 12775884-12775884 on Build GRCh38
rs17853715 Chr.9: 12775899-12775899 on Build GRCh38
rs750628608 Chr.9: 12775843-12775843 on Build GRCh38
rs752525770 Chr.9: 12775835-12775835 on Build GRCh38
rs555618559 Chr.9: 12776115-12776115 on Build GRCh38
rs116787446 Chr.9: 12775804-12775804 on Build GRCh38
rs760866779 Chr.9: 12775955-12775955 on Build GRCh38
rs1026468879 Chr.9: 12776137-12776137 on Build GRCh38
rs752097780 Chr.9: 12775868-12775869 on Build GRCh38
rs775499589 Chr.9: 12775877-12775877 on Build GRCh38
rs112112353 Chr.9: 12776122-12776122 on Build GRCh38
rs952142933 Chr.9: 12775896-12775896 on Build GRCh38
rs776651254 Chr.9: 12775957-12775957 on Build GRCh38
rs778173403 Chr.9: 12775865-12775866 on Build GRCh38
rs760183432 Chr.9: 12775812-12775812 on Build GRCh38
rs755827663 Chr.9: 12776052-12776052 on Build GRCh38
rs759485730 Chr.9: 12775829-12775829 on Build GRCh38
rs73403628 Chr.9: 12776104-12776104 on Build GRCh38
rs201963967 Chr.9: 12775881-12775881 on Build GRCh38
rs1039855195 Chr.9: 12775859-12775859 on Build GRCh38
rs935790832 Chr.9: 12775998-12775998 on Build GRCh38
rs759447438 Chr.9: 12775863-12775863 on Build GRCh38
rs780453238 Chr.9: 12775795-12775795 on Build GRCh38
rs370041576 Chr.9: 12775933-12775933 on Build GRCh38
rs902923494 Chr.9: 12775985-12775985 on Build GRCh38
rs757496573 Chr.9: 12775976-12775976 on Build GRCh38
rs776934630 Chr.9: 12776011-12776011 on Build GRCh38
rs762724934 Chr.9: 12775859-12775860 on Build GRCh38
rs139315731 Chr.9: 12775862-12775863 on Build GRCh38
rs866999155 Chr.9: 12775975-12775975 on Build GRCh38
rs753943046 Chr.9: 12775889-12775889 on Build GRCh38
rs973556094 Chr.9: 12776012-12776012 on Build GRCh38
rs182999630 Chr.9: 12776186-12776186 on Build GRCh38
rs774287296 Chr.9: 12775953-12775953 on Build GRCh38
rs761650878 Chr.9: 12776028-12776031 on Build GRCh38
rs754475530 Chr.9: 12776059-12776059 on Build GRCh38
rs779633177 Chr.9: 12775884-12775884 on Build GRCh38
rs753162910 Chr.9: 12775892-12775892 on Build GRCh38
rs780925893 Chr.9: 12775980-12775980 on Build GRCh38
rs775532901 Chr.9: 12776034-12776034 on Build GRCh38
rs774319753 Chr.9: 12775957-12775957 on Build GRCh38
rs769345710 Chr.9: 12775878-12775879 on Build GRCh38
rs534390977 Chr.9: 12775865-12775865 on Build GRCh38
rs1050992760 Chr.9: 12776179-12776179 on Build GRCh38
rs149696691 Chr.9: 12775771-12775771 on Build GRCh38
rs770473914 Chr.9: 12775820-12775821 on Build GRCh38
rs964705494 Chr.9: 12776212-12776212 on Build GRCh38
rs781623805 Chr.9: 12775873-12775874 on Build GRCh38
rs901043740 Chr.9: 12775774-12775774 on Build GRCh38
rs771702525 Chr.9: 12775808-12775808 on Build GRCh38
rs751967998 Chr.9: 12776044-12776044 on Build GRCh38
rs766439880 Chr.9: 12775887-12775887 on Build GRCh38
rs767853430 Chr.9: 12775839-12775839 on Build GRCh38
rs759694236 Chr.9: 12775962-12775962 on Build GRCh38
rs763929087 Chr.9: 12775882-12775882 on Build GRCh38
rs531772186 Chr.9: 12775792-12775792 on Build GRCh38
rs894176643 Chr.9: 12776085-12776085 on Build GRCh38
rs758744192 Chr.9: 12775864-12775865 on Build GRCh38
rs553686180 Chr.9: 12775850-12775850 on Build GRCh38
rs778941900 Chr.9: 12776074-12776074 on Build GRCh38

View Full Product Details