Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72202475-72203022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDA
Entrez Gene ID
9615
Gene Name
guanine deaminase
Gene Aliases
CYPIN, GUANASE, NEDASIN
UniGene
Hs.494163
Gene Chromosome Location
Chr.9: 72114595-72256045 on Build GRCh38
Transcripts
NM_001242505.2 NM_001242506.2 NM_001242507.2 NM_004293.4 XM_005252317.1 XM_011519213.1 XM_011519214.1 XM_011519215.1 XM_011519216.1 XM_011519217.1 XM_011519218.2 XM_011519219.1 XM_011519220.1 XM_011519221.1 XM_011519222.1 XM_017015333.1 XM_017015334.1 XM_017015335.1 XM_017015336.1 XM_017015337.1 XM_017015338.1 XM_017015339.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1109966 c.230G>A p.G77E Chr.9: 72202588-72202588 on Build GRCh38
4432417 c.251A>G p.H84R Chr.9: 72202609-72202609 on Build GRCh38
1109964 c.219C>A p.F73L Chr.9: 72202577-72202577 on Build GRCh38
5890420 c.325C>T p.P109S Chr.9: 72202683-72202683 on Build GRCh38
4416937 c.326C>T p.P109L Chr.9: 72202684-72202684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766142263 Chr.9: 72202764-72202769 on Build GRCh38
rs77854645 Chr.9: 72202936-72202936 on Build GRCh38
rs143550458 Chr.9: 72202875-72202875 on Build GRCh38
rs938783137 Chr.9: 72202804-72202804 on Build GRCh38
rs529545864 Chr.9: 72202604-72202604 on Build GRCh38
rs763084635 Chr.9: 72202657-72202657 on Build GRCh38
rs1028103822 Chr.9: 72202957-72202957 on Build GRCh38
rs755331133 Chr.9: 72202610-72202610 on Build GRCh38
rs896227192 Chr.9: 72202826-72202826 on Build GRCh38
rs752036338 Chr.9: 72202854-72202854 on Build GRCh38
rs776767059 Chr.9: 72202520-72202520 on Build GRCh38
rs758772378 Chr.9: 72202841-72202841 on Build GRCh38
rs773222504 Chr.9: 72202653-72202653 on Build GRCh38
rs1041723139 Chr.9: 72202788-72202788 on Build GRCh38
rs149388704 Chr.9: 72202761-72202761 on Build GRCh38
rs1031238767 Chr.9: 72202710-72202710 on Build GRCh38
rs762914194 Chr.9: 72202683-72202683 on Build GRCh38
rs761091832 Chr.9: 72202774-72202774 on Build GRCh38
rs762960160 Chr.9: 72202759-72202759 on Build GRCh38
rs1015281861 Chr.9: 72202920-72202920 on Build GRCh38
rs747963673 Chr.9: 72202752-72202752 on Build GRCh38
rs1030894587 Chr.9: 72202906-72202906 on Build GRCh38
rs75382038 Chr.9: 72202758-72202758 on Build GRCh38
rs779979867 Chr.9: 72202715-72202715 on Build GRCh38
rs751048421 Chr.9: 72202784-72202784 on Build GRCh38
rs371223443 Chr.9: 72202659-72202659 on Build GRCh38
rs779961418 Chr.9: 72202644-72202644 on Build GRCh38
rs750486743 Chr.9: 72202557-72202557 on Build GRCh38
rs199540013 Chr.9: 72202539-72202539 on Build GRCh38
rs768112787 Chr.9: 72202767-72202767 on Build GRCh38
rs745358144 Chr.9: 72202642-72202642 on Build GRCh38
rs985092341 Chr.9: 72202769-72202769 on Build GRCh38
rs146422167 Chr.9: 72202651-72202651 on Build GRCh38
rs764800573 Chr.9: 72202775-72202775 on Build GRCh38
rs1029398466 Chr.9: 72202992-72202992 on Build GRCh38
rs771958358 Chr.9: 72202643-72202643 on Build GRCh38
rs753618221 Chr.9: 72202778-72202781 on Build GRCh38
rs375585337 Chr.9: 72202550-72202550 on Build GRCh38
rs986356750 Chr.9: 72202980-72202980 on Build GRCh38
rs566312919 Chr.9: 72202926-72202926 on Build GRCh38
rs1052956672 Chr.9: 72202769-72202791 on Build GRCh38
rs113810444 Chr.9: 72202731-72202731 on Build GRCh38
rs996298846 Chr.9: 72202978-72202978 on Build GRCh38
rs530521050 Chr.9: 72202677-72202677 on Build GRCh38
rs753494302 Chr.9: 72202633-72202633 on Build GRCh38
rs750356146 Chr.9: 72202756-72202757 on Build GRCh38
rs758880816 Chr.9: 72202566-72202566 on Build GRCh38
rs752037406 Chr.9: 72202592-72202592 on Build GRCh38
rs756826268 Chr.9: 72202640-72202640 on Build GRCh38
rs966098008 Chr.9: 72202961-72202961 on Build GRCh38
rs1000952962 Chr.9: 72202880-72202880 on Build GRCh38
rs781016341 Chr.9: 72202741-72202741 on Build GRCh38
rs377678116 Chr.9: 72202556-72202556 on Build GRCh38
rs975141723 Chr.9: 72202757-72202757 on Build GRCh38
rs775031438 Chr.9: 72202762-72202762 on Build GRCh38
rs138792393 Chr.9: 72202678-72202678 on Build GRCh38
rs746898915 Chr.9: 72202727-72202727 on Build GRCh38
rs766896532 Chr.9: 72202581-72202581 on Build GRCh38
rs776859539 Chr.9: 72202646-72202646 on Build GRCh38
rs952725071 Chr.9: 72202511-72202511 on Build GRCh38
rs368567455 Chr.9: 72202782-72202782 on Build GRCh38
rs753059474 Chr.9: 72202768-72202768 on Build GRCh38
rs956509172 Chr.9: 72202720-72202720 on Build GRCh38
rs940817400 Chr.9: 72202800-72202800 on Build GRCh38
rs755554741 Chr.9: 72202868-72202868 on Build GRCh38
rs191364910 Chr.9: 72202979-72202979 on Build GRCh38
rs767384214 Chr.9: 72202718-72202720 on Build GRCh38
rs778323963 Chr.9: 72202641-72202641 on Build GRCh38
rs914861052 Chr.9: 72202780-72202780 on Build GRCh38
rs777883667 Chr.9: 72202785-72202786 on Build GRCh38
rs777586157 Chr.9: 72202613-72202613 on Build GRCh38
rs767957340 Chr.9: 72202673-72202673 on Build GRCh38
rs749965441 Chr.9: 72202707-72202707 on Build GRCh38
rs371294481 Chr.9: 72202754-72202754 on Build GRCh38
rs1048700539 Chr.9: 72202842-72202842 on Build GRCh38
rs767002186 Chr.9: 72202668-72202668 on Build GRCh38
rs1007983619 Chr.9: 72202925-72202925 on Build GRCh38
rs1011883867 Chr.9: 72202514-72202514 on Build GRCh38
rs892384197 Chr.9: 72202912-72202912 on Build GRCh38
rs372372452 Chr.9: 72202783-72202783 on Build GRCh38
rs894546064 Chr.9: 72202835-72202835 on Build GRCh38
rs758608855 Chr.9: 72202783-72202784 on Build GRCh38
rs1023332333 Chr.9: 72202519-72202519 on Build GRCh38
rs1018944322 Chr.9: 72202753-72202753 on Build GRCh38
rs761964345 Chr.9: 72202542-72202542 on Build GRCh38
rs778610772 Chr.9: 72202700-72202700 on Build GRCh38
rs61758972 Chr.9: 72202670-72202670 on Build GRCh38
rs754968351 Chr.9: 72202792-72202792 on Build GRCh38
rs761986169 Chr.9: 72202709-72202709 on Build GRCh38
rs77548027 Chr.9: 72202645-72202645 on Build GRCh38
rs368070896 Chr.9: 72202730-72202730 on Build GRCh38
rs768469943 Chr.9: 72202736-72202736 on Build GRCh38
rs1056653468 Chr.9: 72202814-72202814 on Build GRCh38
rs760664067 Chr.9: 72202759-72202760 on Build GRCh38

View Full Product Details