Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134030189-134030694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD3
Entrez Gene ID
8019
Gene Name
bromodomain containing 3
Gene Aliases
ORFX, RING3L
UniGene
Hs.522472
Gene Chromosome Location
Chr.9: 134030305-134068533 on Build GRCh38
Transcripts
NM_007371.3 XM_017015165.1 XM_017015166.1 NR_015427.2

Gene Symbol
LINC00094
Entrez Gene ID
266655
Gene Name
long intergenic non-protein coding RNA 94
Gene Aliases
LP2477, NCRNA00094, bA374P20.3
UniGene
Hs.654869
Gene Chromosome Location
Chr.9: 134025439-134031597 on Build GRCh38
Transcripts
NM_007371.3 XM_017015165.1 XM_017015166.1 NR_015427.2

SNP Detail

SNP ID Chromosome Location
rs907573265 Chr.9: 134030654-134030654 on Build GRCh38
rs1018845445 Chr.9: 134030590-134030590 on Build GRCh38
rs1044880009 Chr.9: 134030438-134030438 on Build GRCh38
rs796946313 Chr.9: 134030236-134030236 on Build GRCh38
rs543214445 Chr.9: 134030650-134030650 on Build GRCh38
rs930312971 Chr.9: 134030243-134030243 on Build GRCh38
rs950194890 Chr.9: 134030416-134030416 on Build GRCh38
rs1010187309 Chr.9: 134030553-134030553 on Build GRCh38
rs745379292 Chr.9: 134030620-134030620 on Build GRCh38
rs773990406 Chr.9: 134030220-134030220 on Build GRCh38
rs755277498 Chr.9: 134030598-134030598 on Build GRCh38
rs534099703 Chr.9: 134030509-134030509 on Build GRCh38
rs113349010 Chr.9: 134030388-134030388 on Build GRCh38
rs368037755 Chr.9: 134030300-134030300 on Build GRCh38
rs11792352 Chr.9: 134030303-134030303 on Build GRCh38
rs922374218 Chr.9: 134030266-134030266 on Build GRCh38
rs371929806 Chr.9: 134030287-134030288 on Build GRCh38
rs965855341 Chr.9: 134030218-134030218 on Build GRCh38
rs796701551 Chr.9: 134030324-134030325 on Build GRCh38
rs577961572 Chr.9: 134030526-134030526 on Build GRCh38
rs1037230636 Chr.9: 134030308-134030308 on Build GRCh38
rs907544860 Chr.9: 134030253-134030253 on Build GRCh38
rs917426951 Chr.9: 134030366-134030366 on Build GRCh38
rs563751951 Chr.9: 134030544-134030544 on Build GRCh38
rs940299653 Chr.9: 134030300-134030301 on Build GRCh38
rs541268917 Chr.9: 134030484-134030484 on Build GRCh38
rs908380431 Chr.9: 134030233-134030233 on Build GRCh38
rs984507190 Chr.9: 134030248-134030248 on Build GRCh38
rs139953305 Chr.9: 134030326-134030327 on Build GRCh38
rs905915017 Chr.9: 134030471-134030471 on Build GRCh38
rs376809450 Chr.9: 134030300-134030300 on Build GRCh38
rs58289869 Chr.9: 134030241-134030241 on Build GRCh38
rs867784405 Chr.9: 134030287-134030287 on Build GRCh38
rs181816238 Chr.9: 134030523-134030523 on Build GRCh38
rs772997700 Chr.9: 134030536-134030536 on Build GRCh38
rs781707774 Chr.9: 134030371-134030371 on Build GRCh38
rs766934169 Chr.9: 134030299-134030300 on Build GRCh38
rs545466382 Chr.9: 134030537-134030537 on Build GRCh38
rs199899839 Chr.9: 134030327-134030327 on Build GRCh38
rs12554235 Chr.9: 134030672-134030672 on Build GRCh38
rs71503376 Chr.9: 134030304-134030305 on Build GRCh38
rs891783750 Chr.9: 134030542-134030542 on Build GRCh38
rs916224676 Chr.9: 134030326-134030326 on Build GRCh38
rs995989498 Chr.9: 134030598-134030598 on Build GRCh38
rs1046538331 Chr.9: 134030333-134030333 on Build GRCh38
rs575821996 Chr.9: 134030612-134030612 on Build GRCh38
rs78579971 Chr.9: 134030298-134030299 on Build GRCh38
rs961658457 Chr.9: 134030664-134030664 on Build GRCh38
rs771639713 Chr.9: 134030425-134030425 on Build GRCh38

View Full Product Details