Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128959023-128959485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP188
Entrez Gene ID
23511
Gene Name
nucleoporin 188
Gene Aliases
KIAA0169, hNup188
UniGene
Hs.308340
Gene Chromosome Location
Chr.9: 128947693-129007096 on Build GRCh38
Transcripts
NM_015354.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
421943 c.487G>C p.D163H Chr.9: 128959036-128959036 on Build GRCh38
1105777 c.486T>C p.V162V Chr.9: 128959035-128959035 on Build GRCh38
1105778 c.509T>A p.V170D Chr.9: 128959058-128959058 on Build GRCh38
399584 c.563G>T p.W188L Chr.9: 128959112-128959112 on Build GRCh38
5410505 c.575G>A p.G192E Chr.9: 128959124-128959124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927327860 Chr.9: 128959361-128959361 on Build GRCh38
rs1028525297 Chr.9: 128959270-128959270 on Build GRCh38
rs944234641 Chr.9: 128959367-128959367 on Build GRCh38
rs775225785 Chr.9: 128959116-128959116 on Build GRCh38
rs754111677 Chr.9: 128959443-128959443 on Build GRCh38
rs952694773 Chr.9: 128959345-128959345 on Build GRCh38
rs938717979 Chr.9: 128959070-128959070 on Build GRCh38
rs916675653 Chr.9: 128959224-128959224 on Build GRCh38
rs149242943 Chr.9: 128959384-128959384 on Build GRCh38
rs146233640 Chr.9: 128959362-128959362 on Build GRCh38
rs972655504 Chr.9: 128959313-128959313 on Build GRCh38
rs762740256 Chr.9: 128959121-128959121 on Build GRCh38
rs1050618800 Chr.9: 128959435-128959435 on Build GRCh38
rs767685652 Chr.9: 128959168-128959168 on Build GRCh38
rs938764413 Chr.9: 128959368-128959368 on Build GRCh38
rs531582557 Chr.9: 128959285-128959285 on Build GRCh38
rs372344068 Chr.9: 128959157-128959157 on Build GRCh38
rs549693283 Chr.9: 128959456-128959456 on Build GRCh38
rs532359135 Chr.9: 128959080-128959080 on Build GRCh38
rs372052732 Chr.9: 128959132-128959132 on Build GRCh38
rs776348954 Chr.9: 128959163-128959163 on Build GRCh38
rs1010503804 Chr.9: 128959191-128959191 on Build GRCh38
rs767303965 Chr.9: 128959299-128959299 on Build GRCh38
rs935570068 Chr.9: 128959316-128959316 on Build GRCh38
rs1014713251 Chr.9: 128959145-128959145 on Build GRCh38
rs759371132 Chr.9: 128959162-128959163 on Build GRCh38
rs750494053 Chr.9: 128959175-128959175 on Build GRCh38
rs1057153966 Chr.9: 128959083-128959083 on Build GRCh38
rs754201611 Chr.9: 128959179-128959179 on Build GRCh38
rs764210249 Chr.9: 128959147-128959147 on Build GRCh38
rs150109090 Chr.9: 128959290-128959290 on Build GRCh38
rs568882179 Chr.9: 128959433-128959433 on Build GRCh38
rs983298507 Chr.9: 128959108-128959108 on Build GRCh38
rs146286782 Chr.9: 128959427-128959427 on Build GRCh38
rs756651113 Chr.9: 128959177-128959177 on Build GRCh38
rs769744217 Chr.9: 128959093-128959093 on Build GRCh38
rs566532905 Chr.9: 128959428-128959428 on Build GRCh38
rs948139169 Chr.9: 128959287-128959287 on Build GRCh38
rs996714986 Chr.9: 128959432-128959432 on Build GRCh38
rs767002233 Chr.9: 128959178-128959178 on Build GRCh38
rs562532363 Chr.9: 128959252-128959252 on Build GRCh38
rs746484535 Chr.9: 128959061-128959061 on Build GRCh38
rs138409015 Chr.9: 128959294-128959294 on Build GRCh38
rs1000617080 Chr.9: 128959263-128959263 on Build GRCh38
rs148698768 Chr.9: 128959053-128959053 on Build GRCh38
rs569328631 Chr.9: 128959460-128959460 on Build GRCh38
rs1057125372 Chr.9: 128959369-128959369 on Build GRCh38
rs770538635 Chr.9: 128959076-128959076 on Build GRCh38
rs986390768 Chr.9: 128959291-128959291 on Build GRCh38
rs761768950 Chr.9: 128959166-128959166 on Build GRCh38
rs186298929 Chr.9: 128959439-128959439 on Build GRCh38
rs980205382 Chr.9: 128959348-128959348 on Build GRCh38
rs961054159 Chr.9: 128959459-128959459 on Build GRCh38
rs1021787908 Chr.9: 128959197-128959197 on Build GRCh38
rs946261861 Chr.9: 128959385-128959385 on Build GRCh38
rs919753747 Chr.9: 128959328-128959328 on Build GRCh38
rs1019228284 Chr.9: 128959453-128959453 on Build GRCh38
rs1027353455 Chr.9: 128959461-128959461 on Build GRCh38

View Full Product Details