Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129858245-129858745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP20
Entrez Gene ID
10868
Gene Name
ubiquitin specific peptidase 20
Gene Aliases
LSFR3A, VDU2, hVDU2
UniGene
Hs.5452
Gene Chromosome Location
Chr.9: 129835417-129881838 on Build GRCh38
Transcripts
NM_001008563.4 NM_001110303.3 NM_006676.7 XM_005251665.3 XM_005251666.3 XM_011518161.2 XM_011518162.2 XM_017014221.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2149540 c.290_291delCA p.A97fs*12 Chr.9: 129858558-129858559 on Build GRCh38
2149541 c.290_291delCA p.A97fs*12 Chr.9: 129858558-129858559 on Build GRCh38
6837559 c.330G>C p.Q110H Chr.9: 129858598-129858598 on Build GRCh38
5749461 c.199G>A p.A67T Chr.9: 129858467-129858467 on Build GRCh38
6818339 c.239G>A p.R80Q Chr.9: 129858507-129858507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749631422 Chr.9: 129858633-129858633 on Build GRCh38
rs2174156 Chr.9: 129858341-129858341 on Build GRCh38
rs770716278 Chr.9: 129858552-129858552 on Build GRCh38
rs200946773 Chr.9: 129858520-129858520 on Build GRCh38
rs746825367 Chr.9: 129858551-129858551 on Build GRCh38
rs908731161 Chr.9: 129858394-129858394 on Build GRCh38
rs781726860 Chr.9: 129858518-129858519 on Build GRCh38
rs1020100708 Chr.9: 129858567-129858567 on Build GRCh38
rs111838494 Chr.9: 129858364-129858364 on Build GRCh38
rs915011203 Chr.9: 129858347-129858347 on Build GRCh38
rs146591850 Chr.9: 129858460-129858460 on Build GRCh38
rs544534698 Chr.9: 129858393-129858393 on Build GRCh38
rs985846451 Chr.9: 129858677-129858677 on Build GRCh38
rs985513319 Chr.9: 129858372-129858372 on Build GRCh38
rs1031287859 Chr.9: 129858319-129858319 on Build GRCh38
rs142119169 Chr.9: 129858321-129858321 on Build GRCh38
rs922232341 Chr.9: 129858370-129858370 on Build GRCh38
rs941478351 Chr.9: 129858390-129858390 on Build GRCh38
rs963736471 Chr.9: 129858349-129858349 on Build GRCh38
rs577966827 Chr.9: 129858402-129858402 on Build GRCh38
rs975290489 Chr.9: 129858273-129858273 on Build GRCh38
rs764693521 Chr.9: 129858477-129858477 on Build GRCh38
rs762300776 Chr.9: 129858579-129858579 on Build GRCh38
rs763611791 Chr.9: 129858462-129858462 on Build GRCh38
rs182914258 Chr.9: 129858508-129858508 on Build GRCh38
rs771788261 Chr.9: 129858302-129858307 on Build GRCh38
rs35953314 Chr.9: 129858572-129858572 on Build GRCh38
rs746306617 Chr.9: 129858520-129858520 on Build GRCh38
rs36086252 Chr.9: 129858576-129858576 on Build GRCh38
rs983461775 Chr.9: 129858625-129858625 on Build GRCh38
rs768052334 Chr.9: 129858527-129858527 on Build GRCh38
rs201578380 Chr.9: 129858568-129858568 on Build GRCh38
rs746806972 Chr.9: 129858429-129858429 on Build GRCh38
rs75346745 Chr.9: 129858437-129858437 on Build GRCh38
rs758400384 Chr.9: 129858536-129858536 on Build GRCh38
rs191133247 Chr.9: 129858488-129858488 on Build GRCh38
rs773145798 Chr.9: 129858304-129858309 on Build GRCh38
rs771187433 Chr.9: 129858430-129858430 on Build GRCh38
rs908811417 Chr.9: 129858647-129858647 on Build GRCh38
rs374658775 Chr.9: 129858559-129858559 on Build GRCh38
rs752193950 Chr.9: 129858521-129858521 on Build GRCh38
rs982050852 Chr.9: 129858317-129858317 on Build GRCh38
rs368354996 Chr.9: 129858453-129858453 on Build GRCh38
rs752011584 Chr.9: 129858487-129858487 on Build GRCh38
rs1014490692 Chr.9: 129858287-129858287 on Build GRCh38
rs957176004 Chr.9: 129858314-129858314 on Build GRCh38
rs770324038 Chr.9: 129858528-129858530 on Build GRCh38
rs571480207 Chr.9: 129858502-129858502 on Build GRCh38
rs5900868 Chr.9: 129858301-129858306 on Build GRCh38
rs538017151 Chr.9: 129858674-129858674 on Build GRCh38
rs769843480 Chr.9: 129858458-129858458 on Build GRCh38
rs763093813 Chr.9: 129858573-129858573 on Build GRCh38
rs1001252292 Chr.9: 129858303-129858303 on Build GRCh38
rs986915109 Chr.9: 129858610-129858610 on Build GRCh38
rs540572169 Chr.9: 129858409-129858409 on Build GRCh38
rs769893010 Chr.9: 129858564-129858564 on Build GRCh38
rs765971028 Chr.9: 129858493-129858493 on Build GRCh38
rs777660423 Chr.9: 129858550-129858550 on Build GRCh38
rs948819465 Chr.9: 129858700-129858700 on Build GRCh38
rs970638118 Chr.9: 129858311-129858311 on Build GRCh38
rs2134601 Chr.9: 129858428-129858428 on Build GRCh38
rs113893347 Chr.9: 129858612-129858612 on Build GRCh38
rs41279144 Chr.9: 129858392-129858392 on Build GRCh38
rs1024281110 Chr.9: 129858278-129858278 on Build GRCh38
rs558169132 Chr.9: 129858689-129858689 on Build GRCh38
rs759446315 Chr.9: 129858300-129858301 on Build GRCh38
rs775781572 Chr.9: 129858625-129858627 on Build GRCh38
rs932653001 Chr.9: 129858350-129858350 on Build GRCh38
rs950280877 Chr.9: 129858408-129858408 on Build GRCh38
rs909954751 Chr.9: 129858378-129858378 on Build GRCh38
rs746041872 Chr.9: 129858563-129858563 on Build GRCh38
rs187572353 Chr.9: 129858399-129858399 on Build GRCh38
rs759279205 Chr.9: 129858621-129858621 on Build GRCh38
rs916039831 Chr.9: 129858696-129858696 on Build GRCh38
rs560280675 Chr.9: 129858411-129858411 on Build GRCh38
rs77339429 Chr.9: 129858620-129858620 on Build GRCh38
rs1038553835 Chr.9: 129858695-129858695 on Build GRCh38
rs2174157 Chr.9: 129858711-129858711 on Build GRCh38
rs975056541 Chr.9: 129858353-129858353 on Build GRCh38
rs139106007 Chr.9: 129858631-129858631 on Build GRCh38
rs200364798 Chr.9: 129858577-129858577 on Build GRCh38
rs551714401 Chr.9: 129858496-129858496 on Build GRCh38
rs976008762 Chr.9: 129858342-129858342 on Build GRCh38

View Full Product Details