Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153803277-153803776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZD4
Entrez Gene ID
57595
Gene Name
PDZ domain containing 4
Gene Aliases
LNX5, LU1, PDZK4, PDZRN4L
UniGene
Hs.92732
Gene Chromosome Location
Chr.X: 153802166-153830567 on Build GRCh38
Transcripts
NM_001303512.1 NM_001303513.1 NM_001303514.1 NM_001303515.1 NM_001303516.1 NM_032512.3

SNP Detail

SNP ID Chromosome Location
rs782116183 Chr.X: 153803455-153803455 on Build GRCh38
rs375303161 Chr.X: 153803743-153803743 on Build GRCh38
rs782358549 Chr.X: 153803685-153803685 on Build GRCh38
rs781785549 Chr.X: 153803533-153803533 on Build GRCh38
rs376042669 Chr.X: 153803410-153803410 on Build GRCh38
rs782134177 Chr.X: 153803728-153803728 on Build GRCh38
rs782807031 Chr.X: 153803543-153803543 on Build GRCh38
rs781877261 Chr.X: 153803525-153803525 on Build GRCh38
rs375486951 Chr.X: 153803542-153803542 on Build GRCh38
rs782044820 Chr.X: 153803746-153803746 on Build GRCh38
rs782010694 Chr.X: 153803332-153803332 on Build GRCh38
rs781853495 Chr.X: 153803352-153803352 on Build GRCh38
rs782272511 Chr.X: 153803413-153803413 on Build GRCh38
rs782487475 Chr.X: 153803319-153803320 on Build GRCh38
rs140408896 Chr.X: 153803526-153803526 on Build GRCh38
rs781793305 Chr.X: 153803376-153803376 on Build GRCh38
rs782222596 Chr.X: 153803571-153803571 on Build GRCh38
rs782413110 Chr.X: 153803416-153803416 on Build GRCh38
rs782564473 Chr.X: 153803302-153803302 on Build GRCh38
rs926733236 Chr.X: 153803437-153803437 on Build GRCh38
rs782645559 Chr.X: 153803334-153803334 on Build GRCh38
rs782677370 Chr.X: 153803529-153803529 on Build GRCh38
rs782196573 Chr.X: 153803310-153803310 on Build GRCh38
rs888874648 Chr.X: 153803605-153803605 on Build GRCh38
rs377300947 Chr.X: 153803440-153803440 on Build GRCh38
rs940160103 Chr.X: 153803486-153803486 on Build GRCh38
rs781840928 Chr.X: 153803390-153803390 on Build GRCh38
rs781992337 Chr.X: 153803688-153803688 on Build GRCh38
rs146551513 Chr.X: 153803572-153803572 on Build GRCh38
rs782151064 Chr.X: 153803359-153803359 on Build GRCh38
rs370790531 Chr.X: 153803368-153803368 on Build GRCh38
rs992348488 Chr.X: 153803459-153803459 on Build GRCh38
rs141253880 Chr.X: 153803749-153803749 on Build GRCh38
rs781971869 Chr.X: 153803312-153803312 on Build GRCh38
rs151185706 Chr.X: 153803449-153803449 on Build GRCh38
rs371190527 Chr.X: 153803488-153803488 on Build GRCh38
rs782450743 Chr.X: 153803377-153803377 on Build GRCh38
rs199823164 Chr.X: 153803663-153803663 on Build GRCh38
rs782392156 Chr.X: 153803469-153803469 on Build GRCh38
rs782555949 Chr.X: 153803421-153803421 on Build GRCh38
rs968113390 Chr.X: 153803412-153803412 on Build GRCh38
rs782615389 Chr.X: 153803383-153803383 on Build GRCh38
rs782037246 Chr.X: 153803487-153803487 on Build GRCh38
rs782511562 Chr.X: 153803409-153803409 on Build GRCh38
rs781820312 Chr.X: 153803757-153803757 on Build GRCh38
rs371466435 Chr.X: 153803671-153803671 on Build GRCh38
rs781968798 Chr.X: 153803454-153803454 on Build GRCh38
rs781842955 Chr.X: 153803308-153803308 on Build GRCh38
rs782759924 Chr.X: 153803517-153803517 on Build GRCh38
rs781824314 Chr.X: 153803502-153803502 on Build GRCh38
rs956634876 Chr.X: 153803365-153803365 on Build GRCh38
rs782102008 Chr.X: 153803515-153803515 on Build GRCh38
rs994894464 Chr.X: 153803320-153803320 on Build GRCh38
rs782641912 Chr.X: 153803580-153803580 on Build GRCh38
rs144134931 Chr.X: 153803566-153803566 on Build GRCh38
rs908684186 Chr.X: 153803463-153803463 on Build GRCh38
rs367833672 Chr.X: 153803602-153803602 on Build GRCh38
rs782299423 Chr.X: 153803590-153803590 on Build GRCh38
rs782080476 Chr.X: 153803342-153803342 on Build GRCh38
rs368917754 Chr.X: 153803324-153803324 on Build GRCh38
rs375849010 Chr.X: 153803349-153803349 on Build GRCh38
rs782730420 Chr.X: 153803499-153803499 on Build GRCh38

View Full Product Details