Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153805418-153805924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZD4
Entrez Gene ID
57595
Gene Name
PDZ domain containing 4
Gene Aliases
LNX5, LU1, PDZK4, PDZRN4L
UniGene
Hs.92732
Gene Chromosome Location
Chr.X: 153802166-153830567 on Build GRCh38
Transcripts
NM_001303512.1 NM_001303513.1 NM_001303514.1 NM_001303515.1 NM_001303516.1 NM_032512.3

SNP Detail

SNP ID Chromosome Location
rs781839161 Chr.X: 153805531-153805531 on Build GRCh38
rs782404871 Chr.X: 153805611-153805611 on Build GRCh38
rs3747312 Chr.X: 153805544-153805544 on Build GRCh38
rs782725467 Chr.X: 153805520-153805520 on Build GRCh38
rs782203525 Chr.X: 153805571-153805571 on Build GRCh38
rs186738796 Chr.X: 153805874-153805874 on Build GRCh38
rs1010360209 Chr.X: 153805855-153805855 on Build GRCh38
rs578057261 Chr.X: 153805478-153805478 on Build GRCh38
rs369117434 Chr.X: 153805480-153805480 on Build GRCh38
rs782664074 Chr.X: 153805538-153805538 on Build GRCh38
rs1041093958 Chr.X: 153805821-153805821 on Build GRCh38
rs782591441 Chr.X: 153805559-153805559 on Build GRCh38
rs782220399 Chr.X: 153805684-153805684 on Build GRCh38
rs377442810 Chr.X: 153805577-153805577 on Build GRCh38
rs376192609 Chr.X: 153805778-153805778 on Build GRCh38
rs199686127 Chr.X: 153805472-153805472 on Build GRCh38
rs782293794 Chr.X: 153805587-153805587 on Build GRCh38
rs782777662 Chr.X: 153805627-153805627 on Build GRCh38
rs112383108 Chr.X: 153805699-153805699 on Build GRCh38
rs782506536 Chr.X: 153805537-153805537 on Build GRCh38
rs782258050 Chr.X: 153805591-153805591 on Build GRCh38
rs901375612 Chr.X: 153805854-153805854 on Build GRCh38
rs782131092 Chr.X: 153805757-153805757 on Build GRCh38
rs942768972 Chr.X: 153805766-153805766 on Build GRCh38
rs782205628 Chr.X: 153805665-153805665 on Build GRCh38
rs782336806 Chr.X: 153805887-153805887 on Build GRCh38
rs782046403 Chr.X: 153805696-153805696 on Build GRCh38
rs3747310 Chr.X: 153805896-153805896 on Build GRCh38
rs782623327 Chr.X: 153805479-153805480 on Build GRCh38
rs782436345 Chr.X: 153805547-153805547 on Build GRCh38
rs3747311 Chr.X: 153805669-153805669 on Build GRCh38
rs782094201 Chr.X: 153805475-153805475 on Build GRCh38
rs781956893 Chr.X: 153805620-153805620 on Build GRCh38
rs782175269 Chr.X: 153805458-153805458 on Build GRCh38
rs782056937 Chr.X: 153805616-153805616 on Build GRCh38
rs782306702 Chr.X: 153805497-153805497 on Build GRCh38
rs782328094 Chr.X: 153805618-153805618 on Build GRCh38
rs201564498 Chr.X: 153805582-153805582 on Build GRCh38
rs782684948 Chr.X: 153805438-153805438 on Build GRCh38
rs782069105 Chr.X: 153805492-153805492 on Build GRCh38
rs782359050 Chr.X: 153805698-153805698 on Build GRCh38
rs782166390 Chr.X: 153805647-153805647 on Build GRCh38
rs782008059 Chr.X: 153805479-153805479 on Build GRCh38
rs782099194 Chr.X: 153805626-153805626 on Build GRCh38
rs781886187 Chr.X: 153805643-153805643 on Build GRCh38
rs782411376 Chr.X: 153805592-153805592 on Build GRCh38
rs782316198 Chr.X: 153805471-153805471 on Build GRCh38
rs917728863 Chr.X: 153805489-153805489 on Build GRCh38
rs191589260 Chr.X: 153805877-153805877 on Build GRCh38
rs188719613 Chr.X: 153805485-153805485 on Build GRCh38

View Full Product Details