Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70138412-70138811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYSND1
Entrez Gene ID
219743
Gene Name
trypsin domain containing 1
Gene Aliases
NET41
UniGene
Hs.533655
Gene Chromosome Location
Chr.10: 70137981-70146740 on Build GRCh38
Transcripts
NM_001040273.2 NM_173555.3 NR_073580.1 NR_073581.1 NR_073582.1 NR_073591.1 NR_073592.1 NR_073593.1 NR_073594.1 NR_073595.1

SNP Detail

SNP ID Chromosome Location
rs975720274 Chr.10: 70138484-70138484 on Build GRCh38
rs1041007774 Chr.10: 70138470-70138470 on Build GRCh38
rs184005847 Chr.10: 70138529-70138529 on Build GRCh38
rs941949852 Chr.10: 70138700-70138700 on Build GRCh38
rs984150604 Chr.10: 70138697-70138697 on Build GRCh38
rs530166805 Chr.10: 70138580-70138580 on Build GRCh38
rs966666852 Chr.10: 70138781-70138781 on Build GRCh38
rs886825883 Chr.10: 70138442-70138442 on Build GRCh38
rs962864348 Chr.10: 70138475-70138475 on Build GRCh38
rs976768539 Chr.10: 70138675-70138675 on Build GRCh38
rs760292991 Chr.10: 70138485-70138485 on Build GRCh38
rs999203558 Chr.10: 70138600-70138600 on Build GRCh38
rs562531651 Chr.10: 70138556-70138556 on Build GRCh38
rs955414100 Chr.10: 70138489-70138489 on Build GRCh38
rs571061942 Chr.10: 70138770-70138770 on Build GRCh38
rs949758469 Chr.10: 70138562-70138562 on Build GRCh38
rs965433157 Chr.10: 70138654-70138654 on Build GRCh38
rs929999327 Chr.10: 70138512-70138512 on Build GRCh38
rs887717804 Chr.10: 70138514-70138514 on Build GRCh38
rs565041497 Chr.10: 70138474-70138474 on Build GRCh38
rs771046244 Chr.10: 70138775-70138775 on Build GRCh38
rs902272710 Chr.10: 70138778-70138778 on Build GRCh38
rs1020944005 Chr.10: 70138779-70138779 on Build GRCh38
rs913989647 Chr.10: 70138494-70138494 on Build GRCh38
rs532387966 Chr.10: 70138513-70138513 on Build GRCh38
rs944156651 Chr.10: 70138466-70138466 on Build GRCh38
rs767470840 Chr.10: 70138761-70138761 on Build GRCh38
rs915787332 Chr.10: 70138557-70138557 on Build GRCh38
rs935813546 Chr.10: 70138515-70138515 on Build GRCh38
rs754447096 Chr.10: 70138525-70138525 on Build GRCh38
rs1018423294 Chr.10: 70138648-70138648 on Build GRCh38
rs755291941 Chr.10: 70138548-70138548 on Build GRCh38
rs753016785 Chr.10: 70138643-70138643 on Build GRCh38
rs898689295 Chr.10: 70138752-70138752 on Build GRCh38
rs139978097 Chr.10: 70138768-70138768 on Build GRCh38
rs979476413 Chr.10: 70138509-70138509 on Build GRCh38
rs550607849 Chr.10: 70138517-70138517 on Build GRCh38
rs34023753 Chr.10: 70138760-70138760 on Build GRCh38
rs945501927 Chr.10: 70138505-70138505 on Build GRCh38
rs1038114246 Chr.10: 70138727-70138727 on Build GRCh38
rs111311141 Chr.10: 70138692-70138692 on Build GRCh38
rs955773481 Chr.10: 70138767-70138767 on Build GRCh38
rs1004387853 Chr.10: 70138452-70138452 on Build GRCh38
rs41277980 Chr.10: 70138440-70138440 on Build GRCh38
rs1045387793 Chr.10: 70138619-70138619 on Build GRCh38
rs143857056 Chr.10: 70138693-70138693 on Build GRCh38
rs2394647 Chr.10: 70138599-70138599 on Build GRCh38
rs958329621 Chr.10: 70138624-70138624 on Build GRCh38
rs1054993665 Chr.10: 70138524-70138524 on Build GRCh38
rs895014737 Chr.10: 70138725-70138725 on Build GRCh38
rs985582772 Chr.10: 70138632-70138632 on Build GRCh38
rs987171590 Chr.10: 70138493-70138493 on Build GRCh38
rs909108811 Chr.10: 70138698-70138699 on Build GRCh38
rs1054962650 Chr.10: 70138724-70138724 on Build GRCh38

View Full Product Details