Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41480578-41481074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT35
Entrez Gene ID
3886
Gene Name
keratin 35
Gene Aliases
HA5, Ha-5, KRTHA5, hHa5
UniGene
Hs.73082
Gene Chromosome Location
Chr.17: 41476689-41482743 on Build GRCh38
Transcripts
NM_002280.4 XM_011524794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
329686 c.471+5delG p.? Chr.17: 41480622-41480622 on Build GRCh38
6898895 c.398G>A p.C133Y Chr.17: 41480700-41480700 on Build GRCh38
5749773 c.305A>G p.Q102R Chr.17: 41480793-41480793 on Build GRCh38
3819561 c.162G>T p.V54V Chr.17: 41480936-41480936 on Build GRCh38
1285833 c.326C>A p.A109D Chr.17: 41480772-41480772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755005785 Chr.17: 41480972-41480972 on Build GRCh38
rs200189711 Chr.17: 41480875-41480875 on Build GRCh38
rs758575937 Chr.17: 41480986-41480986 on Build GRCh38
rs758100090 Chr.17: 41480828-41480828 on Build GRCh38
rs200451053 Chr.17: 41480710-41480710 on Build GRCh38
rs371550228 Chr.17: 41480969-41480969 on Build GRCh38
rs377000130 Chr.17: 41480770-41480770 on Build GRCh38
rs375205375 Chr.17: 41480970-41480970 on Build GRCh38
rs201523783 Chr.17: 41480743-41480743 on Build GRCh38
rs749718410 Chr.17: 41480701-41480701 on Build GRCh38
rs1036097733 Chr.17: 41480690-41480690 on Build GRCh38
rs1033509581 Chr.17: 41480696-41480696 on Build GRCh38
rs201582083 Chr.17: 41481013-41481013 on Build GRCh38
rs780311443 Chr.17: 41480759-41480759 on Build GRCh38
rs774738086 Chr.17: 41480755-41480758 on Build GRCh38
rs371261817 Chr.17: 41480843-41480843 on Build GRCh38
rs568564860 Chr.17: 41481026-41481026 on Build GRCh38
rs1021195310 Chr.17: 41480919-41480919 on Build GRCh38
rs775007819 Chr.17: 41481018-41481018 on Build GRCh38
rs770754537 Chr.17: 41481031-41481031 on Build GRCh38
rs771618297 Chr.17: 41481006-41481006 on Build GRCh38
rs377455839 Chr.17: 41480775-41480775 on Build GRCh38
rs772308134 Chr.17: 41480881-41480881 on Build GRCh38
rs755641537 Chr.17: 41480841-41480841 on Build GRCh38
rs743686 Chr.17: 41480992-41480992 on Build GRCh38
rs778970011 Chr.17: 41480860-41480860 on Build GRCh38
rs777423088 Chr.17: 41480876-41480890 on Build GRCh38
rs762865803 Chr.17: 41480943-41480943 on Build GRCh38
rs866972687 Chr.17: 41480849-41480849 on Build GRCh38
rs373516449 Chr.17: 41480737-41480737 on Build GRCh38
rs748438525 Chr.17: 41480779-41480779 on Build GRCh38
rs544871925 Chr.17: 41480645-41480645 on Build GRCh38
rs373926552 Chr.17: 41480993-41480993 on Build GRCh38
rs549831235 Chr.17: 41480822-41480822 on Build GRCh38
rs200703006 Chr.17: 41480987-41480987 on Build GRCh38
rs572743417 Chr.17: 41480711-41480711 on Build GRCh38
rs768995554 Chr.17: 41480702-41480702 on Build GRCh38
rs34548975 Chr.17: 41481041-41481042 on Build GRCh38
rs768224452 Chr.17: 41480715-41480715 on Build GRCh38
rs781360780 Chr.17: 41480672-41480672 on Build GRCh38
rs941730671 Chr.17: 41480884-41480884 on Build GRCh38
rs375272934 Chr.17: 41480825-41480825 on Build GRCh38
rs940086104 Chr.17: 41480847-41480847 on Build GRCh38
rs765996793 Chr.17: 41480804-41480804 on Build GRCh38
rs753958348 Chr.17: 41480971-41480971 on Build GRCh38
rs200651620 Chr.17: 41480893-41480893 on Build GRCh38
rs763809681 Chr.17: 41480961-41480961 on Build GRCh38
rs760600823 Chr.17: 41481050-41481050 on Build GRCh38
rs940157351 Chr.17: 41481030-41481030 on Build GRCh38
rs187375349 Chr.17: 41481021-41481021 on Build GRCh38
rs938444694 Chr.17: 41480964-41480964 on Build GRCh38
rs756180436 Chr.17: 41480758-41480758 on Build GRCh38
rs752873489 Chr.17: 41480979-41480979 on Build GRCh38
rs751485990 Chr.17: 41480840-41480840 on Build GRCh38
rs776209725 Chr.17: 41480817-41480817 on Build GRCh38
rs185963879 Chr.17: 41480709-41480709 on Build GRCh38
rs200159958 Chr.17: 41480653-41480653 on Build GRCh38
rs768826142 Chr.17: 41480771-41480771 on Build GRCh38
rs750343970 Chr.17: 41481042-41481042 on Build GRCh38
rs769529602 Chr.17: 41480931-41480931 on Build GRCh38
rs199694376 Chr.17: 41480968-41480968 on Build GRCh38
rs745985116 Chr.17: 41480752-41480752 on Build GRCh38
rs752589926 Chr.17: 41480826-41480826 on Build GRCh38
rs760315641 Chr.17: 41480798-41480798 on Build GRCh38
rs527275228 Chr.17: 41480878-41480878 on Build GRCh38
rs771528504 Chr.17: 41480897-41480897 on Build GRCh38
rs775323065 Chr.17: 41481002-41481002 on Build GRCh38
rs984400628 Chr.17: 41480871-41480871 on Build GRCh38
rs145937229 Chr.17: 41480674-41480674 on Build GRCh38
rs758198541 Chr.17: 41480663-41480663 on Build GRCh38
rs371467079 Chr.17: 41480704-41480704 on Build GRCh38
rs757674828 Chr.17: 41480644-41480644 on Build GRCh38
rs746897330 Chr.17: 41480688-41480688 on Build GRCh38
rs771735371 Chr.17: 41480867-41480867 on Build GRCh38
rs201336508 Chr.17: 41480731-41480731 on Build GRCh38
rs770048800 Chr.17: 41480682-41480682 on Build GRCh38
rs191074954 Chr.17: 41480732-41480732 on Build GRCh38
rs761563340 Chr.17: 41481028-41481028 on Build GRCh38
rs769176495 Chr.17: 41481011-41481011 on Build GRCh38
rs755276315 Chr.17: 41480848-41480848 on Build GRCh38
rs761288172 Chr.17: 41480609-41480609 on Build GRCh38
rs781757077 Chr.17: 41481009-41481009 on Build GRCh38
rs368387676 Chr.17: 41480651-41480651 on Build GRCh38
rs748135071 Chr.17: 41480654-41480654 on Build GRCh38
rs749861425 Chr.17: 41480627-41480627 on Build GRCh38
rs759097864 Chr.17: 41480907-41480907 on Build GRCh38
rs577676650 Chr.17: 41480652-41480652 on Build GRCh38
rs747740969 Chr.17: 41480783-41480783 on Build GRCh38
rs199599074 Chr.17: 41481019-41481019 on Build GRCh38
rs778222747 Chr.17: 41480882-41480882 on Build GRCh38
rs772672108 Chr.17: 41480794-41480794 on Build GRCh38
rs775003917 Chr.17: 41480935-41480935 on Build GRCh38
rs778401284 Chr.17: 41480751-41480751 on Build GRCh38
rs201945790 Chr.17: 41480619-41480619 on Build GRCh38
rs976798843 Chr.17: 41480793-41480793 on Build GRCh38
rs139838007 Chr.17: 41481010-41481010 on Build GRCh38
rs779251289 Chr.17: 41480977-41480977 on Build GRCh38
rs776307492 Chr.17: 41480899-41480899 on Build GRCh38
rs199661421 Chr.17: 41480913-41480913 on Build GRCh38
rs757128527 Chr.17: 41480842-41480842 on Build GRCh38
rs765232461 Chr.17: 41481051-41481051 on Build GRCh38
rs370278283 Chr.17: 41480778-41480778 on Build GRCh38
rs919044461 Chr.17: 41480955-41480955 on Build GRCh38
rs764160696 Chr.17: 41480838-41480838 on Build GRCh38
rs775657586 Chr.17: 41480687-41480687 on Build GRCh38
rs751983782 Chr.17: 41480636-41480636 on Build GRCh38
rs753530971 Chr.17: 41480716-41480716 on Build GRCh38
rs181774285 Chr.17: 41480952-41480952 on Build GRCh38
rs767508558 Chr.17: 41481040-41481040 on Build GRCh38
rs921444698 Chr.17: 41480815-41480815 on Build GRCh38
rs773584728 Chr.17: 41480796-41480796 on Build GRCh38

View Full Product Details