Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133340827-133341323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED22
Entrez Gene ID
6837
Gene Name
mediator complex subunit 22
Gene Aliases
MED24, SURF5, surf-5
UniGene
Hs.78354
Gene Chromosome Location
Chr.9: 133340901-133348156 on Build GRCh38
Transcripts
NM_133640.4 NM_181491.2

SNP Detail

SNP ID Chromosome Location
rs1013394698 Chr.9: 133341240-133341240 on Build GRCh38
rs1034891365 Chr.9: 133341037-133341037 on Build GRCh38
rs943107484 Chr.9: 133341127-133341127 on Build GRCh38
rs1013034755 Chr.9: 133341052-133341052 on Build GRCh38
rs899127619 Chr.9: 133341272-133341272 on Build GRCh38
rs587642444 Chr.9: 133340940-133340940 on Build GRCh38
rs587734004 Chr.9: 133341158-133341158 on Build GRCh38
rs782651199 Chr.9: 133341155-133341155 on Build GRCh38
rs929131589 Chr.9: 133341063-133341063 on Build GRCh38
rs587701080 Chr.9: 133341102-133341102 on Build GRCh38
rs994850124 Chr.9: 133341278-133341278 on Build GRCh38
rs782059995 Chr.9: 133341088-133341088 on Build GRCh38
rs954455275 Chr.9: 133341134-133341134 on Build GRCh38
rs908634626 Chr.9: 133341232-133341232 on Build GRCh38
rs782298421 Chr.9: 133341124-133341124 on Build GRCh38
rs1013137442 Chr.9: 133341096-133341096 on Build GRCh38
rs112461556 Chr.9: 133341285-133341285 on Build GRCh38
rs782163308 Chr.9: 133341005-133341005 on Build GRCh38
rs782771314 Chr.9: 133341214-133341214 on Build GRCh38
rs200859721 Chr.9: 133340921-133340921 on Build GRCh38
rs782109984 Chr.9: 133341083-133341083 on Build GRCh38
rs1042661629 Chr.9: 133341173-133341173 on Build GRCh38
rs1026771133 Chr.9: 133341280-133341280 on Build GRCh38
rs902429179 Chr.9: 133341174-133341174 on Build GRCh38
rs939284097 Chr.9: 133341180-133341180 on Build GRCh38
rs587619203 Chr.9: 133341073-133341073 on Build GRCh38
rs964728024 Chr.9: 133341171-133341171 on Build GRCh38
rs782093086 Chr.9: 133340878-133340878 on Build GRCh38
rs1042322749 Chr.9: 133340998-133340998 on Build GRCh38
rs911642322 Chr.9: 133340918-133340918 on Build GRCh38
rs781828908 Chr.9: 133341132-133341132 on Build GRCh38
rs587604716 Chr.9: 133341159-133341159 on Build GRCh38
rs193089074 Chr.9: 133340988-133340988 on Build GRCh38
rs782788617 Chr.9: 133341053-133341053 on Build GRCh38
rs141908130 Chr.9: 133341286-133341286 on Build GRCh38
rs902312582 Chr.9: 133341004-133341004 on Build GRCh38
rs1023060956 Chr.9: 133341253-133341253 on Build GRCh38
rs987131096 Chr.9: 133340984-133340984 on Build GRCh38
rs974539768 Chr.9: 133341152-133341152 on Build GRCh38
rs987594422 Chr.9: 133341109-133341109 on Build GRCh38
rs950845931 Chr.9: 133341105-133341105 on Build GRCh38
rs147925691 Chr.9: 133340871-133340871 on Build GRCh38
rs782203995 Chr.9: 133341014-133341014 on Build GRCh38
rs943159694 Chr.9: 133340919-133340919 on Build GRCh38
rs149977018 Chr.9: 133340918-133340919 on Build GRCh38
rs782752438 Chr.9: 133341110-133341110 on Build GRCh38
rs893499118 Chr.9: 133340945-133340945 on Build GRCh38
rs1017229356 Chr.9: 133341170-133341170 on Build GRCh38
rs894727408 Chr.9: 133341095-133341095 on Build GRCh38
rs954549339 Chr.9: 133340982-133340982 on Build GRCh38
rs184679506 Chr.9: 133341226-133341226 on Build GRCh38
rs587739471 Chr.9: 133341195-133341195 on Build GRCh38
rs998644753 Chr.9: 133340937-133340937 on Build GRCh38
rs11655 Chr.9: 133340993-133340993 on Build GRCh38
rs929489669 Chr.9: 133340846-133340846 on Build GRCh38
rs895896630 Chr.9: 133341047-133341047 on Build GRCh38
rs974034721 Chr.9: 133341086-133341086 on Build GRCh38
rs621907 Chr.9: 133341194-133341194 on Build GRCh38

View Full Product Details