Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153934884-153935382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAA10
Entrez Gene ID
8260
Gene Name
N(alpha)-acetyltransferase 10, NatA catalytic subunit
Gene Aliases
ARD1, ARD1A, ARD1P, DXS707, MCOPS1, NATD, OGDNS, TE2
UniGene
Hs.433291
Gene Chromosome Location
Chr.X: 153929827-153935154 on Build GRCh38
Transcripts
NM_001256119.1 NM_001256120.1 NM_003491.3 NM_002910.5 XM_017029698.1

Gene Symbol
RENBP
Entrez Gene ID
5973
Gene Name
renin binding protein
Gene Aliases
RBP, RNBP
UniGene
Hs.158331
Gene Chromosome Location
Chr.X: 153935263-153944780 on Build GRCh38
Transcripts
NM_001256119.1 NM_001256120.1 NM_003491.3 NM_002910.5 XM_017029698.1

SNP Detail

SNP ID Chromosome Location
rs782432905 Chr.X: 153935244-153935244 on Build GRCh38
rs782618876 Chr.X: 153935333-153935333 on Build GRCh38
rs781792536 Chr.X: 153935357-153935357 on Build GRCh38
rs782051988 Chr.X: 153935318-153935319 on Build GRCh38
rs781854646 Chr.X: 153935307-153935307 on Build GRCh38
rs782512087 Chr.X: 153935330-153935330 on Build GRCh38
rs782799079 Chr.X: 153935299-153935299 on Build GRCh38
rs72616465 Chr.X: 153935224-153935224 on Build GRCh38
rs192489081 Chr.X: 153935010-153935010 on Build GRCh38

View Full Product Details