Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50622268-50622814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT3
Entrez Gene ID
84258
Gene Name
synaptotagmin 3
Gene Aliases
SytIII
UniGene
Hs.515554
Gene Chromosome Location
Chr.19: 50621977-50658127 on Build GRCh38
Transcripts
NM_001160328.1 NM_001160329.1 NM_032298.2 XM_011527390.2 XM_011527391.2

SNP Detail

SNP ID Chromosome Location
rs1057207696 Chr.19: 50622567-50622602 on Build GRCh38
rs754772190 Chr.19: 50622785-50622785 on Build GRCh38
rs1032446569 Chr.19: 50622387-50622387 on Build GRCh38
rs374263730 Chr.19: 50622790-50622790 on Build GRCh38
rs540849964 Chr.19: 50622581-50622581 on Build GRCh38
rs1009727962 Chr.19: 50622418-50622418 on Build GRCh38
rs534880240 Chr.19: 50622612-50622612 on Build GRCh38
rs575716780 Chr.19: 50622360-50622360 on Build GRCh38
rs1019820583 Chr.19: 50622423-50622423 on Build GRCh38
rs777145059 Chr.19: 50622714-50622714 on Build GRCh38
rs775904871 Chr.19: 50622686-50622686 on Build GRCh38
rs577289321 Chr.19: 50622592-50622592 on Build GRCh38
rs892600393 Chr.19: 50622406-50622406 on Build GRCh38
rs905169112 Chr.19: 50622553-50622553 on Build GRCh38
rs971868046 Chr.19: 50622461-50622461 on Build GRCh38
rs544714755 Chr.19: 50622607-50622607 on Build GRCh38
rs748990166 Chr.19: 50622678-50622678 on Build GRCh38
rs879205122 Chr.19: 50622347-50622347 on Build GRCh38
rs772566603 Chr.19: 50622676-50622676 on Build GRCh38
rs975697801 Chr.19: 50622450-50622450 on Build GRCh38
rs557359416 Chr.19: 50622355-50622355 on Build GRCh38
rs758751227 Chr.19: 50622780-50622780 on Build GRCh38
rs539158192 Chr.19: 50622320-50622320 on Build GRCh38
rs567530929 Chr.19: 50622541-50622541 on Build GRCh38
rs984454871 Chr.19: 50622656-50622656 on Build GRCh38
rs878899591 Chr.19: 50622316-50622316 on Build GRCh38
rs1028585488 Chr.19: 50622645-50622645 on Build GRCh38
rs760966257 Chr.19: 50622695-50622695 on Build GRCh38
rs139672852 Chr.19: 50622696-50622696 on Build GRCh38
rs917782285 Chr.19: 50622467-50622467 on Build GRCh38
rs769309198 Chr.19: 50622667-50622667 on Build GRCh38
rs1016382916 Chr.19: 50622664-50622664 on Build GRCh38
rs201686278 Chr.19: 50622682-50622682 on Build GRCh38
rs553159271 Chr.19: 50622582-50622582 on Build GRCh38
rs118078846 Chr.19: 50622639-50622639 on Build GRCh38
rs372064620 Chr.19: 50622728-50622728 on Build GRCh38
rs370127270 Chr.19: 50622640-50622640 on Build GRCh38
rs908945841 Chr.19: 50622459-50622459 on Build GRCh38
rs984915976 Chr.19: 50622456-50622456 on Build GRCh38
rs573919237 Chr.19: 50622566-50622566 on Build GRCh38
rs1029035744 Chr.19: 50622453-50622453 on Build GRCh38
rs751403482 Chr.19: 50622781-50622781 on Build GRCh38
rs965617720 Chr.19: 50622435-50622435 on Build GRCh38
rs747635509 Chr.19: 50622660-50622660 on Build GRCh38
rs750447303 Chr.19: 50622768-50622768 on Build GRCh38
rs781116196 Chr.19: 50622648-50622648 on Build GRCh38
rs940467474 Chr.19: 50622460-50622460 on Build GRCh38
rs1045036180 Chr.19: 50622336-50622336 on Build GRCh38
rs777659790 Chr.19: 50622777-50622786 on Build GRCh38
rs186522177 Chr.19: 50622289-50622289 on Build GRCh38
rs537303716 Chr.19: 50622538-50622538 on Build GRCh38

View Full Product Details