Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76482839-76483424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX2
Entrez Gene ID
113878
Gene Name
deltex E3 ubiquitin ligase 2
Gene Aliases
RNF58
UniGene
Hs.187058
Gene Chromosome Location
Chr.7: 76461655-76505995 on Build GRCh38
Transcripts
NM_001102594.1 NM_001102595.1 NM_001102596.1 NM_020892.2 XM_005250131.1 XM_005250132.1 XM_011515772.1 XM_011515773.2 XM_011515774.1 XM_011515775.1 XM_017011723.1 XM_017011724.1 XM_017011725.1 XM_017011726.1 XM_017011727.1 XM_017011728.1 XM_017011729.1 XM_017011730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1452121 c.693delA p.H232fs*30 Chr.7: 76482932-76482932 on Build GRCh38
342024 c.701C>T p.T234I Chr.7: 76482940-76482940 on Build GRCh38
145350 c.731T>C p.F244S Chr.7: 76482970-76482970 on Build GRCh38
6650070 c.759C>T p.S253S Chr.7: 76482998-76482998 on Build GRCh38
5906804 c.664C>T p.P222S Chr.7: 76482903-76482903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769421747 Chr.7: 76482943-76482943 on Build GRCh38
rs756977113 Chr.7: 76483080-76483080 on Build GRCh38
rs757469796 Chr.7: 76482930-76482930 on Build GRCh38
rs774852183 Chr.7: 76483191-76483191 on Build GRCh38
rs199598165 Chr.7: 76482915-76482915 on Build GRCh38
rs777517183 Chr.7: 76483005-76483005 on Build GRCh38
rs764704468 Chr.7: 76482864-76482864 on Build GRCh38
rs2258686 Chr.7: 76482932-76482932 on Build GRCh38
rs552061839 Chr.7: 76483258-76483258 on Build GRCh38
rs143934697 Chr.7: 76482926-76482926 on Build GRCh38
rs577518480 Chr.7: 76483129-76483129 on Build GRCh38
rs773838700 Chr.7: 76483121-76483121 on Build GRCh38
rs750414249 Chr.7: 76483159-76483159 on Build GRCh38
rs778090809 Chr.7: 76482886-76482886 on Build GRCh38
rs776357746 Chr.7: 76483105-76483105 on Build GRCh38
rs751158935 Chr.7: 76482876-76482876 on Build GRCh38
rs758494603 Chr.7: 76482925-76482926 on Build GRCh38
rs779399752 Chr.7: 76483084-76483084 on Build GRCh38
rs374522089 Chr.7: 76483165-76483165 on Build GRCh38
rs920873376 Chr.7: 76483275-76483275 on Build GRCh38
rs930963862 Chr.7: 76483299-76483299 on Build GRCh38
rs559738855 Chr.7: 76482911-76482911 on Build GRCh38
rs759623433 Chr.7: 76483192-76483192 on Build GRCh38
rs1035488033 Chr.7: 76483004-76483004 on Build GRCh38
rs559156215 Chr.7: 76483044-76483044 on Build GRCh38
rs138141868 Chr.7: 76483012-76483012 on Build GRCh38
rs373052232 Chr.7: 76483152-76483152 on Build GRCh38
rs768456000 Chr.7: 76483096-76483096 on Build GRCh38
rs968747176 Chr.7: 76482898-76482898 on Build GRCh38
rs978815021 Chr.7: 76483103-76483103 on Build GRCh38
rs766027737 Chr.7: 76482870-76482870 on Build GRCh38
rs759568472 Chr.7: 76483136-76483136 on Build GRCh38
rs544024397 Chr.7: 76483043-76483043 on Build GRCh38
rs755130056 Chr.7: 76483049-76483049 on Build GRCh38
rs778198302 Chr.7: 76482932-76482932 on Build GRCh38
rs142155908 Chr.7: 76483081-76483081 on Build GRCh38
rs750563156 Chr.7: 76482994-76482994 on Build GRCh38
rs552000434 Chr.7: 76482955-76482955 on Build GRCh38
rs886818009 Chr.7: 76483301-76483301 on Build GRCh38
rs747982040 Chr.7: 76482942-76482942 on Build GRCh38
rs576710616 Chr.7: 76483026-76483026 on Build GRCh38
rs142474507 Chr.7: 76482999-76482999 on Build GRCh38
rs368991320 Chr.7: 76482980-76482980 on Build GRCh38
rs774312459 Chr.7: 76483131-76483131 on Build GRCh38
rs781339036 Chr.7: 76483056-76483056 on Build GRCh38
rs769359297 Chr.7: 76483232-76483232 on Build GRCh38
rs555222086 Chr.7: 76483024-76483024 on Build GRCh38
rs188650862 Chr.7: 76482914-76482914 on Build GRCh38
rs765845959 Chr.7: 76483034-76483034 on Build GRCh38
rs773824914 Chr.7: 76482954-76482954 on Build GRCh38
rs530707991 Chr.7: 76482950-76482950 on Build GRCh38
rs751720408 Chr.7: 76483000-76483000 on Build GRCh38
rs550117954 Chr.7: 76482919-76482919 on Build GRCh38
rs144914725 Chr.7: 76482902-76482902 on Build GRCh38
rs767569894 Chr.7: 76483195-76483195 on Build GRCh38
rs2690583 Chr.7: 76483307-76483307 on Build GRCh38
rs761113916 Chr.7: 76482975-76482975 on Build GRCh38
rs773194901 Chr.7: 76482944-76482944 on Build GRCh38
rs770326343 Chr.7: 76483169-76483169 on Build GRCh38
rs762466989 Chr.7: 76482918-76482918 on Build GRCh38
rs148430404 Chr.7: 76482882-76482882 on Build GRCh38
rs939795476 Chr.7: 76483333-76483333 on Build GRCh38
rs770273048 Chr.7: 76483116-76483116 on Build GRCh38
rs886894451 Chr.7: 76483002-76483002 on Build GRCh38
rs758670196 Chr.7: 76482933-76482933 on Build GRCh38
rs749972814 Chr.7: 76482866-76482866 on Build GRCh38
rs368500347 Chr.7: 76482867-76482867 on Build GRCh38
rs546274019 Chr.7: 76483353-76483353 on Build GRCh38
rs140118064 Chr.7: 76482924-76482924 on Build GRCh38
rs979276089 Chr.7: 76482917-76482917 on Build GRCh38
rs149983793 Chr.7: 76482973-76482973 on Build GRCh38
rs374758181 Chr.7: 76483186-76483186 on Build GRCh38
rs369802603 Chr.7: 76482923-76482923 on Build GRCh38
rs376653406 Chr.7: 76482941-76482941 on Build GRCh38
rs767227824 Chr.7: 76482964-76482964 on Build GRCh38
rs535139642 Chr.7: 76483013-76483013 on Build GRCh38
rs1004448935 Chr.7: 76483003-76483003 on Build GRCh38
rs956190351 Chr.7: 76483119-76483119 on Build GRCh38
rs2270253 Chr.7: 76482916-76482916 on Build GRCh38
rs754463201 Chr.7: 76482977-76482977 on Build GRCh38
rs747801084 Chr.7: 76483167-76483167 on Build GRCh38
rs972265913 Chr.7: 76482890-76482890 on Build GRCh38
rs745386317 Chr.7: 76483015-76483015 on Build GRCh38
rs2430309 Chr.7: 76483201-76483201 on Build GRCh38
rs749268963 Chr.7: 76483006-76483006 on Build GRCh38
rs372688420 Chr.7: 76482931-76482931 on Build GRCh38
rs777158978 Chr.7: 76483031-76483031 on Build GRCh38
rs968675104 Chr.7: 76483054-76483054 on Build GRCh38
rs565377740 Chr.7: 76482877-76482877 on Build GRCh38
rs753661686 Chr.7: 76483078-76483078 on Build GRCh38
rs767727619 Chr.7: 76483137-76483137 on Build GRCh38
rs79750972 Chr.7: 76483346-76483346 on Build GRCh38
rs1012728829 Chr.7: 76483357-76483357 on Build GRCh38
rs756390618 Chr.7: 76482928-76482928 on Build GRCh38
rs145151450 Chr.7: 76482982-76482982 on Build GRCh38
rs775753992 Chr.7: 76483197-76483197 on Build GRCh38
rs151329883 Chr.7: 76482929-76482929 on Build GRCh38
rs372226447 Chr.7: 76482884-76482884 on Build GRCh38
rs369794491 Chr.7: 76483110-76483110 on Build GRCh38
rs1799171 Chr.7: 76483385-76483385 on Build GRCh38
rs1638073 Chr.7: 76483170-76483170 on Build GRCh38
rs559605865 Chr.7: 76483153-76483153 on Build GRCh38
rs199811851 Chr.7: 76483083-76483083 on Build GRCh38
rs746479588 Chr.7: 76483089-76483089 on Build GRCh38
rs974972772 Chr.7: 76483266-76483266 on Build GRCh38
rs376471639 Chr.7: 76483161-76483161 on Build GRCh38
rs146537218 Chr.7: 76483001-76483001 on Build GRCh38
rs749612129 Chr.7: 76483184-76483184 on Build GRCh38
rs10247123 Chr.7: 76483163-76483163 on Build GRCh38
rs757338585 Chr.7: 76482891-76482891 on Build GRCh38
rs776464727 Chr.7: 76482912-76482912 on Build GRCh38
rs764351511 Chr.7: 76482976-76482976 on Build GRCh38
rs375142588 Chr.7: 76483300-76483300 on Build GRCh38
rs373466186 Chr.7: 76482998-76482998 on Build GRCh38
rs752826429 Chr.7: 76482863-76482863 on Build GRCh38
rs2530609 Chr.7: 76483042-76483042 on Build GRCh38
rs752856351 Chr.7: 76483142-76483142 on Build GRCh38
rs753180358 Chr.7: 76482883-76482883 on Build GRCh38
rs199743411 Chr.7: 76483166-76483166 on Build GRCh38
rs761778612 Chr.7: 76482863-76482863 on Build GRCh38
rs749409113 Chr.7: 76482887-76482887 on Build GRCh38
rs779291726 Chr.7: 76482895-76482895 on Build GRCh38
rs376375118 Chr.7: 76483019-76483019 on Build GRCh38
rs765864484 Chr.7: 76482921-76482921 on Build GRCh38
rs113825285 Chr.7: 76483345-76483345 on Build GRCh38
rs569884280 Chr.7: 76482949-76482949 on Build GRCh38

View Full Product Details