Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42107854-42108310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX58
Entrez Gene ID
79132
Gene Name
DEXH-box helicase 58
Gene Aliases
D11LGP2, D11lgp2e, LGP2, RLR-3
UniGene
Hs.55918
Gene Chromosome Location
Chr.17: 42101404-42112733 on Build GRCh38
Transcripts
NM_024119.2 XM_017025059.1 XM_017025060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3188341 c.805G>A p.A269T Chr.17: 42107982-42107982 on Build GRCh38
3517612 c.716A>T p.Q239L Chr.17: 42108071-42108071 on Build GRCh38
706328 c.753G>T p.R251R Chr.17: 42108034-42108034 on Build GRCh38
160421 c.742G>C p.E248Q Chr.17: 42108045-42108045 on Build GRCh38
5774652 c.751C>T p.R251W Chr.17: 42108036-42108036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782280357 Chr.17: 42108047-42108047 on Build GRCh38
rs542126610 Chr.17: 42108168-42108168 on Build GRCh38
rs371459098 Chr.17: 42108021-42108021 on Build GRCh38
rs781800937 Chr.17: 42107972-42107972 on Build GRCh38
rs782349280 Chr.17: 42108075-42108075 on Build GRCh38
rs782105599 Chr.17: 42108114-42108114 on Build GRCh38
rs541581567 Chr.17: 42107959-42107959 on Build GRCh38
rs563784554 Chr.17: 42108264-42108264 on Build GRCh38
rs782514778 Chr.17: 42108014-42108014 on Build GRCh38
rs577415123 Chr.17: 42107921-42107921 on Build GRCh38
rs782424594 Chr.17: 42108049-42108049 on Build GRCh38
rs940462130 Chr.17: 42107941-42107941 on Build GRCh38
rs782582597 Chr.17: 42108041-42108041 on Build GRCh38
rs781878338 Chr.17: 42107963-42107963 on Build GRCh38
rs2074154 Chr.17: 42107962-42107962 on Build GRCh38
rs1047714324 Chr.17: 42107908-42107908 on Build GRCh38
rs782790320 Chr.17: 42108108-42108108 on Build GRCh38
rs782227711 Chr.17: 42108022-42108022 on Build GRCh38
rs782690330 Chr.17: 42107994-42107994 on Build GRCh38
rs536762494 Chr.17: 42107919-42107919 on Build GRCh38
rs781813543 Chr.17: 42108145-42108145 on Build GRCh38
rs552110701 Chr.17: 42108224-42108224 on Build GRCh38
rs537454136 Chr.17: 42107885-42107885 on Build GRCh38
rs781838844 Chr.17: 42107976-42107976 on Build GRCh38
rs781970197 Chr.17: 42108078-42108078 on Build GRCh38
rs781992768 Chr.17: 42108054-42108054 on Build GRCh38
rs907615773 Chr.17: 42107918-42107918 on Build GRCh38
rs1019713708 Chr.17: 42108251-42108251 on Build GRCh38
rs374027183 Chr.17: 42108064-42108064 on Build GRCh38
rs781845007 Chr.17: 42108012-42108012 on Build GRCh38
rs782317670 Chr.17: 42108036-42108036 on Build GRCh38
rs937008169 Chr.17: 42108119-42108119 on Build GRCh38
rs530383936 Chr.17: 42108182-42108182 on Build GRCh38
rs374550238 Chr.17: 42107968-42107968 on Build GRCh38
rs782134071 Chr.17: 42108060-42108060 on Build GRCh38
rs782527847 Chr.17: 42107974-42107974 on Build GRCh38
rs35778210 Chr.17: 42107889-42107890 on Build GRCh38
rs113827173 Chr.17: 42108184-42108184 on Build GRCh38
rs114326479 Chr.17: 42108125-42108125 on Build GRCh38
rs528159768 Chr.17: 42108286-42108286 on Build GRCh38
rs764528130 Chr.17: 42107904-42107904 on Build GRCh38
rs782046283 Chr.17: 42107929-42107929 on Build GRCh38
rs782565446 Chr.17: 42108140-42108140 on Build GRCh38
rs782088102 Chr.17: 42107964-42107964 on Build GRCh38
rs374872638 Chr.17: 42108169-42108169 on Build GRCh38
rs782539341 Chr.17: 42108149-42108149 on Build GRCh38
rs782063165 Chr.17: 42108097-42108097 on Build GRCh38
rs559049672 Chr.17: 42107920-42107920 on Build GRCh38
rs1020559131 Chr.17: 42108176-42108176 on Build GRCh38
rs782486185 Chr.17: 42108127-42108127 on Build GRCh38
rs782332630 Chr.17: 42107953-42107953 on Build GRCh38

View Full Product Details