Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11013545-11014120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TARDBP
Entrez Gene ID
23435
Gene Name
TAR DNA binding protein
Gene Aliases
ALS10, TDP-43
UniGene
Hs.300624
Gene Chromosome Location
Chr.1: 11012622-11030528 on Build GRCh38
Transcripts
NM_007375.3 XM_017000863.1 XM_017000864.1 XM_017000865.1 XM_017000866.1 XM_017000867.1 XM_017000868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7121191 c.204G>T p.W68C Chr.1: 11013931-11013931 on Build GRCh38
458739 c.59C>T p.S20L Chr.1: 11013786-11013786 on Build GRCh38
3471037 c.193G>T p.D65Y Chr.1: 11013920-11013920 on Build GRCh38
5496964 c.82C>T p.L28F Chr.1: 11013809-11013809 on Build GRCh38
3975862 c.75G>C p.T25T Chr.1: 11013802-11013802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971750485 Chr.1: 11014023-11014023 on Build GRCh38
rs758092840 Chr.1: 11013681-11013681 on Build GRCh38
rs369718876 Chr.1: 11013811-11013811 on Build GRCh38
rs780873066 Chr.1: 11013893-11013893 on Build GRCh38
rs561330926 Chr.1: 11013677-11013677 on Build GRCh38
rs140008847 Chr.1: 11013757-11013757 on Build GRCh38
rs766620483 Chr.1: 11013799-11013799 on Build GRCh38
rs780770428 Chr.1: 11013703-11013703 on Build GRCh38
rs769520878 Chr.1: 11014011-11014011 on Build GRCh38
rs569266043 Chr.1: 11014040-11014040 on Build GRCh38
rs903048201 Chr.1: 11013662-11013662 on Build GRCh38
rs35714982 Chr.1: 11013920-11013921 on Build GRCh38
rs758344713 Chr.1: 11013847-11013847 on Build GRCh38
rs866353277 Chr.1: 11013905-11013905 on Build GRCh38
rs756772204 Chr.1: 11013892-11013892 on Build GRCh38
rs755357622 Chr.1: 11013928-11013928 on Build GRCh38
rs547926643 Chr.1: 11014022-11014022 on Build GRCh38
rs930946087 Chr.1: 11014090-11014090 on Build GRCh38
rs991682088 Chr.1: 11014058-11014058 on Build GRCh38
rs754798189 Chr.1: 11013679-11013679 on Build GRCh38
rs555955326 Chr.1: 11013597-11013597 on Build GRCh38
rs920886185 Chr.1: 11014032-11014032 on Build GRCh38
rs770967242 Chr.1: 11013980-11013980 on Build GRCh38
rs886045034 Chr.1: 11013706-11013707 on Build GRCh38
rs201693535 Chr.1: 11013814-11013814 on Build GRCh38
rs528142716 Chr.1: 11013725-11013725 on Build GRCh38
rs920782656 Chr.1: 11013618-11013618 on Build GRCh38
rs748698993 Chr.1: 11013769-11013769 on Build GRCh38
rs200517396 Chr.1: 11013723-11013723 on Build GRCh38
rs373450963 Chr.1: 11013981-11013981 on Build GRCh38
rs1055972910 Chr.1: 11013641-11013641 on Build GRCh38
rs773358415 Chr.1: 11013793-11013793 on Build GRCh38
rs111671110 Chr.1: 11013817-11013817 on Build GRCh38
rs759774919 Chr.1: 11014010-11014010 on Build GRCh38
rs55825417 Chr.1: 11014051-11014051 on Build GRCh38
rs751073315 Chr.1: 11013889-11013889 on Build GRCh38
rs61730366 Chr.1: 11013925-11013925 on Build GRCh38
rs532319219 Chr.1: 11013784-11013784 on Build GRCh38
rs893057481 Chr.1: 11013710-11013710 on Build GRCh38
rs535276506 Chr.1: 11013841-11013841 on Build GRCh38
rs950097344 Chr.1: 11013652-11013652 on Build GRCh38
rs953145872 Chr.1: 11013876-11013876 on Build GRCh38
rs760679678 Chr.1: 11013682-11013683 on Build GRCh38
rs1055809664 Chr.1: 11013624-11013624 on Build GRCh38
rs572682145 Chr.1: 11013660-11013660 on Build GRCh38
rs765147873 Chr.1: 11013689-11013689 on Build GRCh38
rs142913423 Chr.1: 11013954-11013954 on Build GRCh38
rs187757140 Chr.1: 11013644-11013644 on Build GRCh38
rs72870030 Chr.1: 11013751-11013751 on Build GRCh38
rs1045773650 Chr.1: 11013661-11013661 on Build GRCh38
rs940971679 Chr.1: 11013583-11013583 on Build GRCh38
rs757365023 Chr.1: 11013642-11013642 on Build GRCh38
rs376560308 Chr.1: 11013974-11013974 on Build GRCh38
rs752580444 Chr.1: 11013822-11013822 on Build GRCh38
rs377639252 Chr.1: 11013700-11013700 on Build GRCh38
rs930843143 Chr.1: 11013626-11013626 on Build GRCh38
rs779446284 Chr.1: 11013934-11013934 on Build GRCh38
rs763441783 Chr.1: 11013868-11013868 on Build GRCh38
rs80356716 Chr.1: 11013922-11013922 on Build GRCh38
rs776777703 Chr.1: 11013808-11013808 on Build GRCh38
rs575825467 Chr.1: 11013704-11013705 on Build GRCh38
rs749647867 Chr.1: 11013705-11013705 on Build GRCh38
rs574282680 Chr.1: 11013615-11013615 on Build GRCh38
rs757862204 Chr.1: 11013692-11013692 on Build GRCh38
rs375490347 Chr.1: 11013697-11013697 on Build GRCh38
rs748612906 Chr.1: 11013938-11013938 on Build GRCh38
rs542695606 Chr.1: 11013667-11013667 on Build GRCh38
rs565091566 Chr.1: 11013763-11013763 on Build GRCh38
rs531845599 Chr.1: 11013719-11013719 on Build GRCh38
rs375089732 Chr.1: 11013794-11013794 on Build GRCh38
rs752670179 Chr.1: 11013691-11013691 on Build GRCh38
rs911530889 Chr.1: 11013580-11013580 on Build GRCh38
rs916051068 Chr.1: 11014078-11014078 on Build GRCh38
rs756934624 Chr.1: 11013701-11013701 on Build GRCh38

View Full Product Details