Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30472513-30473042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGAL
Entrez Gene ID
3683
Gene Name
integrin subunit alpha L
Gene Aliases
CD11A, LFA-1, LFA1A
UniGene
Hs.174103
Gene Chromosome Location
Chr.16: 30472662-30523185 on Build GRCh38
Transcripts
NM_001114380.1 NM_002209.2 XM_005255313.1 XM_006721044.1 XM_011545849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3817790 c.38T>C p.L13P Chr.16: 30472875-30472875 on Build GRCh38
300778 c.43T>C p.S15P Chr.16: 30472880-30472880 on Build GRCh38
7292783 c.35C>T p.A12V Chr.16: 30472872-30472872 on Build GRCh38
1749552 c.57C>T p.F19F Chr.16: 30472894-30472894 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201096086 Chr.16: 30472872-30472872 on Build GRCh38
rs540588332 Chr.16: 30472600-30472600 on Build GRCh38
rs77318464 Chr.16: 30472724-30472724 on Build GRCh38
rs766557890 Chr.16: 30472918-30472918 on Build GRCh38
rs976968976 Chr.16: 30472782-30472782 on Build GRCh38
rs765373900 Chr.16: 30472812-30472812 on Build GRCh38
rs770801392 Chr.16: 30472545-30472545 on Build GRCh38
rs1052952736 Chr.16: 30472671-30472671 on Build GRCh38
rs953992926 Chr.16: 30472802-30472802 on Build GRCh38
rs528194162 Chr.16: 30473019-30473019 on Build GRCh38
rs759216454 Chr.16: 30472641-30472641 on Build GRCh38
rs1056722510 Chr.16: 30472566-30472566 on Build GRCh38
rs748788135 Chr.16: 30472859-30472859 on Build GRCh38
rs747422101 Chr.16: 30472791-30472791 on Build GRCh38
rs748713465 Chr.16: 30472884-30472884 on Build GRCh38
rs765169793 Chr.16: 30472657-30472657 on Build GRCh38
rs763177110 Chr.16: 30472905-30472905 on Build GRCh38
rs376435561 Chr.16: 30472799-30472799 on Build GRCh38
rs765225584 Chr.16: 30472900-30472901 on Build GRCh38
rs369314831 Chr.16: 30472720-30472720 on Build GRCh38
rs942414234 Chr.16: 30472944-30472944 on Build GRCh38
rs373763604 Chr.16: 30472798-30472798 on Build GRCh38
rs370903267 Chr.16: 30472825-30472825 on Build GRCh38
rs773740407 Chr.16: 30472892-30472892 on Build GRCh38
rs752690019 Chr.16: 30472837-30472837 on Build GRCh38
rs1010205947 Chr.16: 30472753-30472753 on Build GRCh38
rs1054500285 Chr.16: 30472717-30472717 on Build GRCh38
rs543472247 Chr.16: 30472640-30472640 on Build GRCh38
rs985326097 Chr.16: 30472805-30472805 on Build GRCh38
rs929781223 Chr.16: 30472621-30472621 on Build GRCh38
rs1027684417 Chr.16: 30472649-30472649 on Build GRCh38
rs563065988 Chr.16: 30472779-30472779 on Build GRCh38
rs1005258092 Chr.16: 30473014-30473014 on Build GRCh38
rs974931841 Chr.16: 30472957-30472957 on Build GRCh38
rs773959642 Chr.16: 30472787-30472787 on Build GRCh38
rs202078829 Chr.16: 30472818-30472818 on Build GRCh38
rs1039192940 Chr.16: 30472659-30472659 on Build GRCh38
rs773118038 Chr.16: 30472817-30472817 on Build GRCh38
rs368954009 Chr.16: 30472930-30472930 on Build GRCh38
rs745515604 Chr.16: 30472869-30472869 on Build GRCh38
rs777371433 Chr.16: 30472853-30472853 on Build GRCh38
rs776804144 Chr.16: 30472873-30472873 on Build GRCh38
rs895376103 Chr.16: 30472603-30472603 on Build GRCh38
rs952005155 Chr.16: 30472708-30472708 on Build GRCh38
rs375748761 Chr.16: 30472824-30472824 on Build GRCh38
rs759770387 Chr.16: 30472925-30472925 on Build GRCh38
rs762759841 Chr.16: 30472921-30472921 on Build GRCh38
rs565246741 Chr.16: 30472988-30472988 on Build GRCh38
rs936233262 Chr.16: 30472714-30472714 on Build GRCh38
rs900784270 Chr.16: 30472666-30472666 on Build GRCh38
rs918227019 Chr.16: 30472537-30472537 on Build GRCh38
rs951127614 Chr.16: 30472897-30472897 on Build GRCh38
rs770161979 Chr.16: 30472888-30472888 on Build GRCh38
rs1056615688 Chr.16: 30472877-30472877 on Build GRCh38
rs149285825 Chr.16: 30472860-30472860 on Build GRCh38
rs374629566 Chr.16: 30472850-30472850 on Build GRCh38
rs778595234 Chr.16: 30472862-30472862 on Build GRCh38
rs1049338554 Chr.16: 30472929-30472930 on Build GRCh38

View Full Product Details