Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27911168-27911692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2B2
Entrez Gene ID
81697
Gene Name
olfactory receptor family 2 subfamily B member 2
Gene Aliases
OR2B2Q, OR2B9, OR6-1, dJ193B12.4, hs6M1-10
UniGene
Hs.307111
Gene Chromosome Location
Chr.6: 27911185-27912396 on Build GRCh38
Transcripts
NM_033057.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
461829 c.879G>C p.R293S Chr.6: 27911441-27911441 on Build GRCh38
6485045 c.956T>G p.M319R Chr.6: 27911364-27911364 on Build GRCh38
3623657 c.889G>A p.V297I Chr.6: 27911431-27911431 on Build GRCh38
6734067 c.925C>T p.L309F Chr.6: 27911395-27911395 on Build GRCh38
451093 c.631G>A p.V211M Chr.6: 27911689-27911689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760915109 Chr.6: 27911330-27911330 on Build GRCh38
rs763459633 Chr.6: 27911363-27911363 on Build GRCh38
rs375078072 Chr.6: 27911471-27911471 on Build GRCh38
rs753463297 Chr.6: 27911387-27911387 on Build GRCh38
rs78401135 Chr.6: 27911511-27911511 on Build GRCh38
rs1026656845 Chr.6: 27911224-27911224 on Build GRCh38
rs144751509 Chr.6: 27911453-27911453 on Build GRCh38
rs760829378 Chr.6: 27911213-27911213 on Build GRCh38
rs759133730 Chr.6: 27911610-27911610 on Build GRCh38
rs754588569 Chr.6: 27911488-27911488 on Build GRCh38
rs753131462 Chr.6: 27911355-27911355 on Build GRCh38
rs774982377 Chr.6: 27911553-27911553 on Build GRCh38
rs775458265 Chr.6: 27911521-27911521 on Build GRCh38
rs755856799 Chr.6: 27911379-27911379 on Build GRCh38
rs776830629 Chr.6: 27911423-27911423 on Build GRCh38
rs779743198 Chr.6: 27911294-27911294 on Build GRCh38
rs771776652 Chr.6: 27911326-27911326 on Build GRCh38
rs143663108 Chr.6: 27911438-27911438 on Build GRCh38
rs144946803 Chr.6: 27911548-27911548 on Build GRCh38
rs868552959 Chr.6: 27911296-27911296 on Build GRCh38
rs762349584 Chr.6: 27911561-27911561 on Build GRCh38
rs771937970 Chr.6: 27911188-27911188 on Build GRCh38
rs749447639 Chr.6: 27911430-27911430 on Build GRCh38
rs73392698 Chr.6: 27911315-27911315 on Build GRCh38
rs755140498 Chr.6: 27911657-27911657 on Build GRCh38
rs1014498479 Chr.6: 27911247-27911247 on Build GRCh38
rs375458136 Chr.6: 27911287-27911287 on Build GRCh38
rs758688905 Chr.6: 27911266-27911266 on Build GRCh38
rs932087346 Chr.6: 27911286-27911286 on Build GRCh38
rs200384354 Chr.6: 27911314-27911314 on Build GRCh38
rs201596071 Chr.6: 27911509-27911509 on Build GRCh38
rs868186131 Chr.6: 27911500-27911500 on Build GRCh38
rs780960606 Chr.6: 27911274-27911274 on Build GRCh38
rs560085003 Chr.6: 27911279-27911279 on Build GRCh38
rs755495558 Chr.6: 27911236-27911236 on Build GRCh38
rs766189013 Chr.6: 27911480-27911480 on Build GRCh38
rs764684180 Chr.6: 27911490-27911490 on Build GRCh38
rs775993954 Chr.6: 27911278-27911278 on Build GRCh38
rs147925033 Chr.6: 27911277-27911277 on Build GRCh38
rs776738415 Chr.6: 27911341-27911341 on Build GRCh38
rs771089412 Chr.6: 27911335-27911335 on Build GRCh38
rs758315013 Chr.6: 27911495-27911495 on Build GRCh38
rs762515906 Chr.6: 27911335-27911335 on Build GRCh38
rs752022664 Chr.6: 27911472-27911472 on Build GRCh38
rs779833061 Chr.6: 27911381-27911381 on Build GRCh38
rs748032869 Chr.6: 27911323-27911323 on Build GRCh38
rs764375625 Chr.6: 27911370-27911370 on Build GRCh38
rs967027358 Chr.6: 27911416-27911416 on Build GRCh38
rs151114349 Chr.6: 27911512-27911512 on Build GRCh38
rs371031773 Chr.6: 27911616-27911616 on Build GRCh38
rs770021461 Chr.6: 27911515-27911515 on Build GRCh38
rs764079255 Chr.6: 27911652-27911652 on Build GRCh38
rs958116141 Chr.6: 27911259-27911259 on Build GRCh38
rs142590029 Chr.6: 27911522-27911522 on Build GRCh38
rs368557270 Chr.6: 27911342-27911342 on Build GRCh38
rs865865291 Chr.6: 27911542-27911542 on Build GRCh38
rs147920845 Chr.6: 27911617-27911617 on Build GRCh38
rs199535361 Chr.6: 27911411-27911411 on Build GRCh38
rs753551195 Chr.6: 27911481-27911481 on Build GRCh38
rs145985903 Chr.6: 27911421-27911421 on Build GRCh38
rs773184342 Chr.6: 27911459-27911459 on Build GRCh38
rs201137841 Chr.6: 27911401-27911401 on Build GRCh38
rs760182947 Chr.6: 27911230-27911230 on Build GRCh38
rs34788973 Chr.6: 27911422-27911422 on Build GRCh38
rs772920671 Chr.6: 27911328-27911328 on Build GRCh38
rs767994392 Chr.6: 27911565-27911565 on Build GRCh38
rs765638294 Chr.6: 27911251-27911251 on Build GRCh38
rs773292384 Chr.6: 27911196-27911196 on Build GRCh38
rs202202321 Chr.6: 27911412-27911412 on Build GRCh38
rs184783783 Chr.6: 27911582-27911582 on Build GRCh38
rs748760215 Chr.6: 27911302-27911302 on Build GRCh38
rs746874756 Chr.6: 27911505-27911505 on Build GRCh38
rs943075828 Chr.6: 27911447-27911447 on Build GRCh38
rs371526433 Chr.6: 27911252-27911252 on Build GRCh38
rs201669058 Chr.6: 27911653-27911653 on Build GRCh38
rs1002218474 Chr.6: 27911571-27911571 on Build GRCh38
rs539471263 Chr.6: 27911623-27911623 on Build GRCh38
rs1045525609 Chr.6: 27911525-27911525 on Build GRCh38
rs148502450 Chr.6: 27911503-27911503 on Build GRCh38
rs769328637 Chr.6: 27911545-27911545 on Build GRCh38
rs958294160 Chr.6: 27911350-27911350 on Build GRCh38
rs765840458 Chr.6: 27911354-27911354 on Build GRCh38
rs34957169 Chr.6: 27911619-27911619 on Build GRCh38
rs372273786 Chr.6: 27911404-27911404 on Build GRCh38
rs752201813 Chr.6: 27911494-27911494 on Build GRCh38

View Full Product Details