Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 82907686-82908253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THAP9
Entrez Gene ID
79725
Gene Name
THAP domain containing 9
Gene Aliases
hTh9
UniGene
Hs.582050
Gene Chromosome Location
Chr.4: 82900650-82920283 on Build GRCh38
Transcripts
NM_001317776.1 NM_024672.5 XM_005263220.2 XM_011532246.2 XM_017008603.1 XM_017008604.1 XM_017008605.1 XM_017008606.1 XM_017008607.1 XM_017008608.1 XM_017008609.1 XM_017008610.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3826333 c.581-1G>A p.? Chr.4: 82907784-82907784 on Build GRCh38
3702648 c.601A>G p.N201D Chr.4: 82907805-82907805 on Build GRCh38
6296259 c.731+1G>T p.? Chr.4: 82907936-82907936 on Build GRCh38
6101297 c.728G>A p.R243K Chr.4: 82907932-82907932 on Build GRCh38
1431398 c.686A>G p.Y229C Chr.4: 82907890-82907890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs180735250 Chr.4: 82907833-82907833 on Build GRCh38
rs11731186 Chr.4: 82907858-82907858 on Build GRCh38
rs373288998 Chr.4: 82907906-82907906 on Build GRCh38
rs768702978 Chr.4: 82907981-82907981 on Build GRCh38
rs747122124 Chr.4: 82907790-82907790 on Build GRCh38
rs779371009 Chr.4: 82907815-82907815 on Build GRCh38
rs755411146 Chr.4: 82907752-82907752 on Build GRCh38
rs985259868 Chr.4: 82907943-82907943 on Build GRCh38
rs754146317 Chr.4: 82907751-82907751 on Build GRCh38
rs774384909 Chr.4: 82907987-82907987 on Build GRCh38
rs778446927 Chr.4: 82907729-82907729 on Build GRCh38
rs3214860 Chr.4: 82908085-82908085 on Build GRCh38
rs941425193 Chr.4: 82908180-82908180 on Build GRCh38
rs781253094 Chr.4: 82907942-82907942 on Build GRCh38
rs751112205 Chr.4: 82907734-82907734 on Build GRCh38
rs186181530 Chr.4: 82907719-82907719 on Build GRCh38
rs1018975963 Chr.4: 82907961-82907961 on Build GRCh38
rs925693510 Chr.4: 82908131-82908131 on Build GRCh38
rs547964009 Chr.4: 82907710-82907710 on Build GRCh38
rs928002733 Chr.4: 82908070-82908070 on Build GRCh38
rs988807705 Chr.4: 82908043-82908043 on Build GRCh38
rs535419993 Chr.4: 82907801-82907801 on Build GRCh38
rs754502176 Chr.4: 82907961-82907962 on Build GRCh38
rs112782823 Chr.4: 82907959-82907959 on Build GRCh38
rs911096071 Chr.4: 82907777-82907777 on Build GRCh38
rs533456083 Chr.4: 82907724-82907724 on Build GRCh38
rs184994232 Chr.4: 82907852-82907852 on Build GRCh38
rs1055347277 Chr.4: 82908182-82908182 on Build GRCh38
rs369267998 Chr.4: 82907802-82907802 on Build GRCh38
rs761606222 Chr.4: 82907931-82907932 on Build GRCh38
rs753060761 Chr.4: 82907879-82907879 on Build GRCh38
rs1042782087 Chr.4: 82907741-82907741 on Build GRCh38
rs753150626 Chr.4: 82907761-82907761 on Build GRCh38
rs774035313 Chr.4: 82907900-82907900 on Build GRCh38
rs751589696 Chr.4: 82907935-82907935 on Build GRCh38
rs994712805 Chr.4: 82907913-82907913 on Build GRCh38
rs761451561 Chr.4: 82907832-82907832 on Build GRCh38
rs765542775 Chr.4: 82907867-82907867 on Build GRCh38
rs895239525 Chr.4: 82908134-82908134 on Build GRCh38
rs751139697 Chr.4: 82907947-82907948 on Build GRCh38
rs775448767 Chr.4: 82907809-82907809 on Build GRCh38
rs373448726 Chr.4: 82907894-82907894 on Build GRCh38
rs772996379 Chr.4: 82908207-82908207 on Build GRCh38
rs1027720704 Chr.4: 82907914-82907914 on Build GRCh38
rs946874060 Chr.4: 82908102-82908102 on Build GRCh38
rs190161285 Chr.4: 82907749-82907749 on Build GRCh38
rs534086102 Chr.4: 82907723-82907723 on Build GRCh38
rs765063227 Chr.4: 82907936-82907937 on Build GRCh38
rs914195527 Chr.4: 82908063-82908063 on Build GRCh38
rs936991736 Chr.4: 82907753-82907753 on Build GRCh38
rs961581102 Chr.4: 82907721-82907721 on Build GRCh38
rs767213553 Chr.4: 82907750-82907750 on Build GRCh38
rs1029355017 Chr.4: 82907938-82907938 on Build GRCh38
rs937107976 Chr.4: 82908175-82908175 on Build GRCh38
rs112138852 Chr.4: 82907731-82907731 on Build GRCh38
rs771048858 Chr.4: 82907794-82907794 on Build GRCh38
rs769765098 Chr.4: 82907806-82907806 on Build GRCh38
rs543527091 Chr.4: 82908173-82908173 on Build GRCh38
rs1006135204 Chr.4: 82907897-82907897 on Build GRCh38
rs989961754 Chr.4: 82907792-82907792 on Build GRCh38
rs556714543 Chr.4: 82908061-82908061 on Build GRCh38
rs779928980 Chr.4: 82907974-82907974 on Build GRCh38
rs771853943 Chr.4: 82908115-82908115 on Build GRCh38
rs776766908 Chr.4: 82907922-82907925 on Build GRCh38
rs548963182 Chr.4: 82908028-82908030 on Build GRCh38
rs573595138 Chr.4: 82908216-82908216 on Build GRCh38
rs560448087 Chr.4: 82907944-82907946 on Build GRCh38
rs945992547 Chr.4: 82908038-82908038 on Build GRCh38
rs530708873 Chr.4: 82908109-82908109 on Build GRCh38
rs747859393 Chr.4: 82908011-82908011 on Build GRCh38
rs953470148 Chr.4: 82907916-82907916 on Build GRCh38
rs368968636 Chr.4: 82907737-82907737 on Build GRCh38
rs767284361 Chr.4: 82907974-82907975 on Build GRCh38
rs1055611781 Chr.4: 82907862-82907862 on Build GRCh38
rs777760610 Chr.4: 82907772-82907772 on Build GRCh38
rs749198187 Chr.4: 82907820-82907820 on Build GRCh38
rs757401092 Chr.4: 82907936-82907936 on Build GRCh38
rs373651819 Chr.4: 82907824-82907824 on Build GRCh38
rs765859219 Chr.4: 82907947-82907947 on Build GRCh38
rs758930726 Chr.4: 82907769-82907769 on Build GRCh38
rs749110742 Chr.4: 82907976-82907976 on Build GRCh38
rs141749914 Chr.4: 82907972-82907972 on Build GRCh38
rs939358343 Chr.4: 82908108-82908108 on Build GRCh38
rs897308239 Chr.4: 82908227-82908227 on Build GRCh38
rs150214667 Chr.4: 82907831-82907831 on Build GRCh38
rs533133126 Chr.4: 82907939-82907940 on Build GRCh38
rs768897545 Chr.4: 82907764-82907765 on Build GRCh38

View Full Product Details