Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147653921-147654413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP6
Entrez Gene ID
51205
Gene Name
acid phosphatase 6, lysophosphatidic
Gene Aliases
ACPL1, LPAP, PACPL1
UniGene
Hs.562154
Gene Chromosome Location
Chr.1: 147629658-147670543 on Build GRCh38
Transcripts
NM_001323625.1 NM_016361.4 NR_136633.1 NR_136634.1 NR_136635.1 NR_136636.1 XM_011509601.2 XM_017001422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4860256 c.679G>T p.G227* Chr.1: 147654295-147654295 on Build GRCh38
3735932 c.652C>T p.R218W Chr.1: 147654322-147654322 on Build GRCh38
3801690 c.704T>G p.V235G Chr.1: 147654270-147654270 on Build GRCh38
4658943 c.760G>A p.D254N Chr.1: 147654214-147654214 on Build GRCh38
5843488 c.718G>A p.G240S Chr.1: 147654256-147654256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782284142 Chr.1: 147654308-147654308 on Build GRCh38
rs373655744 Chr.1: 147654156-147654156 on Build GRCh38
rs1024668648 Chr.1: 147654320-147654320 on Build GRCh38
rs372223758 Chr.1: 147654246-147654246 on Build GRCh38
rs782702420 Chr.1: 147654209-147654209 on Build GRCh38
rs957944725 Chr.1: 147654351-147654351 on Build GRCh38
rs782463000 Chr.1: 147654321-147654321 on Build GRCh38
rs782552755 Chr.1: 147654208-147654208 on Build GRCh38
rs782402251 Chr.1: 147654168-147654168 on Build GRCh38
rs781810026 Chr.1: 147654214-147654214 on Build GRCh38
rs782753747 Chr.1: 147654200-147654200 on Build GRCh38
rs11541487 Chr.1: 147654302-147654302 on Build GRCh38
rs150452101 Chr.1: 147654360-147654360 on Build GRCh38
rs200795602 Chr.1: 147654167-147654167 on Build GRCh38
rs936626654 Chr.1: 147654054-147654054 on Build GRCh38
rs587593500 Chr.1: 147654044-147654044 on Build GRCh38
rs144323712 Chr.1: 147654233-147654233 on Build GRCh38
rs1013731482 Chr.1: 147654312-147654312 on Build GRCh38
rs782047529 Chr.1: 147654172-147654172 on Build GRCh38
rs782246421 Chr.1: 147654241-147654241 on Build GRCh38
rs782095829 Chr.1: 147654199-147654199 on Build GRCh38
rs782724105 Chr.1: 147654176-147654176 on Build GRCh38
rs966387099 Chr.1: 147654031-147654031 on Build GRCh38
rs201892511 Chr.1: 147654177-147654177 on Build GRCh38
rs587667357 Chr.1: 147654251-147654251 on Build GRCh38
rs1037474301 Chr.1: 147654227-147654227 on Build GRCh38
rs1051197879 Chr.1: 147654113-147654113 on Build GRCh38
rs932692690 Chr.1: 147654099-147654099 on Build GRCh38
rs781931066 Chr.1: 147654271-147654271 on Build GRCh38
rs139884200 Chr.1: 147654283-147654283 on Build GRCh38
rs898744174 Chr.1: 147654235-147654235 on Build GRCh38
rs374778975 Chr.1: 147654142-147654142 on Build GRCh38
rs1038845592 Chr.1: 147654062-147654062 on Build GRCh38
rs140258721 Chr.1: 147654205-147654205 on Build GRCh38
rs140566115 Chr.1: 147654322-147654322 on Build GRCh38
rs782068882 Chr.1: 147654328-147654328 on Build GRCh38
rs200278269 Chr.1: 147654141-147654141 on Build GRCh38
rs782211262 Chr.1: 147654291-147654291 on Build GRCh38
rs782776530 Chr.1: 147654362-147654362 on Build GRCh38
rs761876142 Chr.1: 147654108-147654108 on Build GRCh38
rs991884407 Chr.1: 147653962-147653962 on Build GRCh38
rs782360313 Chr.1: 147654158-147654158 on Build GRCh38
rs782731508 Chr.1: 147654086-147654086 on Build GRCh38
rs781992577 Chr.1: 147654165-147654165 on Build GRCh38
rs959081701 Chr.1: 147653941-147653941 on Build GRCh38
rs781895444 Chr.1: 147654371-147654371 on Build GRCh38
rs782473850 Chr.1: 147654229-147654229 on Build GRCh38
rs782214996 Chr.1: 147654147-147654147 on Build GRCh38
rs925218302 Chr.1: 147654049-147654049 on Build GRCh38
rs782043022 Chr.1: 147653968-147653968 on Build GRCh38
rs587742332 Chr.1: 147654265-147654265 on Build GRCh38
rs933120067 Chr.1: 147654387-147654387 on Build GRCh38
rs995723847 Chr.1: 147654240-147654240 on Build GRCh38
rs978191905 Chr.1: 147654035-147654035 on Build GRCh38
rs782289489 Chr.1: 147654269-147654269 on Build GRCh38
rs587671312 Chr.1: 147654083-147654083 on Build GRCh38
rs368780422 Chr.1: 147654186-147654186 on Build GRCh38
rs917559746 Chr.1: 147653966-147653966 on Build GRCh38

View Full Product Details