Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 226995620-226996217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A4
Entrez Gene ID
1286
Gene Name
collagen type IV alpha 4 chain
Gene Aliases
CA44
UniGene
Hs.591645
Gene Chromosome Location
Chr.2: 226970293-227164559 on Build GRCh38
Transcripts
XM_011510559.2 XM_011510560.2 XM_011510561.2 XM_011510562.2 XM_011510565.2 XM_011510566.2 XM_011510567.2 XM_011510569.2 XM_011510570.2 XM_017003298.1 XM_017003300.1 NM_001167608.1 NM_032276.3 XM_017005083.1 XM_017005084.1 XM_017005085.1 XM_017005086.1 XM_017005087.1 XM_017005088.1 XM_017005089.1 XM_017005090.1 XM_017005091.1 XM_017005092.1

Gene Symbol
RHBDD1
Entrez Gene ID
84236
Gene Name
rhomboid domain containing 1
Gene Aliases
RRP4
UniGene
Hs.471514
Gene Chromosome Location
Chr.2: 226800164-226999210 on Build GRCh38
Transcripts
XM_011510559.2 XM_011510560.2 XM_011510561.2 XM_011510562.2 XM_011510565.2 XM_011510566.2 XM_011510567.2 XM_011510569.2 XM_011510570.2 XM_017003298.1 XM_017003300.1 NM_001167608.1 NM_032276.3 XM_017005083.1 XM_017005084.1 XM_017005085.1 XM_017005086.1 XM_017005087.1 XM_017005088.1 XM_017005089.1 XM_017005090.1 XM_017005091.1 XM_017005092.1

SNP Detail

SNP ID Chromosome Location
rs143376654 Chr.2: 226995861-226995861 on Build GRCh38
rs967136603 Chr.2: 226996042-226996042 on Build GRCh38
rs572017762 Chr.2: 226996063-226996063 on Build GRCh38
rs892956584 Chr.2: 226996001-226996001 on Build GRCh38
rs1019610748 Chr.2: 226995665-226995665 on Build GRCh38
rs768924653 Chr.2: 226995907-226995907 on Build GRCh38
rs549576732 Chr.2: 226995923-226995923 on Build GRCh38
rs984856028 Chr.2: 226995782-226995782 on Build GRCh38
rs568598365 Chr.2: 226995801-226995801 on Build GRCh38
rs1049557263 Chr.2: 226996175-226996175 on Build GRCh38
rs112301740 Chr.2: 226996032-226996032 on Build GRCh38
rs952210586 Chr.2: 226995777-226995777 on Build GRCh38
rs118113269 Chr.2: 226996015-226996015 on Build GRCh38
rs527717109 Chr.2: 226995817-226995817 on Build GRCh38
rs762086366 Chr.2: 226995838-226995838 on Build GRCh38
rs78221368 Chr.2: 226996030-226996030 on Build GRCh38
rs1054177020 Chr.2: 226996096-226996096 on Build GRCh38
rs368890892 Chr.2: 226995759-226995759 on Build GRCh38
rs10933164 Chr.2: 226995955-226995955 on Build GRCh38
rs139391430 Chr.2: 226995818-226995818 on Build GRCh38
rs531552732 Chr.2: 226995776-226995776 on Build GRCh38
rs903004496 Chr.2: 226995864-226995864 on Build GRCh38
rs191871396 Chr.2: 226995954-226995954 on Build GRCh38
rs893227191 Chr.2: 226996158-226996158 on Build GRCh38
rs573679622 Chr.2: 226996006-226996006 on Build GRCh38
rs993192025 Chr.2: 226996157-226996157 on Build GRCh38
rs894749784 Chr.2: 226995843-226995844 on Build GRCh38
rs1032835075 Chr.2: 226995660-226995660 on Build GRCh38
rs1045771706 Chr.2: 226995916-226995916 on Build GRCh38
rs559674539 Chr.2: 226996062-226996062 on Build GRCh38
rs916133237 Chr.2: 226996129-226996129 on Build GRCh38
rs991660634 Chr.2: 226996048-226996048 on Build GRCh38
rs996595347 Chr.2: 226996161-226996161 on Build GRCh38
rs1054123914 Chr.2: 226995906-226995906 on Build GRCh38
rs566066387 Chr.2: 226995863-226995863 on Build GRCh38
rs147140400 Chr.2: 226996181-226996181 on Build GRCh38
rs924614863 Chr.2: 226996149-226996149 on Build GRCh38
rs371111949 Chr.2: 226996088-226996088 on Build GRCh38
rs539700979 Chr.2: 226995870-226995870 on Build GRCh38
rs966283335 Chr.2: 226995681-226995681 on Build GRCh38
rs188118173 Chr.2: 226995726-226995726 on Build GRCh38
rs939793796 Chr.2: 226995752-226995752 on Build GRCh38
rs780405895 Chr.2: 226995760-226995760 on Build GRCh38
rs943106140 Chr.2: 226996125-226996125 on Build GRCh38
rs531385317 Chr.2: 226996172-226996172 on Build GRCh38
rs1034178230 Chr.2: 226996009-226996009 on Build GRCh38
rs934685907 Chr.2: 226996152-226996152 on Build GRCh38
rs975281631 Chr.2: 226995676-226995676 on Build GRCh38
rs888763979 Chr.2: 226995647-226995647 on Build GRCh38
rs1028223712 Chr.2: 226995694-226995694 on Build GRCh38
rs955294226 Chr.2: 226996013-226996013 on Build GRCh38
rs76218537 Chr.2: 226995800-226995800 on Build GRCh38
rs957469514 Chr.2: 226995676-226995676 on Build GRCh38
rs575165573 Chr.2: 226995648-226995648 on Build GRCh38
rs972318697 Chr.2: 226995701-226995701 on Build GRCh38
rs993728605 Chr.2: 226995970-226995970 on Build GRCh38
rs113017295 Chr.2: 226996053-226996053 on Build GRCh38
rs181594968 Chr.2: 226996155-226996155 on Build GRCh38
rs11546330 Chr.2: 226996002-226996002 on Build GRCh38
rs73994326 Chr.2: 226996019-226996019 on Build GRCh38
rs910387328 Chr.2: 226996100-226996100 on Build GRCh38
rs986447723 Chr.2: 226995733-226995733 on Build GRCh38
rs901520899 Chr.2: 226995957-226995957 on Build GRCh38
rs926724327 Chr.2: 226995806-226995806 on Build GRCh38
rs917917554 Chr.2: 226995826-226995826 on Build GRCh38
rs935832874 Chr.2: 226996089-226996089 on Build GRCh38
rs930836612 Chr.2: 226995804-226995804 on Build GRCh38
rs973837649 Chr.2: 226996032-226996033 on Build GRCh38
rs955012124 Chr.2: 226995702-226995702 on Build GRCh38
rs552448785 Chr.2: 226995669-226995669 on Build GRCh38
rs908249468 Chr.2: 226995745-226995745 on Build GRCh38
rs183998589 Chr.2: 226996047-226996047 on Build GRCh38
rs1026590626 Chr.2: 226995991-226995991 on Build GRCh38
rs776243785 Chr.2: 226995924-226995924 on Build GRCh38
rs188393451 Chr.2: 226996139-226996139 on Build GRCh38
rs537060903 Chr.2: 226995944-226995944 on Build GRCh38
rs920756337 Chr.2: 226995803-226995803 on Build GRCh38

View Full Product Details